This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01081564_m1
See other HDAC9 GE Assays ›
Gene Symbol:
HDAC9
Gene Name:
histone deacetylase 9
Gene Aliases:
HD7, HD7b, HD9, HDAC, HDAC7, HDAC7B, HDAC9B, HDAC9FL, HDRP, MITR
Chromosome Location:
Chr.7: 18086942 - 19003745 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01081564_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HDAC9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001321868.1 NP_001308797.1 16 - 17 2508 127
NM_001321877.1 NP_001308806.1 15 - 16 2438 127
NM_001321897.1 NP_001308826.1 16 - 17 2335 127
NM_058176.2 NP_478056.1 15 - 16 2247 127
NM_178423.2 NP_848510.1 16 - 17 2392 127
NM_178425.3 NP_848512.1 15 - 16 2256 127
XM_011515629.2 XP_011513931.1 16 - 17 2350 127
XM_011515631.2 XP_011513933.1 17 - 18 2664 127
XM_011515634.2 XP_011513936.1 15 - 16 2218 127
XM_011515635.2 XP_011513937.1 17 - 18 3216 127
XM_011515636.2 XP_011513938.1 16 - 17 2937 127
XM_011515637.2 XP_011513939.1 16 - 17 2361 127
XM_011515638.2 XP_011513940.1 16 - 17 2931 127
XM_011515639.2 XP_011513941.1 16 - 17 3128 127
XM_011515641.2 XP_011513943.1 17 - 18 3213 127
XM_011515642.2 XP_011513944.1 15 - 16 3942 127
XM_011515644.2 XP_011513946.1 17 - 18 3188 127
XM_011515645.2 XP_011513947.1 15 - 16 3927 127
XM_011515646.2 XP_011513948.1 15 - 16 3918 127
XM_011515648.2 XP_011513950.1 16 - 17 2946 127
XM_011515649.2 XP_011513951.1 16 - 17 3150 127
XM_017012826.1 XP_016868315.1 16 - 17 2922 127
XM_017012827.1 XP_016868316.1 17 - 18 3005 127
XM_017012828.1 XP_016868317.1 15 - 16 2998 127
GenBank mRNA AJ459808.1 - 16 - 17 2356 127
AY032737.1 - 15 - 16 2206 127
AY032738.1 - 15 - 16 2206 127
BC111735.1 - 14 - 15 2082 127

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene HDAC9

Panther Classification:

Molecular Function -

histone modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
response to amphetamine
transcription, DNA-templated
inflammatory response
heart development
chromatin modification
histone deacetylation
neuron differentiation
B cell differentiation
cellular response to insulin stimulus
peptidyl-lysine deacetylation
B cell activation
negative regulation of transcription, DNA-templated
regulation of skeletal muscle fiber development
regulation of striated muscle cell differentiation
histone H3 deacetylation
histone H4 deacetylation
positive regulation of cell migration involved in sprouting angiogenesis
transcription corepressor activity
histone deacetylase activity
protein kinase C binding
protein binding
transcription factor binding
NAD-dependent histone deacetylase activity (H3-K14 specific)
protein deacetylase activity
histone deacetylase binding
metal ion binding
repressing transcription factor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline