This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01081577_m1
See other HDAC9 GE Assays ›
Gene Symbol:
HDAC9
Gene Name:
histone deacetylase 9
Gene Aliases:
HD7, HD7b, HD9, HDAC, HDAC7, HDAC7B, HDAC9B, HDAC9FL, HDRP, MITR
Chromosome Location:
Chr.7: 18086942 - 19003745 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01081577_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HDAC9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001204144.2 NP_001191073.1 5 - 6 752 91
NM_001204145.2 NP_001191074.1 3 - 4 444 91
NM_001204147.2 NP_001191076.1 2 - 3 410 91
NM_001321868.1 NP_001308797.1 4 - 5 692 91
NM_001321870.1 NP_001308799.1 4 - 5 692 91
NM_001321871.1 NP_001308800.1 4 - 5 692 91
NM_001321875.1 NP_001308804.1 3 - 4 613 91
NM_001321878.1 NP_001308807.1 3 - 4 613 91
NM_001321886.1 NP_001308815.1 4 - 5 672 91
NM_001321888.1 NP_001308817.1 3 - 4 444 91
NM_001321890.1 NP_001308819.1 3 - 4 613 91
NM_001321891.1 NP_001308820.1 3 - 4 613 91
NM_001321893.1 NP_001308822.1 4 - 5 510 91
NM_001321900.1 NP_001308829.1 3 - 4 613 91
NM_014707.3 NP_055522.1 3 - 4 444 91
NM_058176.2 NP_478056.1 2 - 3 299 91
NM_178423.2 NP_848510.1 3 - 4 444 91
XM_011515636.2 XP_011513938.1 3 - 4 974 91
XM_011515637.2 XP_011513939.1 3 - 4 398 91
XM_011515646.2 XP_011513948.1 2 - 3 1970 91
XM_011515651.1 XP_011513953.1 3 - 4 374 91
XM_017012826.1 XP_016868315.1 3 - 4 974 91
XM_017012830.1 XP_016868319.1 3 - 4 374 91
GenBank mRNA AB018287.1 - 3 - 4 408 91
AJ459808.1 - 3 - 4 408 91
AK297404.1 - 3 - 4 398 91
AK304298.1 - 5 - 6 745 91
AK304410.1 - 2 - 3 373 91
AY032737.1 - 2 - 3 258 91
AY032738.1 - 2 - 3 258 91
BC150328.1 - 3 - 4 419 8860495 91
BC152405.1 - 3 - 4 408 8862508 91

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0700006813.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene HDAC9

Panther Classification:

Molecular Function -

histone modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
response to amphetamine
transcription, DNA-templated
inflammatory response
heart development
chromatin modification
histone deacetylation
neuron differentiation
B cell differentiation
cellular response to insulin stimulus
peptidyl-lysine deacetylation
B cell activation
negative regulation of transcription, DNA-templated
regulation of skeletal muscle fiber development
regulation of striated muscle cell differentiation
histone H3 deacetylation
histone H4 deacetylation
positive regulation of cell migration involved in sprouting angiogenesis
transcription corepressor activity
histone deacetylase activity
protein kinase C binding
protein binding
transcription factor binding
NAD-dependent histone deacetylase activity (H3-K14 specific)
protein deacetylase activity
histone deacetylase binding
metal ion binding
repressing transcription factor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 ENST00000401921.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 ENST00000405010.7 Ensembl
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 ENST00000406072.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 ENST00000406451.8 Ensembl
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 ENST00000413380.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 ENST00000413509.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 ENST00000417496.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 ENST00000428307.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 ENST00000432645.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 ENST00000441542.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 ENST00000441986.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 ENST00000446646.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 ENST00000456174.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 ENST00000461159.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 ENST00000523867.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 ENST00000524023.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 ENST00000622668.4 Ensembl
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 NM_001204144.2 Refseq
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 NM_001204145.2 Refseq
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 NM_001204146.2 Refseq
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 NM_001204147.2 Refseq
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 NM_001204148.2 Refseq
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 NM_014707.3 Refseq
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 NM_058176.2 Refseq
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 NM_178423.2 Refseq
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 NM_178425.3 Refseq
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 ENST00000401921.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 ENST00000405010.7 Ensembl
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 ENST00000406072.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 ENST00000406451.8 Ensembl
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 ENST00000413380.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 ENST00000413509.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 ENST00000417496.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 ENST00000428307.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 ENST00000432645.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 ENST00000441542.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 ENST00000441986.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 ENST00000446646.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 ENST00000456174.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 ENST00000461159.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 ENST00000523867.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 ENST00000524023.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 ENST00000622668.4 Ensembl
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 NM_001204144.2 Refseq
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 NM_001204145.2 Refseq
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 NM_001204146.2 Refseq
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 NM_001204147.2 Refseq
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 NM_001204148.2 Refseq
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 NM_014707.3 Refseq
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 NM_058176.2 Refseq
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 NM_178423.2 Refseq
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 NM_178425.3 Refseq
Affymetrix Human Clariom D Assay PSR0700147083.hg.1 TC0700006813.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7gdsv:80/100.66.79.88:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0