This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01082235_m1
See other SRC GE Assays ›
Gene Symbol:
SRC
Gene Name:
SRC proto-oncogene, non-receptor tyrosine kinase
Gene Aliases:
ASV, SRC1, THC6, c-SRC, p60-Src
Chromosome Location:
Chr.20: 37344685 - 37405432 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01082235_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SRC
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005417.4 NP_005408.1 5 - 6 796 105
NM_198291.2 NP_938033.1 5 - 6 721 105
XM_011529013.2 XP_011527315.1 6 - 7 1065 105
GenBank mRNA BC011566.1 - 3 - 4 386 4548399 105
BC051270.1 - 3 - 4 449 4871614 105

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2000007283.hg.1
This assay targets the same location as HTA transcript cluster(s) TC20000273.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SRC

Gene Ontology Categories:

Function(s) Process(es)

stimulatory C-type lectin receptor signaling pathway
cell cycle
signal transduction
signal complex assembly
epidermal growth factor receptor signaling pathway
transforming growth factor beta receptor signaling pathway
integrin-mediated signaling pathway
central nervous system development
cell proliferation
response to mechanical stimulus
response to virus
response to acidic pH
regulation of epithelial cell migration
positive regulation of epithelial cell migration
positive regulation of platelet-derived growth factor receptor signaling pathway
positive regulation of glucose metabolic process
positive regulation of smooth muscle cell migration
single organismal cell-cell adhesion
peptidyl-serine phosphorylation
peptidyl-tyrosine phosphorylation
regulation of cell-cell adhesion
platelet activation
intracellular estrogen receptor signaling pathway
forebrain development
T cell costimulation
protein destabilization
response to nutrient levels
positive regulation of protein autophosphorylation
activation of protein kinase B activity
negative regulation of telomere maintenance via telomerase
negative regulation of protein homooligomerization
cellular response to insulin stimulus
regulation of intracellular estrogen receptor signaling pathway
positive regulation of integrin activation
adherens junction organization
substrate adhesion-dependent cell spreading
cellular response to reactive oxygen species
intracellular signal transduction
osteoclast development
cellular response to platelet-derived growth factor stimulus
peptidyl-tyrosine autophosphorylation
Fc-gamma receptor signaling pathway involved in phagocytosis
ERBB2 signaling pathway
regulation of cell proliferation
response to drug
response to hydrogen peroxide
positive regulation of apoptotic process
negative regulation of apoptotic process
regulation of vascular permeability
stress fiber assembly
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
regulation of protein binding
positive regulation of MAP kinase activity
positive regulation of phosphatidylinositol 3-kinase activity
transcytosis
innate immune response
regulation of bone resorption
bone resorption
positive regulation of cyclin-dependent protein serine/threonine kinase activity
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of insulin receptor signaling pathway
protein autophosphorylation
platelet-derived growth factor receptor signaling pathway
vascular endothelial growth factor receptor signaling pathway
neurotrophin TRK receptor signaling pathway
ephrin receptor signaling pathway
oogenesis
positive regulation of cytokine secretion
positive regulation of peptidyl-tyrosine phosphorylation
progesterone receptor signaling pathway
leukocyte migration
positive regulation of small GTPase mediated signal transduction
response to mineralocorticoid
response to electrical stimulus
regulation of cell cycle
negative regulation of focal adhesion assembly
positive regulation of protein kinase B signaling
negative regulation of mitochondrial depolarization
negative regulation of telomerase activity
uterus development
branching involved in mammary gland duct morphogenesis
regulation of cell projection assembly
positive regulation of ERK1 and ERK2 cascade
response to interleukin-1
cellular response to lipopolysaccharide
cellular response to peptide hormone stimulus
cellular response to progesterone stimulus
cellular response to fatty acid
cellular response to hypoxia
cellular response to fluid shear stress
regulation of podosome assembly
positive regulation of podosome assembly
positive regulation of protein serine/threonine kinase activity
angiotensin-activated signaling pathway involved in heart process
positive regulation of canonical Wnt signaling pathway
positive regulation of lamellipodium morphogenesis
positive regulation of DNA biosynthetic process
regulation of early endosome to late endosome transport
negative regulation of anoikis
negative regulation of extrinsic apoptotic signaling pathway
negative regulation of intrinsic apoptotic signaling pathway
regulation of caveolin-mediated endocytosis
protein kinase activity
protein tyrosine kinase activity
non-membrane spanning protein tyrosine kinase activity
SH3/SH2 adaptor activity
protein kinase C binding
receptor binding
insulin receptor binding
integrin binding
protein binding
ATP binding
protein C-terminus binding
kinase activity
enzyme binding
kinase binding
heme binding
estrogen receptor binding
SH2 domain binding
ion channel binding
ephrin receptor binding
phosphoprotein binding
hormone receptor binding
growth factor receptor binding
scaffold protein binding
cadherin binding involved in cell-cell adhesion

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR20003870.hg.1 ENST00000358208.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20003870.hg.1 ENST00000360723.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20003870.hg.1 ENST00000373558.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20003870.hg.1 ENST00000373567.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20003870.hg.1 ENST00000373578.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20003870.hg.1 ENST00000445403.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20003870.hg.1 NM_005417.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR20003870.hg.1 NM_198291.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR20003870.hg.1 TC20000273.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR2000150285.hg.1 ENST00000358208.8 Ensembl
Affymetrix Human Clariom D Assay PSR2000150285.hg.1 ENST00000373558.2 Ensembl
Affymetrix Human Clariom D Assay PSR2000150285.hg.1 ENST00000373567.6 Ensembl
Affymetrix Human Clariom D Assay PSR2000150285.hg.1 ENST00000373578.6 Ensembl
Affymetrix Human Clariom D Assay PSR2000150285.hg.1 ENST00000472968.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000150285.hg.1 ENST00000489153.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000150285.hg.1 NM_005417.4 Refseq
Affymetrix Human Clariom D Assay PSR2000150285.hg.1 NM_198291.2 Refseq
Affymetrix Human Clariom D Assay PSR2000150285.hg.1 TC2000007283.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2000150285.hg.1 ENST00000358208.8 Ensembl
Affymetrix Human Clariom D Assay PSR2000150285.hg.1 ENST00000373558.2 Ensembl
Affymetrix Human Clariom D Assay PSR2000150285.hg.1 ENST00000373567.6 Ensembl
Affymetrix Human Clariom D Assay PSR2000150285.hg.1 ENST00000373578.6 Ensembl
Affymetrix Human Clariom D Assay PSR2000150285.hg.1 ENST00000472968.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000150285.hg.1 ENST00000489153.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000150285.hg.1 NM_005417.4 Refseq
Affymetrix Human Clariom D Assay PSR2000150285.hg.1 NM_198291.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0