This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01083665_g1
See other P2RX5-TAX1BP3 GE Assays ›
Gene Symbol:
P2RX5-TAX1BP3 TAX1BP3
Gene Name:
P2RX5-TAX1BP3 readthrough (NMD candidate)
Tax1 binding protein 3
Gene Aliases:
-
TIP-1, TIP1
Chromosome Location:
Chr.17: 3662893 - 3696404 on Build GRCh38
Chromosome Location:
Chr.17: 3662893 - 3668679 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
-
Assay ID Hs01083665_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: P2RX5-TAX1BP3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_037928.1 13 - 14 5213 96
GenBank mRNA AB209623.1 - 13 - 14 5001 96
Gene Symbol: TAX1BP3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_014604.3 NP_055419.1 2 - 3 311 96
GenBank mRNA AF028823.2 - 2 - 3 314 96
AF234997.1 - 2 - 3 205 96
AF277318.1 - 2 - 3 184 96
AK315408.1 - 2 - 3 210 96
BC023980.1 - 2 - 3 214 4069168 96
U90913.1 - 2 - 3 192 96

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1700012336.hg.1, TC1700012338.hg.1
This assay targets the same location as HTA transcript cluster(s) TC17002884.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene P2RX5-TAX1BP3
Click here to find siRNAs for Gene TAX1BP3

Panther Classification:

Molecular Function -

ligand-gated ion channel

Gene Ontology Categories:

Function(s) Process(es)

Rho protein signal transduction
negative regulation of cell proliferation
Wnt signaling pathway
negative regulation of Wnt signaling pathway
activation of GTPase activity
negative regulation of protein localization to cell surface
protein binding
beta-catenin binding
protein C-terminus binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 ENST00000225525.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 ENST00000225525.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 ENST00000225525.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 ENST00000225525.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 ENST00000225525.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 ENST00000225525.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 ENST00000225525.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 ENST00000611779.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 ENST00000611779.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 ENST00000611779.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 ENST00000611779.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 ENST00000611779.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 ENST00000611779.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 NM_001204698.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 NM_001204698.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 NM_001204698.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 NM_001204698.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 NM_001204698.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 NM_001204698.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 NM_014604.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 NM_014604.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 NM_014604.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 NM_014604.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 NM_014604.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 NM_014604.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 NM_014604.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR17013475.hg.1 TC17002884.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR17013475.hg.1 ENST00000225525.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17013475.hg.1 NM_014604.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012338.hg.1 ENST00000550383.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012338.hg.1 ENST00000550383.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012338.hg.1 ENST00000550383.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012338.hg.1 ENST00000550383.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012338.hg.1 ENST00000550383.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012338.hg.1 ENST00000550383.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012338.hg.1 ENST00000550383.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012338.hg.1 ENST00000550383.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012338.hg.1 ENST00000550383.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012338.hg.1 ENST00000550383.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012338.hg.1 ENST00000550383.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012338.hg.1 ENST00000550383.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012338.hg.1 ENST00000550383.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012338.hg.1 ENST00000550383.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012338.hg.1 ENST00000550383.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012338.hg.1 NR_037928.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012338.hg.1 NR_037928.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012338.hg.1 NR_037928.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012338.hg.1 NR_037928.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012338.hg.1 NR_037928.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012338.hg.1 NR_037928.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012338.hg.1 NR_037928.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012338.hg.1 NR_037928.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012338.hg.1 NR_037928.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012338.hg.1 NR_037928.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012338.hg.1 NR_037928.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012338.hg.1 NR_037928.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012338.hg.1 NR_037928.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012338.hg.1 NR_037928.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012338.hg.1 NR_037928.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012338.hg.1 NR_037928.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012338.hg.1 NR_037928.1 Refseq
Affymetrix Human Clariom D Assay PSR1700204773.hg.1 ENST00000550383.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700204773.hg.1 NR_037928.1 Refseq
Affymetrix Human Clariom D Assay PSR1700204773.hg.1 ENST00000550383.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700204773.hg.1 NR_037928.1 Refseq
Affymetrix Human Clariom D Assay PSR1700204773.hg.1 TC1700012338.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1700012338.hg.1 ENST00000550383.1 Ensembl
Affymetrix Human Clariom D Assay TC1700012338.hg.1 ENST00000550383.1 Ensembl
Affymetrix Human Clariom D Assay TC1700012338.hg.1 ENST00000550383.1 Ensembl
Affymetrix Human Clariom D Assay TC1700012338.hg.1 ENST00000550383.1 Ensembl
Affymetrix Human Clariom D Assay TC1700012338.hg.1 ENST00000550383.1 Ensembl
Affymetrix Human Clariom D Assay TC1700012338.hg.1 ENST00000550383.1 Ensembl
Affymetrix Human Clariom D Assay TC1700012338.hg.1 ENST00000550383.1 Ensembl
Affymetrix Human Clariom D Assay TC1700012338.hg.1 ENST00000550383.1 Ensembl
Affymetrix Human Clariom D Assay TC1700012338.hg.1 ENST00000550383.1 Ensembl
Affymetrix Human Clariom D Assay TC1700012338.hg.1 ENST00000550383.1 Ensembl
Affymetrix Human Clariom D Assay TC1700012338.hg.1 ENST00000550383.1 Ensembl
Affymetrix Human Clariom D Assay TC1700012338.hg.1 ENST00000550383.1 Ensembl
Affymetrix Human Clariom D Assay TC1700012338.hg.1 ENST00000550383.1 Ensembl
Affymetrix Human Clariom D Assay TC1700012338.hg.1 ENST00000550383.1 Ensembl
Affymetrix Human Clariom D Assay TC1700012338.hg.1 ENST00000550383.1 Ensembl
Affymetrix Human Clariom D Assay TC1700012338.hg.1 NR_037928.1 Refseq
Affymetrix Human Clariom D Assay TC1700012338.hg.1 NR_037928.1 Refseq
Affymetrix Human Clariom D Assay TC1700012338.hg.1 NR_037928.1 Refseq
Affymetrix Human Clariom D Assay TC1700012338.hg.1 NR_037928.1 Refseq
Affymetrix Human Clariom D Assay TC1700012338.hg.1 NR_037928.1 Refseq
Affymetrix Human Clariom D Assay TC1700012338.hg.1 NR_037928.1 Refseq
Affymetrix Human Clariom D Assay TC1700012338.hg.1 NR_037928.1 Refseq
Affymetrix Human Clariom D Assay TC1700012338.hg.1 NR_037928.1 Refseq
Affymetrix Human Clariom D Assay TC1700012338.hg.1 NR_037928.1 Refseq
Affymetrix Human Clariom D Assay TC1700012338.hg.1 NR_037928.1 Refseq
Affymetrix Human Clariom D Assay TC1700012338.hg.1 NR_037928.1 Refseq
Affymetrix Human Clariom D Assay TC1700012338.hg.1 NR_037928.1 Refseq
Affymetrix Human Clariom D Assay TC1700012338.hg.1 NR_037928.1 Refseq
Affymetrix Human Clariom D Assay TC1700012338.hg.1 NR_037928.1 Refseq
Affymetrix Human Clariom D Assay TC1700012338.hg.1 NR_037928.1 Refseq
Affymetrix Human Clariom D Assay TC1700012338.hg.1 NR_037928.1 Refseq
Affymetrix Human Clariom D Assay TC1700012338.hg.1 NR_037928.1 Refseq
Affymetrix Human Clariom D Assay TC1700012336.hg.1 ENST00000225525.3 Ensembl
Affymetrix Human Clariom D Assay TC1700012336.hg.1 ENST00000225525.3 Ensembl
Affymetrix Human Clariom D Assay TC1700012336.hg.1 ENST00000225525.3 Ensembl
Affymetrix Human Clariom D Assay TC1700012336.hg.1 ENST00000225525.3 Ensembl
Affymetrix Human Clariom D Assay TC1700012336.hg.1 ENST00000225525.3 Ensembl
Affymetrix Human Clariom D Assay TC1700012336.hg.1 ENST00000225525.3 Ensembl
Affymetrix Human Clariom D Assay TC1700012336.hg.1 ENST00000225525.3 Ensembl
Affymetrix Human Clariom D Assay TC1700012336.hg.1 ENST00000611779.4 Ensembl
Affymetrix Human Clariom D Assay TC1700012336.hg.1 ENST00000611779.4 Ensembl
Affymetrix Human Clariom D Assay TC1700012336.hg.1 ENST00000611779.4 Ensembl
Affymetrix Human Clariom D Assay TC1700012336.hg.1 ENST00000611779.4 Ensembl
Affymetrix Human Clariom D Assay TC1700012336.hg.1 ENST00000611779.4 Ensembl
Affymetrix Human Clariom D Assay TC1700012336.hg.1 ENST00000611779.4 Ensembl
Affymetrix Human Clariom D Assay TC1700012336.hg.1 NM_001204698.1 Refseq
Affymetrix Human Clariom D Assay TC1700012336.hg.1 NM_001204698.1 Refseq
Affymetrix Human Clariom D Assay TC1700012336.hg.1 NM_001204698.1 Refseq
Affymetrix Human Clariom D Assay TC1700012336.hg.1 NM_001204698.1 Refseq
Affymetrix Human Clariom D Assay TC1700012336.hg.1 NM_001204698.1 Refseq
Affymetrix Human Clariom D Assay TC1700012336.hg.1 NM_001204698.1 Refseq
Affymetrix Human Clariom D Assay TC1700012336.hg.1 NM_014604.3 Refseq
Affymetrix Human Clariom D Assay TC1700012336.hg.1 NM_014604.3 Refseq
Affymetrix Human Clariom D Assay TC1700012336.hg.1 NM_014604.3 Refseq
Affymetrix Human Clariom D Assay TC1700012336.hg.1 NM_014604.3 Refseq
Affymetrix Human Clariom D Assay TC1700012336.hg.1 NM_014604.3 Refseq
Affymetrix Human Clariom D Assay TC1700012336.hg.1 NM_014604.3 Refseq
Affymetrix Human Clariom D Assay TC1700012336.hg.1 NM_014604.3 Refseq
Affymetrix Human Clariom D Assay PSR1700204728.hg.1 ENST00000225525.3 Ensembl
Affymetrix Human Clariom D Assay PSR1700204728.hg.1 NM_014604.3 Refseq
Affymetrix Human Clariom D Assay PSR1700204728.hg.1 ENST00000225525.3 Ensembl
Affymetrix Human Clariom D Assay PSR1700204728.hg.1 NM_014604.3 Refseq
Affymetrix Human Clariom D Assay PSR1700204728.hg.1 TC1700012336.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0