This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01083666_g1
See other P2RX5-TAX1BP3 GE Assays ›
Gene Symbol:
P2RX5-TAX1BP3 TAX1BP3
Gene Name:
P2RX5-TAX1BP3 readthrough (NMD candidate)
Tax1 binding protein 3
Gene Aliases:
-
TIP-1, TIP1
Chromosome Location:
Chr.17: 3662893 - 3696404 on Build GRCh38
Chromosome Location:
Chr.17: 3662893 - 3668679 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
-
Assay ID Hs01083666_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: P2RX5-TAX1BP3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_037928.1 14 - 15 5295 71
GenBank mRNA AB209623.1 - 14 - 15 5083 71
Gene Symbol: TAX1BP3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_014604.3 NP_055419.1 3 - 4 393 71
GenBank mRNA AF028823.2 - 3 - 4 396 71
AF234997.1 - 3 - 4 287 71
AF277318.1 - 3 - 4 266 71
AK315408.1 - 3 - 4 292 71
BC023980.1 - 3 - 4 296 4069168 71
U90913.1 - 3 - 4 274 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1700012336.hg.1, TC1700012338.hg.1
This assay targets the same location as HTA transcript cluster(s) TC17002884.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene P2RX5-TAX1BP3
Click here to find siRNAs for Gene TAX1BP3

Panther Classification:

Molecular Function -

ligand-gated ion channel

Gene Ontology Categories:

Function(s) Process(es)

Rho protein signal transduction
negative regulation of cell proliferation
Wnt signaling pathway
negative regulation of Wnt signaling pathway
activation of GTPase activity
negative regulation of protein localization to cell surface
protein binding
beta-catenin binding
protein C-terminus binding

MicroArray Probes:

Agilent Affymetrix ArrayStar ABI

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 795657 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_23_P38468 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03204067 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 ENST00000225525.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 ENST00000225525.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 ENST00000225525.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 ENST00000225525.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 ENST00000225525.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 ENST00000225525.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 ENST00000225525.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 ENST00000611779.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 ENST00000611779.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 ENST00000611779.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 ENST00000611779.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 ENST00000611779.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 ENST00000611779.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 NM_001204698.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 NM_001204698.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 NM_001204698.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 NM_001204698.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 NM_001204698.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 NM_001204698.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 NM_014604.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 NM_014604.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 NM_014604.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 NM_014604.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 NM_014604.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 NM_014604.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700012336.hg.1 NM_014604.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR17013473.hg.1 TC17002884.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR17013473.hg.1 ENST00000225525.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17013473.hg.1 NM_001204698.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR17013473.hg.1 NM_014604.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR17013471.hg.1 TC17002884.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR17013471.hg.1 ENST00000225525.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17013471.hg.1 NM_001204698.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR17013471.hg.1 NM_014604.3 Refseq
Affymetrix Human Clariom D Assay PSR1700204726.hg.1 ENST00000225525.3 Ensembl
Affymetrix Human Clariom D Assay PSR1700204726.hg.1 ENST00000611779.4 Ensembl
Affymetrix Human Clariom D Assay PSR1700204726.hg.1 NM_001204698.1 Refseq
Affymetrix Human Clariom D Assay PSR1700204726.hg.1 NM_014604.3 Refseq
Affymetrix Human Clariom D Assay PSR1700204726.hg.1 ENST00000225525.3 Ensembl
Affymetrix Human Clariom D Assay PSR1700204726.hg.1 ENST00000611779.4 Ensembl
Affymetrix Human Clariom D Assay PSR1700204726.hg.1 NM_001204698.1 Refseq
Affymetrix Human Clariom D Assay PSR1700204726.hg.1 NM_014604.3 Refseq
Affymetrix Human Clariom D Assay PSR1700204726.hg.1 TC1700012336.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1700204772.hg.1 ENST00000550383.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700204772.hg.1 NR_037928.1 Refseq
Affymetrix Human Clariom D Assay PSR1700204772.hg.1 ENST00000550383.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700204772.hg.1 NR_037928.1 Refseq
Affymetrix Human Clariom D Assay PSR1700204772.hg.1 TC1700012338.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1700204727.hg.1 ENST00000225525.3 Ensembl
Affymetrix Human Clariom D Assay PSR1700204727.hg.1 ENST00000611779.4 Ensembl
Affymetrix Human Clariom D Assay PSR1700204727.hg.1 NM_001204698.1 Refseq
Affymetrix Human Clariom D Assay PSR1700204727.hg.1 NM_014604.3 Refseq
Affymetrix Human Clariom D Assay PSR1700204727.hg.1 ENST00000225525.3 Ensembl
Affymetrix Human Clariom D Assay PSR1700204727.hg.1 ENST00000611779.4 Ensembl
Affymetrix Human Clariom D Assay PSR1700204727.hg.1 NM_001204698.1 Refseq
Affymetrix Human Clariom D Assay PSR1700204727.hg.1 NM_014604.3 Refseq
Affymetrix Human Clariom D Assay PSR1700204727.hg.1 TC1700012336.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1700012336.hg.1 ENST00000225525.3 Ensembl
Affymetrix Human Clariom D Assay TC1700012336.hg.1 ENST00000225525.3 Ensembl
Affymetrix Human Clariom D Assay TC1700012336.hg.1 ENST00000225525.3 Ensembl
Affymetrix Human Clariom D Assay TC1700012336.hg.1 ENST00000225525.3 Ensembl
Affymetrix Human Clariom D Assay TC1700012336.hg.1 ENST00000225525.3 Ensembl
Affymetrix Human Clariom D Assay TC1700012336.hg.1 ENST00000225525.3 Ensembl
Affymetrix Human Clariom D Assay TC1700012336.hg.1 ENST00000225525.3 Ensembl
Affymetrix Human Clariom D Assay TC1700012336.hg.1 ENST00000611779.4 Ensembl
Affymetrix Human Clariom D Assay TC1700012336.hg.1 ENST00000611779.4 Ensembl
Affymetrix Human Clariom D Assay TC1700012336.hg.1 ENST00000611779.4 Ensembl
Affymetrix Human Clariom D Assay TC1700012336.hg.1 ENST00000611779.4 Ensembl
Affymetrix Human Clariom D Assay TC1700012336.hg.1 ENST00000611779.4 Ensembl
Affymetrix Human Clariom D Assay TC1700012336.hg.1 ENST00000611779.4 Ensembl
Affymetrix Human Clariom D Assay TC1700012336.hg.1 NM_001204698.1 Refseq
Affymetrix Human Clariom D Assay TC1700012336.hg.1 NM_001204698.1 Refseq
Affymetrix Human Clariom D Assay TC1700012336.hg.1 NM_001204698.1 Refseq
Affymetrix Human Clariom D Assay TC1700012336.hg.1 NM_001204698.1 Refseq
Affymetrix Human Clariom D Assay TC1700012336.hg.1 NM_001204698.1 Refseq
Affymetrix Human Clariom D Assay TC1700012336.hg.1 NM_001204698.1 Refseq
Affymetrix Human Clariom D Assay TC1700012336.hg.1 NM_014604.3 Refseq
Affymetrix Human Clariom D Assay TC1700012336.hg.1 NM_014604.3 Refseq
Affymetrix Human Clariom D Assay TC1700012336.hg.1 NM_014604.3 Refseq
Affymetrix Human Clariom D Assay TC1700012336.hg.1 NM_014604.3 Refseq
Affymetrix Human Clariom D Assay TC1700012336.hg.1 NM_014604.3 Refseq
Affymetrix Human Clariom D Assay TC1700012336.hg.1 NM_014604.3 Refseq
Affymetrix Human Clariom D Assay TC1700012336.hg.1 NM_014604.3 Refseq
Affymetrix Human Clariom D Assay PSR1700204771.hg.1 NR_037928.1 Refseq
Affymetrix Human Clariom D Assay PSR1700204771.hg.1 NR_037928.1 Refseq
Affymetrix Human Clariom D Assay PSR1700204771.hg.1 TC1700012338.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
Arraystar Human LncRNA microarray V2.0 ASHG19A3A004043 - -
Arraystar Human LncRNA microarray V2.0 ASHG19A3A055020 - -
Arraystar Human LncRNA microarray V2.0 ASHG19A3A055020 - -
Arraystar Human LncRNA microarray V2.0 ASHG19A3A004043 - -
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 100058 - -
ABI Human Genome Survey Microarray Version 2.0 106378 - -
ABI Human Genome Survey Microarray Version 2.0 119665 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0