This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01083938_m1
See other LSAMP GE Assays ›
Gene Symbol:
LSAMP
Gene Name:
limbic system-associated membrane protein
Gene Aliases:
IGLON3, LAMP
Chromosome Location:
Chr.3: 115802363 - 116445542 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01083938_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LSAMP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001318915.1 NP_001305844.1 6 - 7 1424 79
XM_017006384.1 XP_016861873.1 5 - 6 646 79
GenBank mRNA AK299851.1 - 1239 79

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

immunoglobulin

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
nervous system development
locomotory exploration behavior
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000333617.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000333617.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000333617.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000333617.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000333617.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000333617.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000333617.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000333617.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000333617.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000333617.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000333617.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000333617.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000333617.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000333617.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000473171.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000473171.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000473171.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000473171.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000473171.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000473171.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000474851.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000474851.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000474851.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000474851.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000474851.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000475403.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000475403.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000475403.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000475403.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000475403.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000475403.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000475403.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000490035.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000490035.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000490035.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000490035.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000490035.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000490035.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000490035.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000490035.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000490035.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000490035.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000490035.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000490035.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000490035.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000490035.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000490035.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000490035.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000498645.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000498645.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000498645.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000498645.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000498645.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000498645.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000498645.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000539563.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000539563.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000539563.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000539563.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000539563.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000539563.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000539563.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000539563.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000539563.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000539563.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000539563.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 ENST00000539563.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 NM_002338.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 NM_002338.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 NM_002338.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 NM_002338.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 NM_002338.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 NM_002338.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 NM_002338.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 NM_002338.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 NM_002338.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 NM_002338.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 NM_002338.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 NM_002338.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 NM_002338.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 NM_002338.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 NM_002338.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012073.hg.1 NM_002338.4 Refseq
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000333617.8 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000333617.8 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000333617.8 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000333617.8 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000333617.8 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000333617.8 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000333617.8 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000333617.8 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000333617.8 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000333617.8 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000333617.8 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000333617.8 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000333617.8 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000333617.8 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000473171.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000473171.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000473171.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000473171.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000473171.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000473171.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000474851.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000474851.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000474851.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000474851.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000474851.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000475403.2 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000475403.2 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000475403.2 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000475403.2 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000475403.2 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000475403.2 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000475403.2 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000490035.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000490035.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000490035.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000490035.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000490035.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000490035.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000490035.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000490035.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000490035.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000490035.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000490035.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000490035.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000490035.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000490035.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000490035.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000490035.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000498645.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000498645.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000498645.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000498645.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000498645.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000498645.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000498645.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000539563.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000539563.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000539563.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000539563.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000539563.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000539563.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000539563.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000539563.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000539563.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000539563.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000539563.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 ENST00000539563.5 Ensembl
Affymetrix Human Clariom D Assay TC0300012073.hg.1 NM_002338.4 Refseq
Affymetrix Human Clariom D Assay TC0300012073.hg.1 NM_002338.4 Refseq
Affymetrix Human Clariom D Assay TC0300012073.hg.1 NM_002338.4 Refseq
Affymetrix Human Clariom D Assay TC0300012073.hg.1 NM_002338.4 Refseq
Affymetrix Human Clariom D Assay TC0300012073.hg.1 NM_002338.4 Refseq
Affymetrix Human Clariom D Assay TC0300012073.hg.1 NM_002338.4 Refseq
Affymetrix Human Clariom D Assay TC0300012073.hg.1 NM_002338.4 Refseq
Affymetrix Human Clariom D Assay TC0300012073.hg.1 NM_002338.4 Refseq
Affymetrix Human Clariom D Assay TC0300012073.hg.1 NM_002338.4 Refseq
Affymetrix Human Clariom D Assay TC0300012073.hg.1 NM_002338.4 Refseq
Affymetrix Human Clariom D Assay TC0300012073.hg.1 NM_002338.4 Refseq
Affymetrix Human Clariom D Assay TC0300012073.hg.1 NM_002338.4 Refseq
Affymetrix Human Clariom D Assay TC0300012073.hg.1 NM_002338.4 Refseq
Affymetrix Human Clariom D Assay TC0300012073.hg.1 NM_002338.4 Refseq
Affymetrix Human Clariom D Assay TC0300012073.hg.1 NM_002338.4 Refseq
Affymetrix Human Clariom D Assay TC0300012073.hg.1 NM_002338.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline