This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01084774_g1
See other UCP1 GE Assays ›
Gene Symbol:
UCP1
Gene Name:
uncoupling protein 1
Gene Aliases:
SLC25A7, UCP
Chromosome Location:
Chr.4: 140555770 - 140568914 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01084774_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UCP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_021833.4 NP_068605.1 3 - 4 601 117
XM_005263206.3 XP_005263263.1 3 - 4 707 117
XM_011532228.2 XP_011530530.1 3 - 4 710 117
GenBank mRNA BC069556.1 - 3 - 4 601 117
BC096736.1 - 3 - 4 598 40016003 117
BC098168.1 - 3 - 4 601 40016002 117
BC098258.1 - 3 - 4 601 40016001 117
BC098352.1 - 3 - 4 598 40016000 117
U28480.1 - 3 - 4 525 117

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0400011975.hg.1
This assay targets the same location as HTA transcript cluster(s) TC04001591.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene UCP1

Panther Classification:

Molecular Function -

transporter secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription from RNA polymerase II promoter
translation
mitochondrial transport
proton transport
brown fat cell differentiation
structural constituent of ribosome
oxidative phosphorylation uncoupler activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR04022192.hg.1 TC04001591.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR04022192.hg.1 ENST00000262999.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04022192.hg.1 NM_021833.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011975.hg.1 ENST00000262999.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011975.hg.1 ENST00000262999.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011975.hg.1 ENST00000262999.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011975.hg.1 ENST00000262999.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011975.hg.1 ENST00000262999.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011975.hg.1 ENST00000262999.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011975.hg.1 ENST00000262999.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011975.hg.1 ENST00000262999.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011975.hg.1 NM_021833.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011975.hg.1 NM_021833.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011975.hg.1 NM_021833.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011975.hg.1 NM_021833.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011975.hg.1 NM_021833.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011975.hg.1 NM_021833.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011975.hg.1 NM_021833.4 Refseq
Affymetrix Human Clariom D Assay PSR0400182151.hg.1 ENST00000262999.3 Ensembl
Affymetrix Human Clariom D Assay PSR0400182151.hg.1 NM_021833.4 Refseq
Affymetrix Human Clariom D Assay PSR0400182151.hg.1 ENST00000262999.3 Ensembl
Affymetrix Human Clariom D Assay PSR0400182151.hg.1 NM_021833.4 Refseq
Affymetrix Human Clariom D Assay PSR0400182151.hg.1 TC0400011975.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0400011975.hg.1 ENST00000262999.3 Ensembl
Affymetrix Human Clariom D Assay TC0400011975.hg.1 ENST00000262999.3 Ensembl
Affymetrix Human Clariom D Assay TC0400011975.hg.1 ENST00000262999.3 Ensembl
Affymetrix Human Clariom D Assay TC0400011975.hg.1 ENST00000262999.3 Ensembl
Affymetrix Human Clariom D Assay TC0400011975.hg.1 ENST00000262999.3 Ensembl
Affymetrix Human Clariom D Assay TC0400011975.hg.1 ENST00000262999.3 Ensembl
Affymetrix Human Clariom D Assay TC0400011975.hg.1 ENST00000262999.3 Ensembl
Affymetrix Human Clariom D Assay TC0400011975.hg.1 ENST00000262999.3 Ensembl
Affymetrix Human Clariom D Assay TC0400011975.hg.1 NM_021833.4 Refseq
Affymetrix Human Clariom D Assay TC0400011975.hg.1 NM_021833.4 Refseq
Affymetrix Human Clariom D Assay TC0400011975.hg.1 NM_021833.4 Refseq
Affymetrix Human Clariom D Assay TC0400011975.hg.1 NM_021833.4 Refseq
Affymetrix Human Clariom D Assay TC0400011975.hg.1 NM_021833.4 Refseq
Affymetrix Human Clariom D Assay TC0400011975.hg.1 NM_021833.4 Refseq
Affymetrix Human Clariom D Assay TC0400011975.hg.1 NM_021833.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vh84l:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0