This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01084794_m1
See other FREM3 GE Assays ›
Gene Symbol:
FREM3
Gene Name:
FRAS1 related extracellular matrix 3
Gene Aliases:
-
Chromosome Location:
Chr.4: 143577407 - 143700675 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01084794_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FREM3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001168235.1 NP_001161707.1 2 - 3 5277 87
GenBank mRNA AW953229.1 - 2 - 3 420 87

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0400012011.hg.1
This assay targets the same location as HTA transcript cluster(s) TC04001604.hg.1

Set Membership:

Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

cell communication
cell adhesion
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR04022364.hg.1 TC04001604.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR04022364.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04022364.hg.1 NM_001168235.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04022363.hg.1 TC04001604.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR04022363.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04022363.hg.1 NM_001168235.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012011.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012011.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012011.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012011.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012011.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012011.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012011.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012011.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012011.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012011.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012011.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012011.hg.1 ENST00000508899.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012011.hg.1 ENST00000508899.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400012011.hg.1 NM_001168235.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012011.hg.1 NM_001168235.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012011.hg.1 NM_001168235.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012011.hg.1 NM_001168235.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012011.hg.1 NM_001168235.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012011.hg.1 NM_001168235.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012011.hg.1 NM_001168235.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012011.hg.1 NM_001168235.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012011.hg.1 NM_001168235.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400012011.hg.1 NM_001168235.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04022362.hg.1 TC04001604.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR04022362.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04022362.hg.1 NM_001168235.1 Refseq
Affymetrix Human Clariom D Assay PSR0400182400.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400182400.hg.1 NM_001168235.1 Refseq
Affymetrix Human Clariom D Assay PSR0400182400.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400182400.hg.1 NM_001168235.1 Refseq
Affymetrix Human Clariom D Assay PSR0400182400.hg.1 TC0400012011.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0400012011.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012011.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012011.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012011.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012011.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012011.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012011.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012011.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012011.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012011.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012011.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Clariom D Assay TC0400012011.hg.1 ENST00000508899.1 Ensembl
Affymetrix Human Clariom D Assay TC0400012011.hg.1 ENST00000508899.1 Ensembl
Affymetrix Human Clariom D Assay TC0400012011.hg.1 NM_001168235.1 Refseq
Affymetrix Human Clariom D Assay TC0400012011.hg.1 NM_001168235.1 Refseq
Affymetrix Human Clariom D Assay TC0400012011.hg.1 NM_001168235.1 Refseq
Affymetrix Human Clariom D Assay TC0400012011.hg.1 NM_001168235.1 Refseq
Affymetrix Human Clariom D Assay TC0400012011.hg.1 NM_001168235.1 Refseq
Affymetrix Human Clariom D Assay TC0400012011.hg.1 NM_001168235.1 Refseq
Affymetrix Human Clariom D Assay TC0400012011.hg.1 NM_001168235.1 Refseq
Affymetrix Human Clariom D Assay TC0400012011.hg.1 NM_001168235.1 Refseq
Affymetrix Human Clariom D Assay TC0400012011.hg.1 NM_001168235.1 Refseq
Affymetrix Human Clariom D Assay TC0400012011.hg.1 NM_001168235.1 Refseq
Affymetrix Human Clariom D Assay PSR0400182402.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400182402.hg.1 NM_001168235.1 Refseq
Affymetrix Human Clariom D Assay PSR0400182402.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400182402.hg.1 NM_001168235.1 Refseq
Affymetrix Human Clariom D Assay PSR0400182402.hg.1 TC0400012011.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0400182401.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400182401.hg.1 NM_001168235.1 Refseq
Affymetrix Human Clariom D Assay PSR0400182401.hg.1 ENST00000329798.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400182401.hg.1 NM_001168235.1 Refseq
Affymetrix Human Clariom D Assay PSR0400182401.hg.1 TC0400012011.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-lk2hp:80/100.66.77.182:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0