This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01086077_g1
See other EML2 GE Assays ›
Gene Symbol:
EML2
Gene Name:
echinoderm microtubule associated protein like 2
Gene Aliases:
ELP70, EMAP-2, EMAP2
Chromosome Location:
Chr.19: 45609396 - 45645601 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01086077_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EML2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_012155.2 NP_036287.1 1 - 2 60 63
XM_017026550.1 XP_016882039.1 1 - 2 40 63
GenBank mRNA AF103939.1 - 1 - 2 54 63
AK225043.1 - 1 - 2 46 63
AK304299.1 - 1 - 2 44 63
AK308403.1 - 1 - 2 40 63
AL096717.1 - 1 - 2 32 63
BC032630.1 - 1 - 2 45 5402615 63
DC366776.1 - 1 - 2 60 63

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1900010947.hg.1
This assay targets the same location as HTA transcript cluster(s) TC19001625.hg.1

Additional Information:

This assay is in the 5'UTR region of the AF103939.1,AK304299.1,AL096717.1,BC032630.1,NM_012155.2,XM_017026550.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene EML2

Gene Ontology Categories:

Function(s) Process(es)

visual perception
sensory perception of sound
regulation of microtubule nucleation
negative regulation of microtubule polymerization
receptor binding
protein binding
microtubule binding
protein C-terminus binding
tubulin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR19022221.hg.1 ENST00000245925.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19022221.hg.1 NM_012155.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19022221.hg.1 NR_034098.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19022221.hg.1 TC19001625.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1900190993.hg.1 ENST00000245925.7 Ensembl
Affymetrix Human Clariom D Assay PSR1900190993.hg.1 ENST00000399594.7 Ensembl
Affymetrix Human Clariom D Assay PSR1900190993.hg.1 ENST00000536630.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900190993.hg.1 ENST00000586195.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900190993.hg.1 ENST00000586770.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900190993.hg.1 ENST00000587152.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900190993.hg.1 ENST00000588000.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900190993.hg.1 ENST00000588308.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900190993.hg.1 ENST00000588610.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900190993.hg.1 ENST00000588889.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900190993.hg.1 ENST00000589876.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900190993.hg.1 ENST00000590018.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900190993.hg.1 ENST00000590043.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900190993.hg.1 ENST00000591721.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900190993.hg.1 ENST00000593255.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900190993.hg.1 NM_001193268.2 Refseq
Affymetrix Human Clariom D Assay PSR1900190993.hg.1 NM_001193269.1 Refseq
Affymetrix Human Clariom D Assay PSR1900190993.hg.1 NM_012155.3 Refseq
Affymetrix Human Clariom D Assay PSR1900190993.hg.1 NR_034098.1 Refseq
Affymetrix Human Clariom D Assay PSR1900190993.hg.1 TC1900010947.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900190993.hg.1 ENST00000245925.7 Ensembl
Affymetrix Human Clariom D Assay PSR1900190993.hg.1 ENST00000399594.7 Ensembl
Affymetrix Human Clariom D Assay PSR1900190993.hg.1 ENST00000536630.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900190993.hg.1 ENST00000586195.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900190993.hg.1 ENST00000586770.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900190993.hg.1 ENST00000587152.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900190993.hg.1 ENST00000588000.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900190993.hg.1 ENST00000588308.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900190993.hg.1 ENST00000588610.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900190993.hg.1 ENST00000588889.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900190993.hg.1 ENST00000589876.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900190993.hg.1 ENST00000590018.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900190993.hg.1 ENST00000590043.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900190993.hg.1 ENST00000591721.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900190993.hg.1 ENST00000593255.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900190993.hg.1 NM_001193268.2 Refseq
Affymetrix Human Clariom D Assay PSR1900190993.hg.1 NM_001193269.1 Refseq
Affymetrix Human Clariom D Assay PSR1900190993.hg.1 NM_012155.3 Refseq
Affymetrix Human Clariom D Assay PSR1900190993.hg.1 NR_034098.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline