This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01086363_m1
See other CEACAM21 GE Assays ›
Gene Symbol:
CEACAM21
Gene Name:
carcinoembryonic antigen related cell adhesion molecule 21
Gene Aliases:
CEACAM3, R29124_1
Chromosome Location:
Chr.19: 41549518 - 41586844 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01086363_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CEACAM21
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001098506.3 NP_001091976.3 3 - 4 803 63
NM_001288773.2 NP_001275702.2 4 - 5 1136 63
XM_005278396.4 XP_005278453.1 3 - 4 1005 63
XM_011527450.1 XP_011525752.1 4 - 5 1174 63
XM_011527452.1 XP_011525754.1 4 - 5 1136 63
XM_017027430.1 XP_016882919.1 3 - 4 1377 63
XM_017027431.1 XP_016882920.1 3 - 4 1377 63
XM_017027433.1 XP_016882922.1 5 - 6 1250 63
GenBank mRNA AK023602.1 - 4 - 5 1136 63
BC106727.1 - 3 - 4 733 40015070 63

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1900011740.hg.1
This assay targets the same location as HTA transcript cluster(s) TC19000581.hg.1

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

immunoglobulin receptor superfamily

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR19008074.hg.1 ENST00000187608.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19008074.hg.1 ENST00000401445.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19008074.hg.1 ENST00000407170.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19008074.hg.1 ENST00000482870.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19008074.hg.1 NM_001098506.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19008074.hg.1 NM_033543.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19008074.hg.1 TC19000581.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1900201090.hg.1 ENST00000187608.13 Ensembl
Affymetrix Human Clariom D Assay PSR1900201090.hg.1 ENST00000401445.3 Ensembl
Affymetrix Human Clariom D Assay PSR1900201090.hg.1 ENST00000407170.6 Ensembl
Affymetrix Human Clariom D Assay PSR1900201090.hg.1 ENST00000457737.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900201090.hg.1 ENST00000482870.6 Ensembl
Affymetrix Human Clariom D Assay PSR1900201090.hg.1 ENST00000632983.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900201090.hg.1 NM_001288773.2 Refseq
Affymetrix Human Clariom D Assay PSR1900201090.hg.1 NM_001290113.1 Refseq
Affymetrix Human Clariom D Assay PSR1900201090.hg.1 NM_033543.5 Refseq
Affymetrix Human Clariom D Assay PSR1900201090.hg.1 TC1900011740.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900201090.hg.1 ENST00000187608.13 Ensembl
Affymetrix Human Clariom D Assay PSR1900201090.hg.1 ENST00000401445.3 Ensembl
Affymetrix Human Clariom D Assay PSR1900201090.hg.1 ENST00000407170.6 Ensembl
Affymetrix Human Clariom D Assay PSR1900201090.hg.1 ENST00000457737.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900201090.hg.1 ENST00000482870.6 Ensembl
Affymetrix Human Clariom D Assay PSR1900201090.hg.1 ENST00000632983.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900201090.hg.1 NM_001288773.2 Refseq
Affymetrix Human Clariom D Assay PSR1900201090.hg.1 NM_001290113.1 Refseq
Affymetrix Human Clariom D Assay PSR1900201090.hg.1 NM_033543.5 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0