This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01086471_m1
See other CHD5 GE Assays ›
Gene Symbol:
CHD5
Gene Name:
chromodomain helicase DNA binding protein 5
Gene Aliases:
CHD-5
Chromosome Location:
Chr.1: 6101787 - 6180134 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01086471_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CHD5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_015557.2 NP_056372.1 34 - 35 5115 146
GenBank mRNA AB007913.1 - 16 - 17 2076 146
AF425231.1 - 34 - 35 5104 146
AK126012.1 - 14 - 15 1907 146
AK127046.1 - 26 - 27 3722 146
AL117491.1 - 24 - 25 3308 146

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100012668.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01002144.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CHD5

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
negative regulation of cell proliferation
cerebral cortex neuron differentiation
spermatogenesis, exchange of chromosomal proteins
histone H4 acetylation
regulation of transcription involved in cell fate commitment
histone H3-K27 trimethylation
positive regulation of signal transduction by p53 class mediator
DNA binding
ATP binding
ATP-dependent helicase activity
zinc ion binding
H3K27me3 modified histone binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR01032991.hg.1 TC01002144.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01032991.hg.1 ENST00000262450.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01032991.hg.1 ENST00000377999.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01032991.hg.1 ENST00000378021.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01032991.hg.1 ENST00000462991.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01032991.hg.1 ENST00000536802.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01032991.hg.1 ENST00000538279.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01032991.hg.1 NM_015557.2 Refseq
Affymetrix Human Clariom D Assay PSR0100193741.hg.1 ENST00000262450.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100193741.hg.1 ENST00000377999.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100193741.hg.1 ENST00000462991.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100193741.hg.1 ENST00000496404.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100193741.hg.1 NM_015557.2 Refseq
Affymetrix Human Clariom D Assay PSR0100193741.hg.1 ENST00000262450.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100193741.hg.1 ENST00000377999.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100193741.hg.1 ENST00000462991.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100193741.hg.1 ENST00000496404.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100193741.hg.1 NM_015557.2 Refseq
Affymetrix Human Clariom D Assay PSR0100193741.hg.1 TC0100012668.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9hc65:80/100.66.78.230:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0