This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01087222_m1
See other FAT2 GE Assays ›
Gene Symbol:
FAT2
Gene Name:
FAT atypical cadherin 2
Gene Aliases:
CDHF8, CDHR9, HFAT2, MEGF1
Chromosome Location:
Chr.5: 151504092 - 151594822 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01087222_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FAT2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001447.2 NP_001438.1 10 - 11 9046 158
XM_006714761.3 XP_006714824.1 11 - 12 9617 158
XM_011537600.2 XP_011535902.1 13 - 14 9385 158
XM_011537603.2 XP_011535905.1 11 - 12 9318 158
XM_017009224.1 XP_016864713.1 13 - 14 9387 158
XM_017009225.1 XP_016864714.1 12 - 13 9229 158
GenBank mRNA AF231022.1 - 10 - 11 9046 158

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0500012514.hg.1
This assay targets the same location as HTA transcript cluster(s) TC05001954.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene FAT2

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

homophilic cell adhesion via plasma membrane adhesion molecules
epithelial cell migration
calcium ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 ENST00000520200.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 ENST00000520200.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 ENST00000520200.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 ENST00000520200.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 ENST00000520200.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 ENST00000520200.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 ENST00000520200.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 ENST00000520200.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 ENST00000520200.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 ENST00000520200.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 ENST00000520200.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 ENST00000520200.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05027214.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05027214.hg.1 NM_001447.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05027214.hg.1 TC05001954.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012514.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012514.hg.1 ENST00000520200.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012514.hg.1 ENST00000520200.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012514.hg.1 ENST00000520200.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012514.hg.1 ENST00000520200.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012514.hg.1 ENST00000520200.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012514.hg.1 ENST00000520200.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012514.hg.1 ENST00000520200.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012514.hg.1 ENST00000520200.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012514.hg.1 ENST00000520200.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012514.hg.1 ENST00000520200.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012514.hg.1 ENST00000520200.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012514.hg.1 ENST00000520200.5 Ensembl
Affymetrix Human Clariom D Assay TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Clariom D Assay TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Clariom D Assay TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Clariom D Assay TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Clariom D Assay TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Clariom D Assay TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Clariom D Assay TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Clariom D Assay TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Clariom D Assay TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Clariom D Assay TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Clariom D Assay TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Clariom D Assay TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Clariom D Assay TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Clariom D Assay TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Clariom D Assay TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Clariom D Assay TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Clariom D Assay TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Clariom D Assay TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Clariom D Assay TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Clariom D Assay TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Clariom D Assay TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Clariom D Assay TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Clariom D Assay TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Clariom D Assay TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Clariom D Assay TC0500012514.hg.1 NM_001447.2 Refseq
Affymetrix Human Clariom D Assay PSR0500187338.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500187338.hg.1 NM_001447.2 Refseq
Affymetrix Human Clariom D Assay PSR0500187338.hg.1 ENST00000261800.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500187338.hg.1 NM_001447.2 Refseq
Affymetrix Human Clariom D Assay PSR0500187338.hg.1 TC0500012514.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline