This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01090294_m1
See other VTI1A GE Assays ›
Gene Symbol:
VTI1A
Gene Name:
vesicle transport through interaction with t-SNAREs 1A
Gene Aliases:
MMDS3, MVti1, VTI1RP2, Vti1-rp2
Chromosome Location:
Chr.10: 112446988 - 112947204 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01090294_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: VTI1A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001318205.1 NP_001305134.1 4 - 5 729 64
NM_145206.3 NP_660207.2 4 - 5 729 64
XM_005269544.4 XP_005269601.1 4 - 5 683 64
XM_005269547.4 XP_005269604.1 4 - 5 683 64
XM_017015746.1 XP_016871235.1 4 - 5 683 64
XM_017015748.1 XP_016871237.1 4 - 5 683 64
GenBank mRNA AK303646.1 - 4 - 5 440 64
BC017052.1 - 4 - 5 459 3885564 64
BI010424.1 - 4 - 5 64
BI909044.1 - 4 - 5 487 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000008927.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10000814.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene VTI1A

Panther Classification:

Molecular Function -

SNARE protein

Gene Ontology Categories:

Function(s) Process(es)

protein targeting to vacuole
ER to Golgi vesicle-mediated transport
intra-Golgi vesicle-mediated transport
Golgi to vacuole transport
autophagy
retrograde transport, endosome to Golgi
vesicle fusion with Golgi apparatus
voluntary musculoskeletal movement
Golgi ribbon formation
SNARE binding
SNAP receptor activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR10009810.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10009810.hg.1 ENST00000432306.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10009810.hg.1 NM_145206.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10009810.hg.1 TC10000814.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR10009811.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10009811.hg.1 ENST00000432306.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10009811.hg.1 NM_145206.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10009811.hg.1 TC10000814.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000472892.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000472892.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000472892.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000472892.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000480057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000480057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000480057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000480057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000480057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000483122.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000483122.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000483122.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000483122.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000483122.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000483122.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000489142.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000489142.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000489142.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000489142.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000489142.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000489357.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000489357.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000489357.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000489357.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000489357.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000494728.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000494728.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000494728.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000494728.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000494728.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Clariom D Assay PSR1000161083.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Clariom D Assay PSR1000161083.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000161083.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000161083.hg.1 NM_145206.3 Refseq
Affymetrix Human Clariom D Assay PSR1000161083.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Clariom D Assay PSR1000161083.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000161083.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000161083.hg.1 NM_145206.3 Refseq
Affymetrix Human Clariom D Assay PSR1000161083.hg.1 TC1000008927.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000472892.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000472892.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000472892.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000472892.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000480057.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000480057.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000480057.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000480057.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000480057.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000483122.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000483122.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000483122.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000483122.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000483122.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000483122.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000489142.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000489142.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000489142.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000489142.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000489142.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000489357.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000489357.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000489357.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000489357.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000489357.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000494728.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000494728.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000494728.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000494728.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000494728.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Clariom D Assay TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Clariom D Assay TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Clariom D Assay TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Clariom D Assay TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Clariom D Assay TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Clariom D Assay TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Clariom D Assay TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Clariom D Assay TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Clariom D Assay TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Clariom D Assay TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Clariom D Assay TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Clariom D Assay PSR1000161082.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Clariom D Assay PSR1000161082.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000161082.hg.1 ENST00000489357.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000161082.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000161082.hg.1 NM_145206.3 Refseq
Affymetrix Human Clariom D Assay PSR1000161082.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Clariom D Assay PSR1000161082.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000161082.hg.1 ENST00000489357.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000161082.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000161082.hg.1 NM_145206.3 Refseq
Affymetrix Human Clariom D Assay PSR1000161082.hg.1 TC1000008927.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0