This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01090296_m1
See other VTI1A GE Assays ›
Gene Symbol:
VTI1A
Gene Name:
vesicle transport through interaction with t-SNAREs 1A
Gene Aliases:
MMDS3, MVti1, VTI1RP2, Vti1-rp2
Chromosome Location:
Chr.10: 112446988 - 112947204 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01090296_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: VTI1A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001318203.1 NP_001305132.1 7 - 8 902 113
NM_001318205.1 NP_001305134.1 6 - 7 881 113
NM_145206.3 NP_660207.2 6 - 7 881 113
XM_005269544.4 XP_005269601.1 6 - 7 835 113
XM_005269545.4 XP_005269602.1 7 - 8 902 113
XM_005269546.3 XP_005269603.1 7 - 8 902 113
XM_005269547.4 XP_005269604.1 6 - 7 835 113
XM_006717637.2 XP_006717700.1 7 - 8 902 113
XM_011539328.2 XP_011537630.1 7 - 8 902 113
XM_011539330.2 XP_011537632.1 7 - 8 902 113
XM_011539331.2 XP_011537633.1 7 - 8 902 113
XM_017015745.1 XP_016871234.1 7 - 8 902 113
XM_017015746.1 XP_016871235.1 6 - 7 835 113
XM_017015747.1 XP_016871236.1 7 - 8 902 113
XM_017015748.1 XP_016871237.1 6 - 7 835 113
GenBank mRNA AI610960.1 - 2 - 3 167 113
AI693064.1 - 2 - 3 113
AK303646.1 - 6 - 7 592 113
BC017052.1 - 6 - 7 611 3885564 113
BI010424.1 - 2 - 3 113

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000008927.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10000814.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene VTI1A

Panther Classification:

Molecular Function -

SNARE protein

Gene Ontology Categories:

Function(s) Process(es)

protein targeting to vacuole
ER to Golgi vesicle-mediated transport
intra-Golgi vesicle-mediated transport
Golgi to vacuole transport
autophagy
retrograde transport, endosome to Golgi
vesicle fusion with Golgi apparatus
voluntary musculoskeletal movement
Golgi ribbon formation
SNARE binding
SNAP receptor activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR10009813.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10009813.hg.1 ENST00000432306.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10009813.hg.1 NM_145206.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10009813.hg.1 TC10000814.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000472892.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000472892.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000472892.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000472892.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000480057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000480057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000480057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000480057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000480057.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000483122.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000483122.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000483122.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000483122.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000483122.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000483122.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000489142.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000489142.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000489142.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000489142.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000489142.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000489357.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000489357.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000489357.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000489357.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000489357.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000494728.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000494728.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000494728.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000494728.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000494728.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10009814.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10009814.hg.1 ENST00000432306.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10009814.hg.1 NM_145206.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10009814.hg.1 TC10000814.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1000161085.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Clariom D Assay PSR1000161085.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000161085.hg.1 NM_145206.3 Refseq
Affymetrix Human Clariom D Assay PSR1000161085.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Clariom D Assay PSR1000161085.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000161085.hg.1 NM_145206.3 Refseq
Affymetrix Human Clariom D Assay PSR1000161085.hg.1 TC1000008927.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1000161086.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Clariom D Assay PSR1000161086.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000161086.hg.1 NM_145206.3 Refseq
Affymetrix Human Clariom D Assay PSR1000161086.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Clariom D Assay PSR1000161086.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000161086.hg.1 NM_145206.3 Refseq
Affymetrix Human Clariom D Assay PSR1000161086.hg.1 TC1000008927.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1000161087.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000161087.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000161087.hg.1 TC1000008927.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000393077.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000432306.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000472892.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000472892.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000472892.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000472892.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000480057.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000480057.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000480057.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000480057.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000480057.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000483122.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000483122.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000483122.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000483122.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000483122.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000483122.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000489142.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000489142.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000489142.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000489142.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000489142.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000489357.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000489357.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000489357.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000489357.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000489357.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000494728.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000494728.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000494728.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000494728.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000494728.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 ENST00000496445.5 Ensembl
Affymetrix Human Clariom D Assay TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Clariom D Assay TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Clariom D Assay TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Clariom D Assay TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Clariom D Assay TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Clariom D Assay TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Clariom D Assay TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Clariom D Assay TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Clariom D Assay TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Clariom D Assay TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Clariom D Assay TC1000008927.hg.1 NM_145206.3 Refseq
Affymetrix Human Clariom D Assay TC1000008927.hg.1 NM_145206.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0