This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01090837_m1
See other CPSF3L GE Assays ›
Gene Symbol:
CPSF3L
Gene Name:
cleavage and polyadenylation specific factor 3-like
Gene Aliases:
CPSF73L, INT11, INTS11, RC-68, RC68
Chromosome Location:
Chr.1: 1311585 - 1324687 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01090837_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CPSF3L
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001256463.1 NP_001243392.1 2 - 3 204 66
GenBank mRNA AK021939.1 - 2 - 3 170 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100012465.hg.1

Set Membership:

Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

RNA processing factor

Gene Ontology Categories:

Function(s) Process(es)

snRNA processing
snRNA transcription from RNA polymerase II promoter
protein binding
hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000323275.10 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000411962.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000419704.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000421495.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000430786.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000435064.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000458452.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000462432.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000467408.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000488042.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000525603.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000526113.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000526332.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000526797.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000526904.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000527098.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000527383.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000527719.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000528879.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000530031.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000531377.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000533916.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000540437.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000545578.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000620829.4 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 NM_001256456.1 Refseq
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 NM_001256460.1 Refseq
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 NM_001256462.1 Refseq
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 NM_001256463.1 Refseq
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 NM_017871.5 Refseq
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000323275.10 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000411962.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000419704.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000421495.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000430786.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000435064.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000458452.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000462432.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000467408.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000488042.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000525603.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000526113.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000526332.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000526797.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000526904.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000527098.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000527383.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000527719.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000528879.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000530031.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000531377.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000533916.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000540437.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000545578.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 ENST00000620829.4 Ensembl
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 NM_001256456.1 Refseq
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 NM_001256460.1 Refseq
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 NM_001256462.1 Refseq
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 NM_001256463.1 Refseq
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 NM_017871.5 Refseq
Affymetrix Human Clariom D Assay PSR0100191751.hg.1 TC0100012465.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000323275.10 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000419704.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000429572.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000430786.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000434694.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000435064.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000450926.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000458452.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000470679.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000488042.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000490853.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000493534.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000496353.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000498173.2 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000498476.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000525285.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000526797.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000527098.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000527719.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000528879.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000530031.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000531019.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000532772.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000532952.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000534345.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000540437.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000545578.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000618806.4 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000627245.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 NM_001256456.1 Refseq
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 NM_001256460.1 Refseq
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 NM_001256463.1 Refseq
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 NM_017871.5 Refseq
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000323275.10 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000419704.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000429572.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000430786.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000434694.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000435064.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000450926.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000458452.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000470679.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000488042.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000490853.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000493534.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000496353.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000498173.2 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000498476.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000525285.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000526797.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000527098.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000527719.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000528879.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000530031.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000531019.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000532772.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000532952.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000534345.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000540437.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000545578.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000618806.4 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 ENST00000627245.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 NM_001256456.1 Refseq
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 NM_001256460.1 Refseq
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 NM_001256463.1 Refseq
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 NM_017871.5 Refseq
Affymetrix Human Clariom D Assay PSR0100191784.hg.1 TC0100012465.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-grw2s:80/100.66.79.65:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline