This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01091099_m1
See other GALE GE Assays ›
Gene Symbol:
GALE
Gene Name:
UDP-galactose-4-epimerase
Gene Aliases:
SDR1E1
Chromosome Location:
Chr.1: 23795599 - 23800804 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01091099_m1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GALE
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001008216.1 NP_001008217.1 1 - 2 90 79
GenBank mRNA AK057302.1 - 1 - 2 68 79
AK307354.1 - 1 - 2 40 79

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK057302.1, NM_001008216.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GALE

Panther Classification:

Molecular Function -

epimerase/racemase

Gene Ontology Categories:

Function(s) Process(es)

galactose catabolic process
UDP-N-acetylglucosamine 4-epimerase activity
UDP-glucose 4-epimerase activity
protein homodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR01036251.hg.1 TC01002350.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01036251.hg.1 NM_001008216.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01036246.hg.1 TC01002350.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01036246.hg.1 NM_001008216.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01036247.hg.1 TC01002350.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01036247.hg.1 NM_001008216.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01036249.hg.1 TC01002350.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01036249.hg.1 NM_001008216.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01036250.hg.1 TC01002350.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01036250.hg.1 NM_001008216.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01036248.hg.1 TC01002350.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01036248.hg.1 NM_001008216.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0