This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01091675_g1
See other GSTT1 GE Assays ›
Gene Symbol:
GSTT1
Gene Name:
glutathione S-transferase theta 1
Gene Aliases:
-
Chromosome Location:
Chr.22_KI270879v1: 270308 - 278486 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01091675_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GSTT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000853.3 NP_000844.2 4 - 5 612 90
NM_001293807.1 NP_001280736.1 4 - 5 570 90
NM_001293808.1 NP_001280737.1 3 - 4 463 90
NM_001293809.1 NP_001280738.1 2 - 3 373 90
NM_001293810.1 NP_001280739.1 3 - 4 524 90
NM_001293811.1 NP_001280740.1 3 - 4 461 90
NM_001293812.1 NP_001280741.1 4 - 5 551 90
GenBank mRNA AF435971.1 - 4 - 5 532 90
AJ867757.1 - 2 - 3 316 90
AJ874719.1 - 3 - 4 404 90
BC007065.1 - 4 - 5 572 3995846 90
BF343733.1 - 3 - 4 380 90
BG536639.1 - 4 - 5 504 90
BT019951.1 - 4 - 5 532 90
BX401239.2 - 4 - 5 499 90
CA313466.1 - 1 - 2 90
CD701199.1 - 2 - 3 358 90
CR456499.1 - 3 - 4 455 90
X79389.1 - 4 - 5 532 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC22000583.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GSTT1

Gene Ontology Categories:

Function(s) Process(es)

glutathione metabolic process
cellular oxidant detoxification
glutathione derivative biosynthetic process
glutathione transferase activity
glutathione peroxidase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR22010282.hg.1 ENST00000248935.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22010282.hg.1 ENST00000382792.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22010282.hg.1 ENST00000439996.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22010282.hg.1 ENST00000452369.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22010282.hg.1 NM_000853.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR22010282.hg.1 TC22000583.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR22010283.hg.1 ENST00000248935.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22010283.hg.1 ENST00000382792.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22010283.hg.1 ENST00000439996.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22010283.hg.1 NM_000853.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR22010283.hg.1 TC22000583.hg.1 HTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rr9bx:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0