This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01091755_g1
See other EIF4H GE Assays ›
Gene Symbol:
EIF4H
Gene Name:
eukaryotic translation initiation factor 4H
Gene Aliases:
WBSCR1, WSCR1, eIF-4H
Chromosome Location:
Chr.7: 74174376 - 74197101 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01091755_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EIF4H
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_022170.1 NP_071496.1 1 - 2 71 75
NM_031992.1 NP_114381.1 1 - 2 71 75
GenBank mRNA AK290676.1 - 1 - 2 91 75
AK297651.1 - 1 - 2 116 75
AK314749.1 - 1 - 2 173 75
BC010021.1 - 1 69 2960402 75
BC021214.2 - 1 - 2 69 4139578 75
BC066928.1 - 1 88 5263716 75
D26068.1 - 1 - 2 71 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0700007998.hg.1
This assay targets the same location as HTA transcript cluster(s) TC07000451.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene EIF4H

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

formation of translation preinitiation complex
cytoplasmic translation
translational initiation
regulation of translational initiation
viral process
sexual reproduction
developmental growth
eukaryotic translation initiation factor 4F complex assembly
cell-cell adhesion
nucleotide binding
RNA binding
translation initiation factor activity
helicase activity
protein binding
translation factor activity, RNA binding
RNA strand annealing activity
RNA strand-exchange activity
ribosomal small subunit binding
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 ENST00000265753.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 ENST00000265753.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 ENST00000265753.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 ENST00000265753.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 ENST00000265753.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 ENST00000265753.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 ENST00000265753.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 ENST00000265753.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 ENST00000265753.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 ENST00000265753.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 ENST00000265753.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 ENST00000353999.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 ENST00000353999.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 ENST00000353999.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 ENST00000353999.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 ENST00000353999.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 ENST00000353999.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 ENST00000353999.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 ENST00000353999.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 ENST00000479338.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 ENST00000479338.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 ENST00000479338.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 ENST00000479338.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 ENST00000479338.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 ENST00000479338.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 ENST00000489484.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 ENST00000489484.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 ENST00000489484.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 ENST00000489484.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 ENST00000489484.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 ENST00000489484.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 ENST00000489484.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 ENST00000489484.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 ENST00000495187.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 ENST00000495187.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 ENST00000495187.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 ENST00000495187.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 ENST00000495187.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 ENST00000495187.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 NM_022170.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 NM_022170.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 NM_022170.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 NM_022170.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 NM_022170.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 NM_022170.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 NM_022170.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 NM_022170.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 NM_022170.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 NM_031992.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 NM_031992.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 NM_031992.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 NM_031992.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 NM_031992.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 NM_031992.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 NM_031992.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007998.hg.1 NM_031992.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007075.hg.1 ENST00000424380.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700010578.hg.1 ENST00000453115.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07006104.hg.1 TC07000451.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR07006104.hg.1 ENST00000265753.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07006104.hg.1 ENST00000353999.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07006104.hg.1 ENST00000495187.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07006104.hg.1 NM_022170.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07006104.hg.1 NM_031992.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07006102.hg.1 TC07000451.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR07006102.hg.1 ENST00000265753.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07006102.hg.1 ENST00000353999.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07006102.hg.1 NM_022170.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07006102.hg.1 NM_031992.1 Refseq
Affymetrix Human Clariom D Assay PSR0700155234.hg.1 ENST00000265753.12 Ensembl
Affymetrix Human Clariom D Assay PSR0700155234.hg.1 ENST00000353999.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700155234.hg.1 ENST00000479338.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700155234.hg.1 ENST00000489484.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700155234.hg.1 NM_022170.1 Refseq
Affymetrix Human Clariom D Assay PSR0700155234.hg.1 NM_031992.1 Refseq
Affymetrix Human Clariom D Assay PSR0700155234.hg.1 ENST00000265753.12 Ensembl
Affymetrix Human Clariom D Assay PSR0700155234.hg.1 ENST00000353999.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700155234.hg.1 ENST00000479338.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700155234.hg.1 ENST00000489484.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700155234.hg.1 NM_022170.1 Refseq
Affymetrix Human Clariom D Assay PSR0700155234.hg.1 NM_031992.1 Refseq
Affymetrix Human Clariom D Assay PSR0700155234.hg.1 TC0700007998.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0700010578.hg.1 ENST00000453115.1 Ensembl
Affymetrix Human Clariom D Assay TC2200007075.hg.1 ENST00000424380.1 Ensembl
Affymetrix Human Clariom D Assay TC0700007998.hg.1 ENST00000265753.12 Ensembl
Affymetrix Human Clariom D Assay TC0700007998.hg.1 ENST00000265753.12 Ensembl
Affymetrix Human Clariom D Assay TC0700007998.hg.1 ENST00000265753.12 Ensembl
Affymetrix Human Clariom D Assay TC0700007998.hg.1 ENST00000265753.12 Ensembl
Affymetrix Human Clariom D Assay TC0700007998.hg.1 ENST00000265753.12 Ensembl
Affymetrix Human Clariom D Assay TC0700007998.hg.1 ENST00000265753.12 Ensembl
Affymetrix Human Clariom D Assay TC0700007998.hg.1 ENST00000265753.12 Ensembl
Affymetrix Human Clariom D Assay TC0700007998.hg.1 ENST00000265753.12 Ensembl
Affymetrix Human Clariom D Assay TC0700007998.hg.1 ENST00000265753.12 Ensembl
Affymetrix Human Clariom D Assay TC0700007998.hg.1 ENST00000265753.12 Ensembl
Affymetrix Human Clariom D Assay TC0700007998.hg.1 ENST00000265753.12 Ensembl
Affymetrix Human Clariom D Assay TC0700007998.hg.1 ENST00000353999.6 Ensembl
Affymetrix Human Clariom D Assay TC0700007998.hg.1 ENST00000353999.6 Ensembl
Affymetrix Human Clariom D Assay TC0700007998.hg.1 ENST00000353999.6 Ensembl
Affymetrix Human Clariom D Assay TC0700007998.hg.1 ENST00000353999.6 Ensembl
Affymetrix Human Clariom D Assay TC0700007998.hg.1 ENST00000353999.6 Ensembl
Affymetrix Human Clariom D Assay TC0700007998.hg.1 ENST00000353999.6 Ensembl
Affymetrix Human Clariom D Assay TC0700007998.hg.1 ENST00000353999.6 Ensembl
Affymetrix Human Clariom D Assay TC0700007998.hg.1 ENST00000353999.6 Ensembl
Affymetrix Human Clariom D Assay TC0700007998.hg.1 ENST00000479338.5 Ensembl
Affymetrix Human Clariom D Assay TC0700007998.hg.1 ENST00000479338.5 Ensembl
Affymetrix Human Clariom D Assay TC0700007998.hg.1 ENST00000479338.5 Ensembl
Affymetrix Human Clariom D Assay TC0700007998.hg.1 ENST00000479338.5 Ensembl
Affymetrix Human Clariom D Assay TC0700007998.hg.1 ENST00000479338.5 Ensembl
Affymetrix Human Clariom D Assay TC0700007998.hg.1 ENST00000479338.5 Ensembl
Affymetrix Human Clariom D Assay TC0700007998.hg.1 ENST00000489484.5 Ensembl
Affymetrix Human Clariom D Assay TC0700007998.hg.1 ENST00000489484.5 Ensembl
Affymetrix Human Clariom D Assay TC0700007998.hg.1 ENST00000489484.5 Ensembl
Affymetrix Human Clariom D Assay TC0700007998.hg.1 ENST00000489484.5 Ensembl
Affymetrix Human Clariom D Assay TC0700007998.hg.1 ENST00000489484.5 Ensembl
Affymetrix Human Clariom D Assay TC0700007998.hg.1 ENST00000489484.5 Ensembl
Affymetrix Human Clariom D Assay TC0700007998.hg.1 ENST00000489484.5 Ensembl
Affymetrix Human Clariom D Assay TC0700007998.hg.1 ENST00000489484.5 Ensembl
Affymetrix Human Clariom D Assay TC0700007998.hg.1 ENST00000495187.1 Ensembl
Affymetrix Human Clariom D Assay TC0700007998.hg.1 ENST00000495187.1 Ensembl
Affymetrix Human Clariom D Assay TC0700007998.hg.1 ENST00000495187.1 Ensembl
Affymetrix Human Clariom D Assay TC0700007998.hg.1 ENST00000495187.1 Ensembl
Affymetrix Human Clariom D Assay TC0700007998.hg.1 ENST00000495187.1 Ensembl
Affymetrix Human Clariom D Assay TC0700007998.hg.1 ENST00000495187.1 Ensembl
Affymetrix Human Clariom D Assay TC0700007998.hg.1 NM_022170.1 Refseq
Affymetrix Human Clariom D Assay TC0700007998.hg.1 NM_022170.1 Refseq
Affymetrix Human Clariom D Assay TC0700007998.hg.1 NM_022170.1 Refseq
Affymetrix Human Clariom D Assay TC0700007998.hg.1 NM_022170.1 Refseq
Affymetrix Human Clariom D Assay TC0700007998.hg.1 NM_022170.1 Refseq
Affymetrix Human Clariom D Assay TC0700007998.hg.1 NM_022170.1 Refseq
Affymetrix Human Clariom D Assay TC0700007998.hg.1 NM_022170.1 Refseq
Affymetrix Human Clariom D Assay TC0700007998.hg.1 NM_022170.1 Refseq
Affymetrix Human Clariom D Assay TC0700007998.hg.1 NM_022170.1 Refseq
Affymetrix Human Clariom D Assay TC0700007998.hg.1 NM_031992.1 Refseq
Affymetrix Human Clariom D Assay TC0700007998.hg.1 NM_031992.1 Refseq
Affymetrix Human Clariom D Assay TC0700007998.hg.1 NM_031992.1 Refseq
Affymetrix Human Clariom D Assay TC0700007998.hg.1 NM_031992.1 Refseq
Affymetrix Human Clariom D Assay TC0700007998.hg.1 NM_031992.1 Refseq
Affymetrix Human Clariom D Assay TC0700007998.hg.1 NM_031992.1 Refseq
Affymetrix Human Clariom D Assay TC0700007998.hg.1 NM_031992.1 Refseq
Affymetrix Human Clariom D Assay TC0700007998.hg.1 NM_031992.1 Refseq
Affymetrix Human Clariom D Assay PSR0700155236.hg.1 ENST00000265753.12 Ensembl
Affymetrix Human Clariom D Assay PSR0700155236.hg.1 ENST00000353999.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700155236.hg.1 ENST00000479338.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700155236.hg.1 ENST00000489484.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700155236.hg.1 ENST00000495187.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700155236.hg.1 NM_022170.1 Refseq
Affymetrix Human Clariom D Assay PSR0700155236.hg.1 NM_031992.1 Refseq
Affymetrix Human Clariom D Assay PSR0700155236.hg.1 ENST00000265753.12 Ensembl
Affymetrix Human Clariom D Assay PSR0700155236.hg.1 ENST00000353999.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700155236.hg.1 ENST00000479338.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700155236.hg.1 ENST00000489484.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700155236.hg.1 ENST00000495187.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700155236.hg.1 NM_022170.1 Refseq
Affymetrix Human Clariom D Assay PSR0700155236.hg.1 NM_031992.1 Refseq
Affymetrix Human Clariom D Assay PSR0700155236.hg.1 TC0700007998.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0