Assay Details
| Gene Symbol: DTX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004416.2 | NP_004407.2 | 7 - 8 | 1890 | 83 | |
| XM_011538009.2 | XP_011536311.1 | 8 - 9 | 2532 | 83 | ||
| GenBank mRNA | AF053700.1 | - | 7 - 8 | 2057 | 83 | |
| BC005816.2 | - | 3 - 4 | 369 | 83 | ||
| BC048216.1 | - | 7 - 8 | 1832 | 5931062 | 83 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000887.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
transcription from RNA polymerase II promoter
cell surface receptor signaling pathway
Notch signaling pathway
regulation of Notch signaling pathway
glial cell differentiation
protein ubiquitination
negative regulation of T cell differentiation
negative regulation of neuron differentiation
regulation of nucleic acid-templated transcription
transcription coactivator activity
Notch binding
protein binding
zinc ion binding
ligase activity
SH3 domain binding
ubiquitin protein ligase binding
transcription, DNA-templated
transcription from RNA polymerase II promoter
cell surface receptor signaling pathway
Notch signaling pathway
regulation of Notch signaling pathway
glial cell differentiation
protein ubiquitination
negative regulation of T cell differentiation
negative regulation of neuron differentiation
regulation of nucleic acid-templated transcription
transcription coactivator activity
Notch binding
protein binding
zinc ion binding
ligase activity
SH3 domain binding
ubiquitin protein ligase binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12011733.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011733.hg.1 | ENST00000547974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011733.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011733.hg.1 | TC12000887.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011732.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011732.hg.1 | ENST00000547974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011732.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011732.hg.1 | TC12000887.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000547730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000547730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000547730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000547974.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000547974.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000547974.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000547974.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000547974.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000547974.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000547974.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000547974.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000547974.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000548759.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000548759.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000548759.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000548759.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000548759.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000553140.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000553140.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000553140.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000553140.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200166364.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166364.hg.1 | ENST00000547730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166364.hg.1 | ENST00000547974.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166364.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200166364.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166364.hg.1 | ENST00000547730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166364.hg.1 | ENST00000547974.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166364.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200166364.hg.1 | TC1200008924.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200166363.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166363.hg.1 | ENST00000547730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166363.hg.1 | ENST00000547974.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166363.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200166363.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166363.hg.1 | ENST00000547730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166363.hg.1 | ENST00000547974.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200166363.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200166363.hg.1 | TC1200008924.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000547730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000547730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000547730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000547974.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000547974.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000547974.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000547974.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000547974.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000547974.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000547974.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000547974.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000547974.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000548759.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000548759.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000548759.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000548759.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000548759.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000553140.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000553140.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000553140.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000553140.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | NM_004416.2 | Refseq |

DTX1
Genomic Map