This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01093682_m1
See other STARD8 GE Assays ›
Gene Symbol:
STARD8
Gene Name:
StAR related lipid transfer domain containing 8
Gene Aliases:
ARHGAP38, DLC3, STARTGAP3
Chromosome Location:
Chr.X: 68647669 - 68725842 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01093682_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: STARD8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001142503.2 NP_001135975.1 2 - 3 290 60
NM_014725.4 NP_055540.2 2 - 3 290 60
XM_005262314.4 XP_005262371.1 2 - 3 264 60
GenBank mRNA BC035587.1 - 2 - 3 301 5518429 60
CR749411.1 - 2 - 3 191 60

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0X00007504.hg.1
This assay targets the same location as HTA transcript cluster(s) TC0X000361.hg.1

Additional Information:

This assay is in the 5'UTR region of the BC035587.1,NM_014725.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene STARD8

Panther Classification:

Molecular Function -

G-protein modulator protein-binding activity modulator GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
positive regulation of GTPase activity
regulation of small GTPase mediated signal transduction
GTPase activator activity
lipid binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR0X004681.hg.1 TC0X000361.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR0X004681.hg.1 ENST00000252336.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X004681.hg.1 ENST00000374597.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X004681.hg.1 ENST00000374599.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X004681.hg.1 NM_001142503.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X004681.hg.1 NM_001142504.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X004681.hg.1 NM_014725.4 Refseq
Affymetrix Human Clariom D Assay PSR0X00151398.hg.1 ENST00000252336.10 Ensembl
Affymetrix Human Clariom D Assay PSR0X00151398.hg.1 ENST00000374597.3 Ensembl
Affymetrix Human Clariom D Assay PSR0X00151398.hg.1 ENST00000374599.7 Ensembl
Affymetrix Human Clariom D Assay PSR0X00151398.hg.1 ENST00000523864.5 Ensembl
Affymetrix Human Clariom D Assay PSR0X00151398.hg.1 NM_001142503.2 Refseq
Affymetrix Human Clariom D Assay PSR0X00151398.hg.1 NM_001142504.2 Refseq
Affymetrix Human Clariom D Assay PSR0X00151398.hg.1 NM_014725.4 Refseq
Affymetrix Human Clariom D Assay PSR0X00151398.hg.1 ENST00000252336.10 Ensembl
Affymetrix Human Clariom D Assay PSR0X00151398.hg.1 ENST00000374597.3 Ensembl
Affymetrix Human Clariom D Assay PSR0X00151398.hg.1 ENST00000374599.7 Ensembl
Affymetrix Human Clariom D Assay PSR0X00151398.hg.1 ENST00000523864.5 Ensembl
Affymetrix Human Clariom D Assay PSR0X00151398.hg.1 NM_001142503.2 Refseq
Affymetrix Human Clariom D Assay PSR0X00151398.hg.1 NM_001142504.2 Refseq
Affymetrix Human Clariom D Assay PSR0X00151398.hg.1 NM_014725.4 Refseq
Affymetrix Human Clariom D Assay PSR0X00151398.hg.1 TC0X00007504.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0