This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01095334_m1
See other ATF2 GE Assays ›
Gene Symbol:
ATF2
Gene Name:
activating transcription factor 2
Gene Aliases:
CRE-BP1, CREB-2, CREB2, HB16, TREB7
Chromosome Location:
Chr.2: 175072250 - 175168206 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01095334_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATF2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001256090.1 NP_001243019.1 13 - 14 1525 130
NM_001256091.1 NP_001243020.1 12 - 13 1450 130
NM_001256092.1 NP_001243021.1 11 - 12 1393 130
NM_001256093.1 NP_001243022.1 11 - 12 1393 130
NM_001880.3 NP_001871.2 12 - 13 1490 130
NR_045774.1 11 - 12 1375 130
GenBank mRNA AK128731.1 - 12 - 13 1390 130
BC107698.1 - 11 - 12 1360 6456836 130
BC130335.1 - 11 - 12 1256 40146366 130
BC130337.1 - 11 - 12 1256 40146368 130
DQ003037.1 - 12 - 13 1453 130
DQ003041.1 - 13 - 14 1488 130
DQ003042.1 - 12 - 13 1391 130
DQ003045.1 - 11 - 12 1356 130
DQ003047.1 - 11 - 12 1338 130
DQ003048.1 - 11 - 12 1356 130
DQ003049.1 - 12 - 13 1413 130
M31630.1 - 4 - 5 561 130
X15875.1 - 10 - 11 1217 130

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200015009.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02002545.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ATF2

Panther Classification:

Molecular Function -

basic leucine zipper transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
outflow tract morphogenesis
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
response to osmotic stress
cellular response to DNA damage stimulus
response to water deprivation
histone acetylation
intra-S DNA damage checkpoint
positive regulation of transforming growth factor beta2 production
positive regulation of neuron apoptotic process
fat cell differentiation
positive regulation of transcription from RNA polymerase II promoter
negative regulation of epithelial cell proliferation
regulation of sequence-specific DNA binding transcription factor activity
positive regulation of sequence-specific DNA binding transcription factor activity
adipose tissue development
amelogenesis
positive regulation of mitochondrial membrane permeability involved in apoptotic process
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
transcription factor activity, RNA polymerase II transcription factor binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II activating transcription factor binding
enhancer sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
transcription coactivator activity
histone acetyltransferase activity
protein binding
cAMP response element binding protein binding
protein kinase binding
cAMP response element binding
metal ion binding
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR02040474.hg.1 TC02002545.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR02040474.hg.1 ENST00000264110.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02040474.hg.1 ENST00000345739.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02040474.hg.1 ENST00000392543.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02040474.hg.1 ENST00000392544.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02040474.hg.1 ENST00000409437.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02040474.hg.1 ENST00000409499.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02040474.hg.1 ENST00000409635.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02040474.hg.1 ENST00000426833.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02040474.hg.1 ENST00000487334.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02040474.hg.1 ENST00000538946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02040474.hg.1 ENST00000542046.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02040474.hg.1 NM_001256090.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02040474.hg.1 NM_001256091.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02040474.hg.1 NM_001256092.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02040474.hg.1 NM_001256093.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02040474.hg.1 NM_001880.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02040474.hg.1 NR_045768.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02040474.hg.1 NR_045769.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02040474.hg.1 NR_045770.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02040474.hg.1 NR_045771.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02040474.hg.1 NR_045772.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02040474.hg.1 NR_045773.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02040474.hg.1 NR_045774.1 Refseq
Affymetrix Human Clariom D Assay PSR0200209566.hg.1 ENST00000417080.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200209566.hg.1 NM_001256093.1 Refseq
Affymetrix Human Clariom D Assay PSR0200209566.hg.1 NR_045769.1 Refseq
Affymetrix Human Clariom D Assay PSR0200209566.hg.1 ENST00000417080.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200209566.hg.1 NM_001256093.1 Refseq
Affymetrix Human Clariom D Assay PSR0200209566.hg.1 NR_045769.1 Refseq
Affymetrix Human Clariom D Assay PSR0200209566.hg.1 TC0200015009.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 ENST00000264110.6 Ensembl
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 ENST00000345739.9 Ensembl
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 ENST00000392544.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 ENST00000409437.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 ENST00000409499.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 ENST00000409635.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 ENST00000415955.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 ENST00000417080.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 ENST00000421438.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 ENST00000426833.7 Ensembl
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 ENST00000428760.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 ENST00000429579.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 ENST00000456655.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 ENST00000478905.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 ENST00000538946.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 NM_001256090.1 Refseq
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 NM_001256091.1 Refseq
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 NM_001256092.1 Refseq
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 NM_001256093.1 Refseq
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 NM_001880.3 Refseq
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 NR_045768.1 Refseq
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 NR_045769.1 Refseq
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 NR_045770.1 Refseq
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 NR_045771.1 Refseq
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 NR_045772.1 Refseq
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 NR_045773.1 Refseq
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 NR_045774.1 Refseq
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 ENST00000264110.6 Ensembl
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 ENST00000345739.9 Ensembl
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 ENST00000392544.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 ENST00000409437.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 ENST00000409499.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 ENST00000409635.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 ENST00000415955.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 ENST00000417080.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 ENST00000421438.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 ENST00000426833.7 Ensembl
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 ENST00000428760.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 ENST00000429579.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 ENST00000456655.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 ENST00000478905.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 ENST00000538946.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 NM_001256090.1 Refseq
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 NM_001256091.1 Refseq
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 NM_001256092.1 Refseq
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 NM_001256093.1 Refseq
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 NM_001880.3 Refseq
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 NR_045768.1 Refseq
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 NR_045769.1 Refseq
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 NR_045770.1 Refseq
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 NR_045771.1 Refseq
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 NR_045772.1 Refseq
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 NR_045773.1 Refseq
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 NR_045774.1 Refseq
Affymetrix Human Clariom D Assay PSR0200209569.hg.1 TC0200015009.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0