This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01095338_m1
See other ATF2 GE Assays ›
Gene Symbol:
ATF2
Gene Name:
activating transcription factor 2
Gene Aliases:
CRE-BP1, CREB-2, CREB2, HB16, TREB7
Chromosome Location:
Chr.2: 175072250 - 175168206 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01095338_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATF2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001256092.1 NP_001243021.1 3 - 4 330 148
NM_001256093.1 NP_001243022.1 3 - 4 330 148
NM_001256094.1 NP_001243023.1 3 - 4 330 148
NM_001880.3 NP_001871.2 3 - 4 330 148
NR_045771.1 3 - 4 330 148
NR_045773.1 3 - 4 330 148
NR_045774.1 3 - 4 330 148
GenBank mRNA AK128731.1 - 3 - 4 299 148
BC026175.1 - 3 - 4 297 148
BC107698.1 - 3 - 4 297 6456836 148
BC130335.1 - 2 - 3 96 40146366 148
BC130337.1 - 2 - 3 96 40146368 148
DA995464.1 - 3 - 4 330 148
DQ003037.1 - 3 - 4 293 148
DQ003038.1 - 3 - 4 293 148
DQ003039.1 - 3 - 4 293 148
DQ003045.1 - 3 - 4 293 148
DQ003046.1 - 3 - 4 293 148
DQ003047.1 - 3 - 4 293 148
DQ003048.1 - 3 - 4 293 148

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK128731.1, BC026175.1, BC107698.1, BC130335.1, BC130337.1, DQ003037.1, DQ003038.1, DQ003039.1, DQ003045.1, DQ003046.1, DQ003047.1, DQ003048.1, NM_001256092.1, NM_001256093.1, NM_001256094.1, NM_001880.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ATF2

Panther Classification:

Molecular Function -

basic leucine zipper transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
outflow tract morphogenesis
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
response to osmotic stress
cellular response to DNA damage stimulus
response to water deprivation
histone acetylation
intra-S DNA damage checkpoint
positive regulation of transforming growth factor beta2 production
positive regulation of neuron apoptotic process
fat cell differentiation
positive regulation of transcription from RNA polymerase II promoter
negative regulation of epithelial cell proliferation
regulation of sequence-specific DNA binding transcription factor activity
positive regulation of sequence-specific DNA binding transcription factor activity
adipose tissue development
amelogenesis
positive regulation of mitochondrial membrane permeability involved in apoptotic process
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
transcription factor activity, RNA polymerase II transcription factor binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II activating transcription factor binding
enhancer sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
transcription coactivator activity
histone acetyltransferase activity
protein binding
cAMP response element binding protein binding
protein kinase binding
cAMP response element binding
metal ion binding
protein heterodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0