This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01097978_s1
See other PDE4B GE Assays ›
Gene Symbol:
PDE4B
Gene Name:
phosphodiesterase 4B
Gene Aliases:
DPDE4, PDEIVB
Chromosome Location:
Chr.1: 65792510 - 66374579 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01097978_s1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PDE4B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK000989.1 - 1 1012 115

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000262961.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000262961.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000262961.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000262961.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000262961.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000262961.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000262961.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000262961.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000262961.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000262961.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000262961.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000262961.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000262961.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000262961.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000262961.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000262961.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000262961.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000262961.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000262961.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000262961.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000262961.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000262961.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000262961.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000262961.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000438164.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000438164.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000438164.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000438164.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000438164.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000438164.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000438164.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000438164.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000438164.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000438164.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000438164.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000438164.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000439086.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000439086.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000439086.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000439086.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000586578.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000586578.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000586578.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000586578.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000586578.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000587212.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000587212.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000587212.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000587212.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000591712.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000591712.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000591712.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000591965.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000591965.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000591965.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000591965.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000591965.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000591965.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000592398.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000592398.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000592398.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000592398.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000592398.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000592807.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000592807.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000592807.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000592807.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 ENST00000592807.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 NM_001145640.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 NM_001145640.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 NM_001145640.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 NM_001145640.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 NM_015174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 NM_015174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 NM_015174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 NM_015174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 NM_015174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 NM_015174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 NM_015174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 NM_015174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 NM_015174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 NM_015174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 NM_015174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 NM_015174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 NM_015174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 NM_015174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 NM_015174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 NM_015174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 NM_015174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 NM_015174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 NM_015174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 NM_015174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 NM_015174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 NM_015174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 NM_015174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 NM_015174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 NR_027149.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 NR_027149.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 NR_027149.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 NR_027149.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009311.hg.1 NR_027149.1 Refseq
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000262961.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000262961.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000262961.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000262961.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000262961.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000262961.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000262961.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000262961.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000262961.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000262961.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000262961.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000262961.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000262961.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000262961.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000262961.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000262961.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000262961.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000262961.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000262961.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000262961.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000262961.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000262961.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000262961.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000262961.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000438164.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000438164.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000438164.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000438164.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000438164.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000438164.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000438164.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000438164.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000438164.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000438164.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000438164.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000438164.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000439086.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000439086.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000439086.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000439086.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000586578.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000586578.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000586578.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000586578.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000586578.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000587212.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000587212.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000587212.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000587212.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000591712.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000591712.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000591712.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000591965.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000591965.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000591965.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000591965.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000591965.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000591965.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000592398.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000592398.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000592398.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000592398.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000592398.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000592807.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000592807.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000592807.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000592807.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 ENST00000592807.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009311.hg.1 NM_001145640.1 Refseq
Affymetrix Human Clariom D Assay TC1900009311.hg.1 NM_001145640.1 Refseq
Affymetrix Human Clariom D Assay TC1900009311.hg.1 NM_001145640.1 Refseq
Affymetrix Human Clariom D Assay TC1900009311.hg.1 NM_001145640.1 Refseq
Affymetrix Human Clariom D Assay TC1900009311.hg.1 NM_015174.1 Refseq
Affymetrix Human Clariom D Assay TC1900009311.hg.1 NM_015174.1 Refseq
Affymetrix Human Clariom D Assay TC1900009311.hg.1 NM_015174.1 Refseq
Affymetrix Human Clariom D Assay TC1900009311.hg.1 NM_015174.1 Refseq
Affymetrix Human Clariom D Assay TC1900009311.hg.1 NM_015174.1 Refseq
Affymetrix Human Clariom D Assay TC1900009311.hg.1 NM_015174.1 Refseq
Affymetrix Human Clariom D Assay TC1900009311.hg.1 NM_015174.1 Refseq
Affymetrix Human Clariom D Assay TC1900009311.hg.1 NM_015174.1 Refseq
Affymetrix Human Clariom D Assay TC1900009311.hg.1 NM_015174.1 Refseq
Affymetrix Human Clariom D Assay TC1900009311.hg.1 NM_015174.1 Refseq
Affymetrix Human Clariom D Assay TC1900009311.hg.1 NM_015174.1 Refseq
Affymetrix Human Clariom D Assay TC1900009311.hg.1 NM_015174.1 Refseq
Affymetrix Human Clariom D Assay TC1900009311.hg.1 NM_015174.1 Refseq
Affymetrix Human Clariom D Assay TC1900009311.hg.1 NM_015174.1 Refseq
Affymetrix Human Clariom D Assay TC1900009311.hg.1 NM_015174.1 Refseq
Affymetrix Human Clariom D Assay TC1900009311.hg.1 NM_015174.1 Refseq
Affymetrix Human Clariom D Assay TC1900009311.hg.1 NM_015174.1 Refseq
Affymetrix Human Clariom D Assay TC1900009311.hg.1 NM_015174.1 Refseq
Affymetrix Human Clariom D Assay TC1900009311.hg.1 NM_015174.1 Refseq
Affymetrix Human Clariom D Assay TC1900009311.hg.1 NM_015174.1 Refseq
Affymetrix Human Clariom D Assay TC1900009311.hg.1 NM_015174.1 Refseq
Affymetrix Human Clariom D Assay TC1900009311.hg.1 NM_015174.1 Refseq
Affymetrix Human Clariom D Assay TC1900009311.hg.1 NM_015174.1 Refseq
Affymetrix Human Clariom D Assay TC1900009311.hg.1 NM_015174.1 Refseq
Affymetrix Human Clariom D Assay TC1900009311.hg.1 NR_027149.1 Refseq
Affymetrix Human Clariom D Assay TC1900009311.hg.1 NR_027149.1 Refseq
Affymetrix Human Clariom D Assay TC1900009311.hg.1 NR_027149.1 Refseq
Affymetrix Human Clariom D Assay TC1900009311.hg.1 NR_027149.1 Refseq
Affymetrix Human Clariom D Assay TC1900009311.hg.1 NR_027149.1 Refseq
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 270143 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline