This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01098121_m1
See other NPAT GE Assays ›
Gene Symbol:
NPAT
Gene Name:
nuclear protein, coactivator of histone transcription
Gene Aliases:
E14, E14/NPAT, p220
Chromosome Location:
Chr.11: 108155280 - 108222638 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01098121_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NPAT
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001321307.1 NP_001308236.1 15 - 16 3106 75
NM_002519.2 NP_002510.2 15 - 16 3106 75
XM_011542854.2 XP_011541156.1 15 - 16 3106 75
XM_017017802.1 XP_016873291.1 15 - 16 3844 75
XM_017017803.1 XP_016873292.1 15 - 16 3287 75
GenBank mRNA AK290020.1 - 15 - 16 3106 75
AK291687.1 - 15 - 16 75
AK301256.1 - 2 - 3 377 75
BC040356.1 - 15 - 16 3097 75
BC050561.2 - 15 - 16 3091 75
BC136270.1 - 15 - 16 9020257 75
D83243.1 - 15 - 16 3070 75
U58852.1 - 15 - 16 3004 75
X97186.1 - 15 - 16 3038 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100012225.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11002264.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NPAT

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription involved in G1/S transition of mitotic cell cycle
transcription, DNA-templated
regulation of gene expression
positive regulation of transcription, DNA-templated
negative regulation of nucleic acid-templated transcription
transcription coactivator activity
transcription corepressor activity
protein binding
protein C-terminus binding
protein N-terminus binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR11027128.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11027128.hg.1 NM_002519.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11027128.hg.1 TC11002264.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR11027127.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11027127.hg.1 NM_002519.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11027127.hg.1 TC11002264.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000527296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000527296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000527296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000527296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000530859.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000530859.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000530859.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000530859.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000530859.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000530859.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000530859.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000530859.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000530859.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000530859.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000530926.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000530926.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000530926.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000530926.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000531384.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000531384.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000531384.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000531384.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000531384.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000531384.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000531384.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000610253.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000610253.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000610253.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000610253.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000610253.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000610253.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000610253.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000610253.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000610253.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000610253.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000610253.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000610253.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000610253.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000610253.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 ENST00000610253.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Clariom D Assay PSR1100194012.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Clariom D Assay PSR1100194012.hg.1 ENST00000530859.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100194012.hg.1 NM_002519.2 Refseq
Affymetrix Human Clariom D Assay PSR1100194012.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Clariom D Assay PSR1100194012.hg.1 ENST00000530859.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100194012.hg.1 NM_002519.2 Refseq
Affymetrix Human Clariom D Assay PSR1100194012.hg.1 TC1100012225.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1100194010.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Clariom D Assay PSR1100194010.hg.1 ENST00000527296.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100194010.hg.1 ENST00000530859.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100194010.hg.1 NM_002519.2 Refseq
Affymetrix Human Clariom D Assay PSR1100194010.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Clariom D Assay PSR1100194010.hg.1 ENST00000527296.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100194010.hg.1 ENST00000530859.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100194010.hg.1 NM_002519.2 Refseq
Affymetrix Human Clariom D Assay PSR1100194010.hg.1 TC1100012225.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000278612.8 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000527296.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000527296.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000527296.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000527296.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000530859.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000530859.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000530859.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000530859.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000530859.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000530859.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000530859.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000530859.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000530859.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000530859.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000530926.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000530926.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000530926.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000530926.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000531384.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000531384.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000531384.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000531384.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000531384.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000531384.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000531384.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000610253.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000610253.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000610253.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000610253.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000610253.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000610253.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000610253.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000610253.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000610253.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000610253.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000610253.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000610253.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000610253.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000610253.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 ENST00000610253.5 Ensembl
Affymetrix Human Clariom D Assay TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Clariom D Assay TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Clariom D Assay TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Clariom D Assay TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Clariom D Assay TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Clariom D Assay TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Clariom D Assay TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Clariom D Assay TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Clariom D Assay TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Clariom D Assay TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Clariom D Assay TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Clariom D Assay TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Clariom D Assay TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Clariom D Assay TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Clariom D Assay TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Clariom D Assay TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Clariom D Assay TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Clariom D Assay TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Clariom D Assay TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Clariom D Assay TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Clariom D Assay TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Clariom D Assay TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Clariom D Assay TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Clariom D Assay TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Clariom D Assay TC1100012225.hg.1 NM_002519.2 Refseq
Affymetrix Human Clariom D Assay TC1100012225.hg.1 NM_002519.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline