This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01098505_m1
See other MEG3 GE Assays ›
Gene Symbol:
MEG3
Gene Name:
maternally expressed 3 (non-protein coding)
Gene Aliases:
FP504, GTL2, LINC00023, NCRNA00023, PRO0518, PRO2160, onco-lncRNA-83, prebp1
Chromosome Location:
Chr.14: 100826108 - 100861026 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01098505_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MEG3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_002766.2 4 - 5 1049 81
NR_046465.2 4 - 5 1087 81
NR_046473.1 4 - 5 1049 81
GenBank mRNA AB032607.1 - 4 - 5 80
AF052114.1 - 2 - 3 791 81
BC092509.1 - 2 - 3 244 81
GQ183496.1 - 4 - 5 1080 81
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BM679015.1 - 3 - 4 Chr. 14 - 100831445 - 100861026 [-] on Build GRCh38 81
GenBankmRNA BX407692.2 - 3 - 4 Chr. 14 - 100829293 - 100860987 [-] on Build GRCh38 81
GenBankmRNA BI913162.1 - 1 - 2 97 Chr. 14 - 100831438 - 100861007 [+] on Build GRCh38 81
GenBankmRNA BX412723.1 - Chr. - - [-] on Build 81
GenBankmRNA BU579722.1 - 1 - 2 57 Chr. 14 - 100831478 - 100860862 [+] on Build GRCh38 81
GenBankmRNA BF932705.1 - 293 Chr. - - [-] on Build 81
GenBankmRNA BX462854.2 - 3 - 4 Chr. 14 - 100829294 - 100860989 [-] on Build GRCh38 81
GenBankmRNA BG703541.1 - 2 - 3 249 Chr. 14 - 100829615 - 100846219 [+] on Build GRCh38 81
GenBankmRNA BY796127.2 - 2 - 3 418 Chr. 14 - 100829446 - 100845508 [+] on Build GRCh38 81
GenBankmRNA CA943189.1 - 2 - 3 343 Chr. 14 - 100829522 - 100845520 [+] on Build GRCh38 81
GenBankmRNA BX426730.2 - 1 - 2 44 Chr. 14 - 100831491 - 100845533 [+] on Build GRCh38 80
GenBankmRNA BX453065.2 - 97 Chr. - - [-] on Build 81
GenBankmRNA BX408784.2 - 2 - 3 288 Chr. 14 - 100829579 - 100861025 [+] on Build GRCh38 81
GenBankmRNA AI174720.1 - 2 - 3 467 Chr. 14 - 100829397 - 100836304 [+] on Build GRCh38 81
GenBankmRNA BX368158.1 - 2 - 3 491 Chr. 14 - 100829371 - 100860859 [+] on Build GRCh38 81
GenBankmRNA BU787606.1 - 1 - 2 57 Chr. 14 - 100831478 - 100861005 [+] on Build GRCh38 81
GenBankmRNA BX449880.2 - 3 - 4 Chr. 14 - 100829411 - 100860977 [-] on Build GRCh38 81
GenBankmRNA BX332214.2 - 3 - 4 Chr. 14 - 100829643 - 100860945 [-] on Build GRCh38 81
GenBankmRNA BX412855.2 - 3 - 4 Chr. 14 - 100829526 - 100861011 [-] on Build GRCh38 81
GenBankmRNA H59650.1 - 244 Chr. - - [-] on Build 80
GenBankmRNA BX367746.1 - 2 - 3 456 Chr. 14 - 100829409 - 100860888 [+] on Build GRCh38 81
GenBankmRNA BU071298.1 - 3 - 4 Chr. 14 - 100831482 - 100861026 [-] on Build GRCh38 81
GenBankmRNA BX408219.1 - 2 - 3 356 Chr. 14 - 100829507 - 100860967 [+] on Build GRCh38 81
GenBankmRNA BX450276.1 - 2 - 3 380 Chr. 14 - 100829484 - 100846142 [+] on Build GRCh38 81
GenBankmRNA BI524541.1 - 3 - 4 Chr. 14 - 100831499 - 100860949 [-] on Build GRCh38 81
GenBankmRNA BX434722.2 - 3 - 4 Chr. 14 - 100829536 - 100861014 [-] on Build GRCh38 81
GenBankmRNA BX428872.2 - 1 - 2 98 Chr. 14 - 100831437 - 100861026 [+] on Build GRCh38 81
GenBankmRNA BX408790.2 - 2 - 3 195 Chr. 14 - 100829668 - 100860997 [+] on Build GRCh38 81
GenBankmRNA BU783773.1 - 1 - 2 87 Chr. 14 - 100831456 - 100860960 [+] on Build GRCh38 81
GenBankmRNA DA361098.1 - 2 - 3 228 Chr. 14 - 100829636 - 100845997 [+] on Build GRCh38 81
GenBankmRNA BX435163.2 - 3 - 4 Chr. 14 - 100829623 - 100861014 [-] on Build GRCh38 81
GenBankmRNA DA083332.1 - 2 - 3 351 Chr. 14 - 100829513 - 100836289 [+] on Build GRCh38 81
GenBankmRNA AV723467.1 - 2 - 3 204 Chr. 14 - 100829658 - 100860731 [+] on Build GRCh38 81
GenBankmRNA CB154926.1 - 2 - 3 296 Chr. 14 - 100829568 - 100860741 [+] on Build GRCh38 81
GenBankmRNA AA029246.1 - 1 - 2 87 Chr. 14 - 100831448 - 100860775 [+] on Build GRCh38 80
GenBankmRNA BU075431.1 - 2 - 3 207 Chr. 14 - 100829679 - 100860862 [+] on Build GRCh38 81
GenBankmRNA BX429998.2 - 2 - 3 174 Chr. 14 - 100829691 - 100846355 [+] on Build GRCh38 81
GenBankmRNA AV727351.1 - 3 - 4 485 Chr. 14 - 100829325 - 100845483 [+] on Build GRCh38 81
GenBankmRNA BX429896.2 - 2 - 3 239 Chr. 14 - 100829639 - 100846333 [+] on Build GRCh38 81
GenBankmRNA AV753036.1 - 390 Chr. - - [-] on Build 81
GenBankmRNA BX453064.2 - 1 - 2 98 Chr. 14 - 100831437 - 100861026 [+] on Build GRCh38 81
GenBankmRNA N56264.1 - 1 - 2 Chr. 14 - 100831464 - 100845534 [+] on Build GRCh38 77
GenBankmRNA BX455272.2 - 3 - 4 Chr. 14 - 100829516 - 100861016 [-] on Build GRCh38 81
GenBankmRNA BE222690.1 - 3 - 4 Chr. 14 - 100831498 - 100861026 [-] on Build GRCh38 81
GenBankmRNA BX408226.1 - 2 - 3 373 Chr. 14 - 100829491 - 100860979 [+] on Build GRCh38 81
GenBankmRNA BX429546.1 - 3 - 4 Chr. 14 - 100829241 - 100845925 [-] on Build GRCh38 81
GenBankmRNA BI524001.1 - 36 Chr. - - [-] on Build 81
GenBankmRNA BX429869.2 - 2 - 3 236 Chr. 14 - 100829629 - 100861023 [+] on Build GRCh38 81
GenBankmRNA BX408189.1 - 2 - 3 402 Chr. 14 - 100829460 - 100860925 [+] on Build GRCh38 81
GenBankmRNA AU309923.1 - 2 - 3 225 Chr. 14 - 100829639 - 100846016 [+] on Build GRCh38 81
GenBankmRNA BX394256.2 - 3 - 4 Chr. 14 - 100831004 - 100860977 [-] on Build GRCh38 81
GenBankmRNA BX423733.2 - 1 - 2 44 Chr. 14 - 100831491 - 100861026 [+] on Build GRCh38 81
GenBankmRNA BX405425.2 - 3 - 4 Chr. 14 - 100831088 - 100860993 [-] on Build GRCh38 81
GenBankmRNA BX450687.2 - 2 - 3 280 Chr. 14 - 100829584 - 100860988 [+] on Build GRCh38 81

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream microRNA 2392 MIR2392 -16.8kb Sense Chr. 14 - 100814491 - 100814574 on NCBI Build GRCh38
Downstream microRNA 770 MIR770 MIRN770,
hsa-mir-770
16.1kb Sense Chr. 14 - 100852390 - 100852487 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline