This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01099228_m1
See other DHX15 GE Assays ›
Gene Symbol:
DHX15
Gene Name:
DEAH-box helicase 15
Gene Aliases:
DBP1, DDX15, HRH2, PRP43, PRPF43, PrPp43p
Chromosome Location:
Chr.4: 24527464 - 24584603 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01099228_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DHX15
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001358.2 NP_001349.2 10 - 11 1954 141
GenBank mRNA AB001636.1 - 10 - 11 1947 141
AF279891.1 - 10 - 11 1786 141
AK303506.1 - 10 - 11 1894 141
BC035974.1 - 10 - 11 1941 4806964 141

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0400010251.hg.1
This assay targets the same location as HTA transcript cluster(s) TC04001086.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene DHX15

Panther Classification:

Molecular Function -

RNA helicase

Gene Ontology Categories:

Function(s) Process(es)

mRNA splicing, via spliceosome
mRNA processing
RNA splicing
response to toxic substance
response to alkaloid
RNA helicase activity
double-stranded RNA binding
ATP-dependent RNA helicase activity
protein binding
ATP binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR04014931.hg.1 ENST00000336812.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04014931.hg.1 ENST00000508032.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04014931.hg.1 ENST00000535946.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04014931.hg.1 NM_001358.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04014931.hg.1 TC04001086.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000336812.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000336812.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000336812.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000336812.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000336812.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000336812.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000336812.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000336812.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000336812.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000336812.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000336812.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000336812.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000336812.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000336812.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000336812.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000336812.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000336812.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000336812.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000336812.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000336812.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000336812.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000336812.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000476316.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000503562.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000503562.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000503562.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000503562.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000504279.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000504279.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000504279.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000504279.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000508032.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000508032.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000508032.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000508032.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000508032.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000508032.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000508032.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000508032.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000508032.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000508368.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000508368.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000508368.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000508368.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000510645.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000510645.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000510645.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000511553.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000511553.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000511553.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000511553.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000512903.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000512903.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000512903.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000512903.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000512903.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000512903.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000512903.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000512903.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000513036.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000513036.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000513036.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000513036.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000513036.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000513036.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000513036.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000513092.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000513092.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000513092.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 ENST00000513092.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 NM_001358.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 NM_001358.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 NM_001358.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 NM_001358.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 NM_001358.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 NM_001358.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 NM_001358.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 NM_001358.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 NM_001358.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 NM_001358.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 NM_001358.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 NM_001358.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 NM_001358.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 NM_001358.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 NM_001358.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 NM_001358.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 NM_001358.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 NM_001358.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 NM_001358.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 NM_001358.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 NM_001358.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010251.hg.1 NM_001358.2 Refseq
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000336812.4 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000336812.4 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000336812.4 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000336812.4 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000336812.4 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000336812.4 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000336812.4 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000336812.4 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000336812.4 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000336812.4 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000336812.4 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000336812.4 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000336812.4 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000336812.4 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000336812.4 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000336812.4 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000336812.4 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000336812.4 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000336812.4 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000336812.4 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000336812.4 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000336812.4 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000476316.3 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000503562.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000503562.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000503562.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000503562.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000504279.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000504279.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000504279.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000504279.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000508032.5 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000508032.5 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000508032.5 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000508032.5 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000508032.5 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000508032.5 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000508032.5 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000508032.5 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000508032.5 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000508368.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000508368.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000508368.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000508368.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000510645.5 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000510645.5 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000510645.5 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000511553.5 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000511553.5 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000511553.5 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000511553.5 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000512903.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000512903.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000512903.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000512903.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000512903.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000512903.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000512903.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000512903.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000513036.5 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000513036.5 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000513036.5 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000513036.5 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000513036.5 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000513036.5 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000513036.5 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000513092.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000513092.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000513092.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 ENST00000513092.1 Ensembl
Affymetrix Human Clariom D Assay TC0400010251.hg.1 NM_001358.2 Refseq
Affymetrix Human Clariom D Assay TC0400010251.hg.1 NM_001358.2 Refseq
Affymetrix Human Clariom D Assay TC0400010251.hg.1 NM_001358.2 Refseq
Affymetrix Human Clariom D Assay TC0400010251.hg.1 NM_001358.2 Refseq
Affymetrix Human Clariom D Assay TC0400010251.hg.1 NM_001358.2 Refseq
Affymetrix Human Clariom D Assay TC0400010251.hg.1 NM_001358.2 Refseq
Affymetrix Human Clariom D Assay TC0400010251.hg.1 NM_001358.2 Refseq
Affymetrix Human Clariom D Assay TC0400010251.hg.1 NM_001358.2 Refseq
Affymetrix Human Clariom D Assay TC0400010251.hg.1 NM_001358.2 Refseq
Affymetrix Human Clariom D Assay TC0400010251.hg.1 NM_001358.2 Refseq
Affymetrix Human Clariom D Assay TC0400010251.hg.1 NM_001358.2 Refseq
Affymetrix Human Clariom D Assay TC0400010251.hg.1 NM_001358.2 Refseq
Affymetrix Human Clariom D Assay TC0400010251.hg.1 NM_001358.2 Refseq
Affymetrix Human Clariom D Assay TC0400010251.hg.1 NM_001358.2 Refseq
Affymetrix Human Clariom D Assay TC0400010251.hg.1 NM_001358.2 Refseq
Affymetrix Human Clariom D Assay TC0400010251.hg.1 NM_001358.2 Refseq
Affymetrix Human Clariom D Assay TC0400010251.hg.1 NM_001358.2 Refseq
Affymetrix Human Clariom D Assay TC0400010251.hg.1 NM_001358.2 Refseq
Affymetrix Human Clariom D Assay TC0400010251.hg.1 NM_001358.2 Refseq
Affymetrix Human Clariom D Assay TC0400010251.hg.1 NM_001358.2 Refseq
Affymetrix Human Clariom D Assay TC0400010251.hg.1 NM_001358.2 Refseq
Affymetrix Human Clariom D Assay TC0400010251.hg.1 NM_001358.2 Refseq
Affymetrix Human Clariom D Assay PSR0400170149.hg.1 ENST00000336812.4 Ensembl
Affymetrix Human Clariom D Assay PSR0400170149.hg.1 ENST00000508032.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400170149.hg.1 NM_001358.2 Refseq
Affymetrix Human Clariom D Assay PSR0400170149.hg.1 ENST00000336812.4 Ensembl
Affymetrix Human Clariom D Assay PSR0400170149.hg.1 ENST00000508032.5 Ensembl
Affymetrix Human Clariom D Assay PSR0400170149.hg.1 NM_001358.2 Refseq
Affymetrix Human Clariom D Assay PSR0400170149.hg.1 TC0400010251.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline