This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01099539_m1
See other IGFALS GE Assays ›
Gene Symbol:
IGFALS
Gene Name:
insulin like growth factor binding protein acid labile subunit
Gene Aliases:
ACLSD, ALS
Chromosome Location:
Chr.16: 1790413 - 1794908 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01099539_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IGFALS
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_004970.2 NP_004961.1 1 - 2 101 109
GenBank mRNA AK315808.1 - 1 - 2 94 109
BC025681.1 - 1 - 2 101 5205012 109

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1600011461.hg.1
This assay targets the same location as HTA transcript cluster(s) TC16000761.hg.1

Additional Information:

This assay is in the 5'UTR region of the AK315808.1,BC025681.1,NM_004970.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene IGFALS

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
signal transduction
cellular protein metabolic process
insulin-like growth factor binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR16011126.hg.1 ENST00000215539.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16011126.hg.1 ENST00000415638.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16011126.hg.1 NM_001146006.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16011126.hg.1 NM_004970.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16011126.hg.1 TC16000761.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 ENST00000215539.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 ENST00000215539.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 ENST00000215539.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 ENST00000215539.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 ENST00000215539.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 ENST00000215539.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 ENST00000415638.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 ENST00000415638.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 ENST00000415638.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 ENST00000415638.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 ENST00000415638.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 ENST00000568221.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 ENST00000568221.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 ENST00000568221.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 NM_001146006.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 NM_001146006.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 NM_001146006.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 NM_001146006.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 NM_001146006.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 NM_001146006.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 NM_004970.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 NM_004970.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 NM_004970.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 NM_004970.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 NM_004970.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 NR_027389.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 NR_027389.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 NR_027389.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 NR_027389.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011461.hg.1 NR_027389.1 Refseq
Affymetrix Human Clariom D Assay PSR1600192183.hg.1 ENST00000215539.3 Ensembl
Affymetrix Human Clariom D Assay PSR1600192183.hg.1 ENST00000415638.3 Ensembl
Affymetrix Human Clariom D Assay PSR1600192183.hg.1 NM_001146006.1 Refseq
Affymetrix Human Clariom D Assay PSR1600192183.hg.1 NM_004970.2 Refseq
Affymetrix Human Clariom D Assay PSR1600192183.hg.1 ENST00000215539.3 Ensembl
Affymetrix Human Clariom D Assay PSR1600192183.hg.1 ENST00000415638.3 Ensembl
Affymetrix Human Clariom D Assay PSR1600192183.hg.1 NM_001146006.1 Refseq
Affymetrix Human Clariom D Assay PSR1600192183.hg.1 NM_004970.2 Refseq
Affymetrix Human Clariom D Assay PSR1600192183.hg.1 TC1600011461.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1600011461.hg.1 ENST00000215539.3 Ensembl
Affymetrix Human Clariom D Assay TC1600011461.hg.1 ENST00000215539.3 Ensembl
Affymetrix Human Clariom D Assay TC1600011461.hg.1 ENST00000215539.3 Ensembl
Affymetrix Human Clariom D Assay TC1600011461.hg.1 ENST00000215539.3 Ensembl
Affymetrix Human Clariom D Assay TC1600011461.hg.1 ENST00000215539.3 Ensembl
Affymetrix Human Clariom D Assay TC1600011461.hg.1 ENST00000215539.3 Ensembl
Affymetrix Human Clariom D Assay TC1600011461.hg.1 ENST00000415638.3 Ensembl
Affymetrix Human Clariom D Assay TC1600011461.hg.1 ENST00000415638.3 Ensembl
Affymetrix Human Clariom D Assay TC1600011461.hg.1 ENST00000415638.3 Ensembl
Affymetrix Human Clariom D Assay TC1600011461.hg.1 ENST00000415638.3 Ensembl
Affymetrix Human Clariom D Assay TC1600011461.hg.1 ENST00000415638.3 Ensembl
Affymetrix Human Clariom D Assay TC1600011461.hg.1 ENST00000568221.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011461.hg.1 ENST00000568221.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011461.hg.1 ENST00000568221.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011461.hg.1 NM_001146006.1 Refseq
Affymetrix Human Clariom D Assay TC1600011461.hg.1 NM_001146006.1 Refseq
Affymetrix Human Clariom D Assay TC1600011461.hg.1 NM_001146006.1 Refseq
Affymetrix Human Clariom D Assay TC1600011461.hg.1 NM_001146006.1 Refseq
Affymetrix Human Clariom D Assay TC1600011461.hg.1 NM_001146006.1 Refseq
Affymetrix Human Clariom D Assay TC1600011461.hg.1 NM_001146006.1 Refseq
Affymetrix Human Clariom D Assay TC1600011461.hg.1 NM_004970.2 Refseq
Affymetrix Human Clariom D Assay TC1600011461.hg.1 NM_004970.2 Refseq
Affymetrix Human Clariom D Assay TC1600011461.hg.1 NM_004970.2 Refseq
Affymetrix Human Clariom D Assay TC1600011461.hg.1 NM_004970.2 Refseq
Affymetrix Human Clariom D Assay TC1600011461.hg.1 NM_004970.2 Refseq
Affymetrix Human Clariom D Assay TC1600011461.hg.1 NR_027389.1 Refseq
Affymetrix Human Clariom D Assay TC1600011461.hg.1 NR_027389.1 Refseq
Affymetrix Human Clariom D Assay TC1600011461.hg.1 NR_027389.1 Refseq
Affymetrix Human Clariom D Assay TC1600011461.hg.1 NR_027389.1 Refseq
Affymetrix Human Clariom D Assay TC1600011461.hg.1 NR_027389.1 Refseq
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 154339 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0