This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01100455_g1
See other DSE GE Assays ›
Gene Symbol:
DSE
Gene Name:
dermatan sulfate epimerase
Gene Aliases:
DS-epi1, DSEP, DSEPI, EDSMC2, SART-2, SART2
Chromosome Location:
Chr.6: 116254152 - 116441261 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01100455_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DSE
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001080976.2 NP_001074445.1 2 - 3 662 70
NM_001322937.1 NP_001309866.1 3 - 4 1218 70
NM_001322938.1 NP_001309867.1 3 - 4 1130 70
NM_001322939.1 NP_001309868.1 2 - 3 545 70
NM_001322943.1 NP_001309872.1 2 - 3 716 70
NM_001322944.1 NP_001309873.1 2 - 3 716 70
NM_013352.3 NP_037484.1 2 - 3 716 70
XM_017010795.1 XP_016866284.1 4 - 5 851 70
XM_017010796.1 XP_016866285.1 1 - 2 472 70
XM_017010797.1 XP_016866286.1 2 - 3 1396 70
GenBank mRNA AF098066.1 - 2 - 3 560 70
AK301513.1 - 2 - 3 545 70
BC039245.1 - 2 - 3 607 5272885 70

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene DSE

Gene Ontology Categories:

Function(s) Process(es)

heparan sulfate proteoglycan biosynthetic process
chondroitin sulfate biosynthetic process
dermatan sulfate biosynthetic process
chondroitin-glucuronate 5-epimerase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline