This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01101016_m1
See other CLPX GE Assays ›
Gene Symbol:
CLPX
Gene Name:
caseinolytic mitochondrial matrix peptidase chaperone subunit
Gene Aliases:
-
Chromosome Location:
Chr.15: 65148219 - 65185420 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01101016_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CLPX
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_006660.4 NP_006651.2 5 - 6 941 73
NR_133680.1 5 - 6 941 73
GenBank mRNA AJ006267.3 - 5 - 6 746 73
AK292316.1 - 5 - 6 867 73
AK308133.1 - 5 - 6 861 73
AK310614.1 - 5 - 6 863 73
AL136922.1 - 5 - 6 844 73
BC130373.1 - 5 - 6 737 40146404 73
BC136487.1 - 5 - 6 737 9020476 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1500009785.hg.1
This assay targets the same location as HTA transcript cluster(s) TC15001561.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CLPX

Panther Classification:

Molecular Function -

protease

Gene Ontology Categories:

Function(s) Process(es)

protein folding
positive regulation of peptidase activity
proteolysis involved in cellular protein catabolic process
ATP-dependent peptidase activity
protein binding
ATP binding
peptidase activator activity
ATPase activity
metal ion binding
unfolded protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000558103.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000558103.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000558103.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000558103.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000559218.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000559218.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000559218.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000559218.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000560166.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000560166.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000560166.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR15014895.hg.1 ENST00000300107.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR15014895.hg.1 ENST00000546194.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR15014895.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR15014895.hg.1 NM_006660.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR15014895.hg.1 TC15001561.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1500172311.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Clariom D Assay PSR1500172311.hg.1 ENST00000558103.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500172311.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500172311.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500172311.hg.1 NM_006660.4 Refseq
Affymetrix Human Clariom D Assay PSR1500172311.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Clariom D Assay PSR1500172311.hg.1 ENST00000558103.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500172311.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500172311.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500172311.hg.1 NM_006660.4 Refseq
Affymetrix Human Clariom D Assay PSR1500172311.hg.1 TC1500009785.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000558103.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000558103.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000558103.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000558103.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000559218.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000559218.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000559218.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000559218.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000560166.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000560166.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000560166.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Clariom D Assay TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Clariom D Assay TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Clariom D Assay TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Clariom D Assay TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Clariom D Assay TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Clariom D Assay TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Clariom D Assay TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Clariom D Assay TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Clariom D Assay TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Clariom D Assay TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Clariom D Assay TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Clariom D Assay TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Clariom D Assay TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Clariom D Assay TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Clariom D Assay TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Clariom D Assay TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Clariom D Assay TC1500009785.hg.1 NM_006660.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nbrrd:80/100.66.77.69:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0