This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01101019_m1
See other CLPX GE Assays ›
Gene Symbol:
CLPX
Gene Name:
caseinolytic mitochondrial matrix peptidase chaperone subunit
Gene Aliases:
-
Chromosome Location:
Chr.15: 65148219 - 65185420 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01101019_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CLPX
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_006660.4 NP_006651.2 8 - 9 1322 104
NR_133680.1 8 - 9 1210 104
XM_011521164.2 XP_011519466.1 7 - 8 1264 104
GenBank mRNA AJ006267.3 - 8 - 9 1127 104
AK292316.1 - 8 - 9 1248 104
AK308133.1 - 8 - 9 1242 104
AK310614.1 - 8 - 9 1244 104
AL136922.1 - 8 - 9 1225 104
BC130373.1 - 8 - 9 1118 40146404 104
BC136487.1 - 8 - 9 1118 9020476 104

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1500009785.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CLPX

Panther Classification:

Molecular Function -

protease

Gene Ontology Categories:

Function(s) Process(es)

protein folding
positive regulation of peptidase activity
proteolysis involved in cellular protein catabolic process
ATP-dependent peptidase activity
protein binding
ATP binding
peptidase activator activity
ATPase activity
metal ion binding
unfolded protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000558103.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000558103.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000558103.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000558103.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000559218.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000559218.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000559218.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000559218.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000560166.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000560166.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 ENST00000560166.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000558103.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000558103.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000558103.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000558103.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000559218.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000559218.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000559218.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000559218.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000560166.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000560166.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 ENST00000560166.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Clariom D Assay TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Clariom D Assay TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Clariom D Assay TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Clariom D Assay TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Clariom D Assay TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Clariom D Assay TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Clariom D Assay TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Clariom D Assay TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Clariom D Assay TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Clariom D Assay TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Clariom D Assay TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Clariom D Assay TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Clariom D Assay TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Clariom D Assay TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Clariom D Assay TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Clariom D Assay TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Clariom D Assay TC1500009785.hg.1 NM_006660.4 Refseq
Affymetrix Human Clariom D Assay PSR1500172306.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Clariom D Assay PSR1500172306.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500172306.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500172306.hg.1 NM_006660.4 Refseq
Affymetrix Human Clariom D Assay PSR1500172306.hg.1 ENST00000300107.7 Ensembl
Affymetrix Human Clariom D Assay PSR1500172306.hg.1 ENST00000558958.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500172306.hg.1 ENST00000559152.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500172306.hg.1 NM_006660.4 Refseq
Affymetrix Human Clariom D Assay PSR1500172306.hg.1 TC1500009785.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-flftk:80/100.66.79.227:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0