This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01102514_m1
See other LAIR2 GE Assays ›
Gene Symbol:
LAIR2
Gene Name:
leukocyte associated immunoglobulin like receptor 2
Gene Aliases:
CD306
Chromosome Location:
Chr.19: 54502797 - 54510693 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01102514_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LAIR2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_021270.4 NP_067154.1 3 - 4 493 112
GenBank mRNA AF013250.1 - 3 - 4 394 112

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1900008862.hg.1
This assay targets the same location as HTA transcript cluster(s) TC19000879.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene LAIR2

Panther Classification:

Molecular Function -

immunoglobulin receptor superfamily

Gene Ontology Categories:

Function(s) Process(es)

regulation of immune response
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR19011946.hg.1 ENST00000301202.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19011946.hg.1 ENST00000351841.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19011946.hg.1 NM_002288.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19011946.hg.1 NM_021270.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19011946.hg.1 TC19000879.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR19011947.hg.1 ENST00000301202.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19011947.hg.1 ENST00000351841.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19011947.hg.1 NM_002288.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19011947.hg.1 NM_021270.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19011947.hg.1 TC19000879.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1900008862.hg.1 ENST00000301202.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008862.hg.1 ENST00000301202.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008862.hg.1 ENST00000301202.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008862.hg.1 ENST00000301202.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008862.hg.1 ENST00000301202.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008862.hg.1 ENST00000301202.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008862.hg.1 ENST00000301202.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008862.hg.1 ENST00000301202.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008862.hg.1 ENST00000301202.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008862.hg.1 ENST00000351841.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008862.hg.1 ENST00000351841.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008862.hg.1 ENST00000351841.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008862.hg.1 ENST00000351841.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008862.hg.1 ENST00000351841.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008862.hg.1 ENST00000351841.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008862.hg.1 ENST00000351841.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008862.hg.1 ENST00000412608.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008862.hg.1 ENST00000412608.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008862.hg.1 ENST00000412608.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008862.hg.1 ENST00000610651.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008862.hg.1 ENST00000610651.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008862.hg.1 NM_002288.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008862.hg.1 NM_002288.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008862.hg.1 NM_002288.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008862.hg.1 NM_002288.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008862.hg.1 NM_002288.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008862.hg.1 NM_002288.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008862.hg.1 NM_002288.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008862.hg.1 NM_002288.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008862.hg.1 NM_002288.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008862.hg.1 NM_021270.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008862.hg.1 NM_021270.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008862.hg.1 NM_021270.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008862.hg.1 NM_021270.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008862.hg.1 NM_021270.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008862.hg.1 NM_021270.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008862.hg.1 NM_021270.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008862.hg.1 NM_021270.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19011944.hg.1 ENST00000301202.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19011944.hg.1 ENST00000351841.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19011944.hg.1 NM_002288.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19011944.hg.1 NM_021270.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19011944.hg.1 TC19000879.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1900169080.hg.1 ENST00000301202.6 Ensembl
Affymetrix Human Clariom D Assay PSR1900169080.hg.1 ENST00000351841.2 Ensembl
Affymetrix Human Clariom D Assay PSR1900169080.hg.1 NM_002288.5 Refseq
Affymetrix Human Clariom D Assay PSR1900169080.hg.1 NM_021270.4 Refseq
Affymetrix Human Clariom D Assay PSR1900169080.hg.1 TC1900008862.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900169080.hg.1 ENST00000301202.6 Ensembl
Affymetrix Human Clariom D Assay PSR1900169080.hg.1 ENST00000351841.2 Ensembl
Affymetrix Human Clariom D Assay PSR1900169080.hg.1 NM_002288.5 Refseq
Affymetrix Human Clariom D Assay PSR1900169080.hg.1 NM_021270.4 Refseq
Affymetrix Human Clariom D Assay PSR1900169081.hg.1 ENST00000301202.6 Ensembl
Affymetrix Human Clariom D Assay PSR1900169081.hg.1 ENST00000351841.2 Ensembl
Affymetrix Human Clariom D Assay PSR1900169081.hg.1 NM_002288.5 Refseq
Affymetrix Human Clariom D Assay PSR1900169081.hg.1 NM_021270.4 Refseq
Affymetrix Human Clariom D Assay PSR1900169081.hg.1 TC1900008862.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900169081.hg.1 ENST00000301202.6 Ensembl
Affymetrix Human Clariom D Assay PSR1900169081.hg.1 ENST00000351841.2 Ensembl
Affymetrix Human Clariom D Assay PSR1900169081.hg.1 NM_002288.5 Refseq
Affymetrix Human Clariom D Assay PSR1900169081.hg.1 NM_021270.4 Refseq
Affymetrix Human Clariom D Assay PSR1900169077.hg.1 ENST00000301202.6 Ensembl
Affymetrix Human Clariom D Assay PSR1900169077.hg.1 ENST00000351841.2 Ensembl
Affymetrix Human Clariom D Assay PSR1900169077.hg.1 NM_002288.5 Refseq
Affymetrix Human Clariom D Assay PSR1900169077.hg.1 NM_021270.4 Refseq
Affymetrix Human Clariom D Assay PSR1900169077.hg.1 TC1900008862.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900169077.hg.1 ENST00000301202.6 Ensembl
Affymetrix Human Clariom D Assay PSR1900169077.hg.1 ENST00000351841.2 Ensembl
Affymetrix Human Clariom D Assay PSR1900169077.hg.1 NM_002288.5 Refseq
Affymetrix Human Clariom D Assay PSR1900169077.hg.1 NM_021270.4 Refseq
Affymetrix Human Clariom D Assay TC1900008862.hg.1 ENST00000301202.6 Ensembl
Affymetrix Human Clariom D Assay TC1900008862.hg.1 ENST00000301202.6 Ensembl
Affymetrix Human Clariom D Assay TC1900008862.hg.1 ENST00000301202.6 Ensembl
Affymetrix Human Clariom D Assay TC1900008862.hg.1 ENST00000301202.6 Ensembl
Affymetrix Human Clariom D Assay TC1900008862.hg.1 ENST00000301202.6 Ensembl
Affymetrix Human Clariom D Assay TC1900008862.hg.1 ENST00000301202.6 Ensembl
Affymetrix Human Clariom D Assay TC1900008862.hg.1 ENST00000301202.6 Ensembl
Affymetrix Human Clariom D Assay TC1900008862.hg.1 ENST00000301202.6 Ensembl
Affymetrix Human Clariom D Assay TC1900008862.hg.1 ENST00000301202.6 Ensembl
Affymetrix Human Clariom D Assay TC1900008862.hg.1 ENST00000351841.2 Ensembl
Affymetrix Human Clariom D Assay TC1900008862.hg.1 ENST00000351841.2 Ensembl
Affymetrix Human Clariom D Assay TC1900008862.hg.1 ENST00000351841.2 Ensembl
Affymetrix Human Clariom D Assay TC1900008862.hg.1 ENST00000351841.2 Ensembl
Affymetrix Human Clariom D Assay TC1900008862.hg.1 ENST00000351841.2 Ensembl
Affymetrix Human Clariom D Assay TC1900008862.hg.1 ENST00000351841.2 Ensembl
Affymetrix Human Clariom D Assay TC1900008862.hg.1 ENST00000351841.2 Ensembl
Affymetrix Human Clariom D Assay TC1900008862.hg.1 ENST00000412608.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008862.hg.1 ENST00000412608.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008862.hg.1 ENST00000412608.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008862.hg.1 ENST00000610651.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008862.hg.1 ENST00000610651.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008862.hg.1 NM_002288.5 Refseq
Affymetrix Human Clariom D Assay TC1900008862.hg.1 NM_002288.5 Refseq
Affymetrix Human Clariom D Assay TC1900008862.hg.1 NM_002288.5 Refseq
Affymetrix Human Clariom D Assay TC1900008862.hg.1 NM_002288.5 Refseq
Affymetrix Human Clariom D Assay TC1900008862.hg.1 NM_002288.5 Refseq
Affymetrix Human Clariom D Assay TC1900008862.hg.1 NM_002288.5 Refseq
Affymetrix Human Clariom D Assay TC1900008862.hg.1 NM_002288.5 Refseq
Affymetrix Human Clariom D Assay TC1900008862.hg.1 NM_002288.5 Refseq
Affymetrix Human Clariom D Assay TC1900008862.hg.1 NM_002288.5 Refseq
Affymetrix Human Clariom D Assay TC1900008862.hg.1 NM_021270.4 Refseq
Affymetrix Human Clariom D Assay TC1900008862.hg.1 NM_021270.4 Refseq
Affymetrix Human Clariom D Assay TC1900008862.hg.1 NM_021270.4 Refseq
Affymetrix Human Clariom D Assay TC1900008862.hg.1 NM_021270.4 Refseq
Affymetrix Human Clariom D Assay TC1900008862.hg.1 NM_021270.4 Refseq
Affymetrix Human Clariom D Assay TC1900008862.hg.1 NM_021270.4 Refseq
Affymetrix Human Clariom D Assay TC1900008862.hg.1 NM_021270.4 Refseq
Affymetrix Human Clariom D Assay TC1900008862.hg.1 NM_021270.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0