This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01103779_m1
See other EPB41L1 GE Assays ›
Gene Symbol:
EPB41L1
Gene Name:
erythrocyte membrane protein band 4.1 like 1
Gene Aliases:
4.1N, MRD11
Chromosome Location:
Chr.20: 36064891 - 36232799 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01103779_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EPB41L1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001258329.1 NP_001245258.1 17 - 18 2407 86
XM_011528681.1 XP_011526983.1 14 - 15 2160 86
XM_011528682.1 XP_011526984.1 14 - 15 2157 86
XM_017027716.1 XP_016883205.1 17 - 18 2386 86
XM_017027717.1 XP_016883206.1 16 - 17 2344 86
GenBank mRNA AY049789.1 - 15 - 16 2191 86
BC040259.1 - 17 - 18 2385 5195986 86
BX537978.1 - 6 - 7 2773 86

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2000007239.hg.1

Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

cortical actin cytoskeleton organization
cell-cell adhesion
actin binding
structural molecule activity
protein binding
cadherin binding involved in cell-cell adhesion

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Clariom D Assay PSR2000149971.hg.1 ENST00000338074.6 Ensembl
Affymetrix Human Clariom D Assay PSR2000149971.hg.1 ENST00000373941.5 Ensembl
Affymetrix Human Clariom D Assay PSR2000149971.hg.1 ENST00000373946.7 Ensembl
Affymetrix Human Clariom D Assay PSR2000149971.hg.1 ENST00000373950.6 Ensembl
Affymetrix Human Clariom D Assay PSR2000149971.hg.1 ENST00000432603.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000149971.hg.1 ENST00000451082.5 Ensembl
Affymetrix Human Clariom D Assay PSR2000149971.hg.1 ENST00000454226.5 Ensembl
Affymetrix Human Clariom D Assay PSR2000149971.hg.1 ENST00000628415.2 Ensembl
Affymetrix Human Clariom D Assay PSR2000149971.hg.1 NM_001258329.1 Refseq
Affymetrix Human Clariom D Assay PSR2000149971.hg.1 NM_001258330.1 Refseq
Affymetrix Human Clariom D Assay PSR2000149971.hg.1 NM_012156.2 Refseq
Affymetrix Human Clariom D Assay PSR2000149971.hg.1 TC2000007239.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2000149971.hg.1 ENST00000338074.6 Ensembl
Affymetrix Human Clariom D Assay PSR2000149971.hg.1 ENST00000373941.5 Ensembl
Affymetrix Human Clariom D Assay PSR2000149971.hg.1 ENST00000373946.7 Ensembl
Affymetrix Human Clariom D Assay PSR2000149971.hg.1 ENST00000373950.6 Ensembl
Affymetrix Human Clariom D Assay PSR2000149971.hg.1 ENST00000432603.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000149971.hg.1 ENST00000451082.5 Ensembl
Affymetrix Human Clariom D Assay PSR2000149971.hg.1 ENST00000454226.5 Ensembl
Affymetrix Human Clariom D Assay PSR2000149971.hg.1 ENST00000628415.2 Ensembl
Affymetrix Human Clariom D Assay PSR2000149971.hg.1 NM_001258329.1 Refseq
Affymetrix Human Clariom D Assay PSR2000149971.hg.1 NM_001258330.1 Refseq
Affymetrix Human Clariom D Assay PSR2000149971.hg.1 NM_012156.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0