This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01104725_m1
See other ABL1 GE Assays ›
Gene Symbol:
ABL1
Gene Name:
ABL proto-oncogene 1, non-receptor tyrosine kinase
Gene Aliases:
ABL, JTK7, bcr/abl, c-ABL, c-ABL1, p150, v-abl
Chromosome Location:
Chr.9: 130713881 - 130887675 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01104725_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ABL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005157.5 NP_005148.2 5 - 6 1097 104
NM_007313.2 NP_009297.2 5 - 6 1401 104
GenBank mRNA AB209456.1 - 5 - 6 1107 104
AB209642.1 - 5 - 6 1329 104
BC117451.1 - 5 - 6 1027 40126002 104
FJ785401.1 - 2 - 3 139 104
JF272500.1 - 2 - 3 194 104
JX565024.1 - 2 - 3 248 104
JX565025.1 - 2 - 3 248 104
M14752.1 - 5 - 6 104
X16416.1 - 5 - 6 1052 104

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Protein Kinase Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ABL1

Gene Ontology Categories:

Function(s) Process(es)

B-1 B cell homeostasis
positive regulation of protein phosphorylation
B cell proliferation involved in immune response
transitional one stage B cell differentiation
mismatch repair
regulation of transcription, DNA-templated
cellular protein modification process
autophagy
cellular response to DNA damage stimulus
DNA damage induced protein phosphorylation
response to oxidative stress
cell cycle arrest
mitotic nuclear division
epidermal growth factor receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
intrinsic apoptotic signaling pathway in response to DNA damage
regulation of autophagy
peptidyl-tyrosine phosphorylation
cerebellum morphogenesis
negative regulation of cell-cell adhesion
microspike assembly
actin cytoskeleton organization
regulation of endocytosis
regulation of cell adhesion
negative regulation of BMP signaling pathway
regulation of axon extension
regulation of microtubule polymerization
regulation of actin cytoskeleton organization
positive regulation of osteoblast proliferation
substrate adhesion-dependent cell spreading
cellular response to oxidative stress
platelet-derived growth factor receptor-beta signaling pathway
peptidyl-tyrosine autophosphorylation
Fc-gamma receptor signaling pathway involved in phagocytosis
regulation of cell proliferation
signal transduction in response to DNA damage
positive regulation of apoptotic process
positive regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of I-kappaB kinase/NF-kappaB signaling
innate immune response
establishment of protein localization
negative regulation of mitotic cell cycle
positive regulation of mitotic cell cycle
alpha-beta T cell differentiation
protein autophosphorylation
spleen development
thymus development
collateral sprouting
positive regulation of peptidyl-tyrosine phosphorylation
activated T cell proliferation
B cell receptor signaling pathway
neuromuscular process controlling balance
positive regulation of muscle cell differentiation
positive regulation of release of sequestered calcium ion into cytosol
positive regulation of oxidoreductase activity
negative regulation of ubiquitin-protein transferase activity
mitochondrial depolarization
Bergmann glial cell differentiation
cellular response to hydrogen peroxide
negative regulation of ERK1 and ERK2 cascade
positive regulation of ERK1 and ERK2 cascade
cellular response to lipopolysaccharide
negative regulation of protein serine/threonine kinase activity
actin filament branching
positive regulation of interleukin-2 secretion
negative regulation of phospholipase C activity
positive regulation of neuron death
positive regulation of interferon-gamma secretion
regulation of extracellular matrix organization
cellular response to dopamine
positive regulation of microtubule binding
positive regulation of actin filament binding
activation of protein kinase C activity
positive regulation of Wnt signaling pathway, planar cell polarity pathway
regulation of cell motility
regulation of actin cytoskeleton reorganization
negative regulation of endothelial cell apoptotic process
negative regulation of cellular senescence
regulation of response to DNA damage stimulus
magnesium ion binding
DNA binding
actin monomer binding
nicotinate-nucleotide adenylyltransferase activity
protein kinase activity
protein tyrosine kinase activity
non-membrane spanning protein tyrosine kinase activity
protein kinase C binding
receptor binding
protein binding
ATP binding
protein C-terminus binding
SH3 domain binding
syntaxin binding
manganese ion binding
actin filament binding
mitogen-activated protein kinase binding
proline-rich region binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0