This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01104933_m1
See other GPR161 GE Assays ›
Gene Symbol:
GPR161
Gene Name:
G protein-coupled receptor 161
Gene Aliases:
RE2
Chromosome Location:
Chr.1: 168079542 - 168137667 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01104933_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPR161
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001267611.1 NP_001254540.1 1 - 2 185 105
GenBank mRNA AK310019.1 - 1 - 2 102 105

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100016322.hg.1

Additional Information:

This assay is in the 5'UTR region of the NM_001267611.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 G-protein coupled receptor Homo sapiens 1700 Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
multicellular organism development
positive regulation of cAMP biosynthetic process
negative regulation of smoothened signaling pathway involved in dorsal/ventral neural tube patterning
G-protein coupled receptor activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000271357.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000271357.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000271357.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000271357.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000271357.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000271357.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000271357.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000271357.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000271357.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000271357.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000271357.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000271357.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367835.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367835.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367835.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367835.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367835.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367835.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367835.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367835.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367835.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367835.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367835.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367836.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367836.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367836.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367836.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367836.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367836.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367836.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367836.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000478868.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000478868.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000478868.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000478868.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000485232.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000485232.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000485232.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000493800.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000493800.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000493800.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000493800.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000493800.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000537209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000537209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000537209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000537209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000537209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000537209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000537209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000537209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000537209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000537209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000537209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000537209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000537209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000537209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000537209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000539777.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000539777.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000539777.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000539777.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000539777.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000539777.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000539777.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000539777.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000539777.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000539777.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000539777.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000546300.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000546300.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000546300.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000546300.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000546300.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000546300.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000546300.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000546300.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000546300.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000546300.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267609.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267609.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267609.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267609.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267609.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267609.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267609.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267609.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267609.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267609.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267609.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267609.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267609.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267609.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267609.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267611.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267611.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267611.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267611.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267611.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267611.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267611.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267611.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267611.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267611.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267611.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267611.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267612.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267612.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267612.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267612.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267612.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267612.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267612.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267612.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267612.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267612.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267612.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267612.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267613.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267613.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267613.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267613.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267613.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267613.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267613.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267613.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267613.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267613.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267613.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267614.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267614.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267614.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267614.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267614.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267614.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267614.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267614.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267614.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267614.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_153832.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_153832.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_153832.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_153832.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_153832.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_153832.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_153832.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_153832.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_153832.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_153832.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_153832.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_153832.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_153832.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_153832.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_153832.2 Refseq
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 190566 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0