This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01104936_m1
See other GPR161 GE Assays ›
Gene Symbol:
GPR161
Gene Name:
G protein-coupled receptor 161
Gene Aliases:
RE2
Chromosome Location:
Chr.1: 168079542 - 168137667 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01104936_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPR161
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001267609.1 NP_001254538.1 5 - 6 1882 120
NM_001267610.1 NP_001254539.1 5 - 6 1845 120
NM_001267611.1 NP_001254540.1 4 - 5 1432 120
NM_001267612.1 NP_001254541.1 4 - 5 1388 120
NM_001267613.1 NP_001254542.1 3 - 4 1281 120
NM_001267614.1 NP_001254543.1 3 - 4 1014 120
NM_153832.2 NP_722561.1 6 - 7 1839 120
XM_005245055.2 XP_005245112.1 5 - 6 1806 120
XM_005245056.2 XP_005245113.1 4 - 5 1413 120
XM_005245057.4 XP_005245114.1 4 - 5 1430 120
XM_006711251.1 XP_006711314.1 6 - 7 1852 120
XM_006711253.2 XP_006711316.2 4 - 5 1645 120
XM_011509370.1 XP_011507672.1 6 - 7 2150 120
XM_011509371.2 XP_011507673.1 6 - 7 1596 120
XM_011509372.1 XP_011507674.1 6 - 7 1928 120
XM_011509373.2 XP_011507675.1 6 - 7 1854 120
XM_011509375.2 XP_011507677.1 6 - 7 1640 120
XM_011509376.2 XP_011507678.1 5 - 6 1771 120
XM_011509377.1 XP_011507679.1 4 - 5 1727 120
XM_011509378.1 XP_011507680.1 4 - 5 1699 120
XM_017000858.1 XP_016856347.1 5 - 6 2299 120
XM_017000859.1 XP_016856348.1 4 - 5 2253 120
XM_017000860.1 XP_016856349.1 5 - 6 2347 120
XM_017000861.1 XP_016856350.1 6 - 7 2393 120
XM_017000862.1 XP_016856351.1 6 - 7 1976 120
GenBank mRNA AK056040.1 - 4 - 5 1335 120
AK122656.1 - 5 - 6 1761 120
AK298797.1 - 3 - 4 1239 120
AK298850.1 - 3 - 4 1014 120
AK299634.1 - 5 - 6 1513 120
AK310019.1 - 4 - 5 1349 120
AY275468.1 - 3 - 4 1204 120
BC028163.1 - 6 - 7 1518 5246359 120

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100016322.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GPR161

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
multicellular organism development
positive regulation of cAMP biosynthetic process
negative regulation of smoothened signaling pathway involved in dorsal/ventral neural tube patterning
G-protein coupled receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000271357.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000271357.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000271357.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000271357.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000271357.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000271357.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000271357.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000271357.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000271357.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000271357.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000271357.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000271357.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367835.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367835.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367835.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367835.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367835.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367835.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367835.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367835.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367835.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367835.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367835.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367836.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367836.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367836.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367836.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367836.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367836.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367836.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367836.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000478868.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000478868.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000478868.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000478868.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000485232.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000485232.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000485232.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000493800.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000493800.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000493800.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000493800.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000493800.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000537209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000537209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000537209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000537209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000537209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000537209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000537209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000537209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000537209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000537209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000537209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000537209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000537209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000537209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000537209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000539777.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000539777.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000539777.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000539777.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000539777.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000539777.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000539777.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000539777.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000539777.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000539777.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000539777.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000546300.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000546300.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000546300.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000546300.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000546300.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000546300.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000546300.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000546300.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000546300.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000546300.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267609.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267609.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267609.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267609.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267609.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267609.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267609.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267609.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267609.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267609.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267609.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267609.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267609.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267609.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267609.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267611.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267611.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267611.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267611.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267611.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267611.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267611.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267611.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267611.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267611.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267611.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267611.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267612.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267612.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267612.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267612.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267612.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267612.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267612.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267612.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267612.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267612.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267612.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267612.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267613.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267613.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267613.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267613.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267613.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267613.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267613.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267613.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267613.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267613.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267613.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267614.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267614.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267614.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267614.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267614.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267614.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267614.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267614.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267614.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267614.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_153832.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_153832.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_153832.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_153832.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_153832.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_153832.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_153832.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_153832.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_153832.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_153832.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_153832.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_153832.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_153832.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_153832.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_153832.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rzthw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0