This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01104937_m1
See other GPR161 GE Assays ›
Gene Symbol:
GPR161
Gene Name:
G protein-coupled receptor 161
Gene Aliases:
RE2
Chromosome Location:
Chr.1: 168079542 - 168137667 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01104937_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPR161
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001267609.1 NP_001254538.1 6 - 7 2008 88
NM_001267610.1 NP_001254539.1 6 - 7 1971 88
NM_001267611.1 NP_001254540.1 5 - 6 1558 88
NM_001267612.1 NP_001254541.1 5 - 6 1514 88
NM_001267613.1 NP_001254542.1 4 - 5 1407 88
NM_001267614.1 NP_001254543.1 4 - 5 1140 88
NM_153832.2 NP_722561.1 7 - 8 1965 88
XM_005245055.2 XP_005245112.1 6 - 7 1932 88
XM_005245056.2 XP_005245113.1 5 - 6 1539 88
XM_005245057.4 XP_005245114.1 5 - 6 1556 88
XM_006711251.1 XP_006711314.1 7 - 8 1978 88
XM_006711253.2 XP_006711316.2 5 - 6 1771 88
XM_011509370.1 XP_011507672.1 7 - 8 2276 88
XM_011509371.2 XP_011507673.1 7 - 8 1722 88
XM_011509372.1 XP_011507674.1 7 - 8 2054 88
XM_011509373.2 XP_011507675.1 7 - 8 1980 88
XM_011509375.2 XP_011507677.1 7 - 8 1766 88
XM_011509376.2 XP_011507678.1 6 - 7 1897 88
XM_011509377.1 XP_011507679.1 5 - 6 1853 88
XM_011509378.1 XP_011507680.1 5 - 6 1825 88
XM_017000858.1 XP_016856347.1 6 - 7 2425 88
XM_017000859.1 XP_016856348.1 5 - 6 2379 88
XM_017000860.1 XP_016856349.1 6 - 7 2473 88
XM_017000861.1 XP_016856350.1 7 - 8 2519 88
XM_017000862.1 XP_016856351.1 7 - 8 2102 88
GenBank mRNA AK056040.1 - 5 - 6 1461 88
AK091869.1 - 1 - 2 1986 88
AK122656.1 - 6 - 7 1887 88
AK298797.1 - 4 - 5 1365 88
AK298850.1 - 4 - 5 1140 88
AK299634.1 - 6 - 7 1639 88
AK310019.1 - 5 - 6 1475 88
AY275468.1 - 4 - 5 1330 88
BC028163.1 - 7 - 8 1644 5246359 88

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100016322.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01003485.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GPR161

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
multicellular organism development
positive regulation of cAMP biosynthetic process
negative regulation of smoothened signaling pathway involved in dorsal/ventral neural tube patterning
G-protein coupled receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000271357.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000271357.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000271357.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000271357.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000271357.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000271357.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000271357.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000271357.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000271357.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000271357.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000271357.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000271357.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367835.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367835.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367835.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367835.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367835.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367835.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367835.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367835.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367835.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367835.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367835.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367836.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367836.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367836.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367836.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367836.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367836.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367836.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367836.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000367838.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000478868.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000478868.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000478868.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000478868.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000485232.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000485232.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000485232.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000493800.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000493800.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000493800.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000493800.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000493800.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000537209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000537209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000537209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000537209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000537209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000537209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000537209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000537209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000537209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000537209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000537209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000537209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000537209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000537209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000537209.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000539777.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000539777.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000539777.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000539777.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000539777.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000539777.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000539777.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000539777.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000539777.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000539777.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000539777.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000546300.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000546300.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000546300.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000546300.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000546300.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000546300.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000546300.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000546300.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000546300.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 ENST00000546300.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267609.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267609.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267609.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267609.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267609.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267609.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267609.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267609.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267609.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267609.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267609.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267609.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267609.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267609.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267609.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267610.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267611.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267611.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267611.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267611.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267611.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267611.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267611.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267611.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267611.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267611.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267611.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267611.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267612.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267612.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267612.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267612.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267612.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267612.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267612.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267612.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267612.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267612.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267612.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267612.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267613.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267613.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267613.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267613.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267613.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267613.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267613.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267613.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267613.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267613.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267613.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267614.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267614.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267614.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267614.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267614.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267614.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267614.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267614.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267614.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_001267614.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_153832.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_153832.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_153832.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_153832.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_153832.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_153832.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_153832.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_153832.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_153832.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_153832.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_153832.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_153832.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_153832.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_153832.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016322.hg.1 NM_153832.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0