This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01105483_g1
See other GLB1L GE Assays ›
Gene Symbol:
GLB1L
Gene Name:
galactosidase beta 1 like
Gene Aliases:
-
Chromosome Location:
Chr.2: 219236598 - 219245429 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01105483_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GLB1L
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001286423.1 NP_001273352.1 10 - 11 1220 78
NM_001286427.1 NP_001273356.1 7 - 8 737 78
NM_024506.4 NP_078782.3 10 - 11 1208 78
XM_011511813.2 XP_011510115.1 8 - 9 797 78
XM_017004895.1 XP_016860384.1 8 - 9 904 78
GenBank mRNA AK055098.1 - 5 - 6 395 78
AK074393.1 - 4 - 5 388 78
AK299404.1 - 9 - 10 969 78
AK300215.1 - 7 - 8 720 78
AY358639.1 - 10 - 11 1188 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200015770.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GLB1L

Panther Classification:

Molecular Function -

galactosidase

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
beta-galactosidase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Clariom D Assay PSR0200215997.hg.1 ENST00000295759.11 Ensembl
Affymetrix Human Clariom D Assay PSR0200215997.hg.1 ENST00000392089.6 Ensembl
Affymetrix Human Clariom D Assay PSR0200215997.hg.1 ENST00000409640.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200215997.hg.1 ENST00000497855.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200215997.hg.1 NM_001286423.1 Refseq
Affymetrix Human Clariom D Assay PSR0200215997.hg.1 NM_001286427.1 Refseq
Affymetrix Human Clariom D Assay PSR0200215997.hg.1 NM_024506.4 Refseq
Affymetrix Human Clariom D Assay PSR0200215997.hg.1 ENST00000295759.11 Ensembl
Affymetrix Human Clariom D Assay PSR0200215997.hg.1 ENST00000392089.6 Ensembl
Affymetrix Human Clariom D Assay PSR0200215997.hg.1 ENST00000409640.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200215997.hg.1 ENST00000497855.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200215997.hg.1 NM_001286423.1 Refseq
Affymetrix Human Clariom D Assay PSR0200215997.hg.1 NM_001286427.1 Refseq
Affymetrix Human Clariom D Assay PSR0200215997.hg.1 NM_024506.4 Refseq
Affymetrix Human Clariom D Assay PSR0200215997.hg.1 TC0200015770.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline