This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01106050_g1
See other UCP3 GE Assays ›
Gene Symbol:
UCP3
Gene Name:
uncoupling protein 3
Gene Aliases:
SLC25A9
Chromosome Location:
Chr.11: 74000281 - 74009237 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01106050_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UCP3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_003356.3 NP_003347.1 2 - 3 481 56
NM_022803.2 NP_073714.1 2 - 3 481 56
GenBank mRNA AF001787.1 - 2 - 3 309 56
AF011449.1 - 1 - 2 224 56
AK290630.1 - 2 - 3 329 56
AK291889.1 - 2 - 3 295 56
AK309873.1 - 2 - 3 329 56
BC008392.1 - 2 - 3 329 4291158 56
KU178371.1 - 1 - 2 126 56
KU178372.1 - 1 - 2 126 56
U82818.1 - 2 - 3 279 56
U84763.1 - 2 - 3 279 56

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100011605.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11002075.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene UCP3

Panther Classification:

Molecular Function -

transporter secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

response to superoxide
response to hypoxia
translation
lipid metabolic process
fatty acid metabolic process
mitochondrial transport
aging
response to nutrient
respiratory gaseous exchange
response to cold
response to activity
proton transport
response to insulin
cellular response to hormone stimulus
response to glucocorticoid
structural constituent of ribosome
transporter activity
protein binding
oxidative phosphorylation uncoupler activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR11024867.hg.1 ENST00000314032.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11024867.hg.1 ENST00000348534.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11024867.hg.1 ENST00000426995.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11024867.hg.1 NM_003356.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11024867.hg.1 NM_022803.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11024867.hg.1 TC11002075.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000426995.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000426995.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000426995.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000426995.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000426995.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000426995.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000426995.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000426995.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000426995.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000544614.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000544614.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000544614.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000544614.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000545271.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000545271.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 NM_003356.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 NM_003356.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 NM_003356.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 NM_003356.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 NM_003356.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 NM_003356.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 NM_003356.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 NM_003356.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 NM_003356.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 NM_003356.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 NM_003356.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 NM_003356.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 NM_022803.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 NM_022803.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 NM_022803.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 NM_022803.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 NM_022803.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 NM_022803.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 NM_022803.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 NM_022803.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 NM_022803.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 NM_022803.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 NM_022803.2 Refseq
Affymetrix Human Clariom D Assay PSR1100189510.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Clariom D Assay PSR1100189510.hg.1 ENST00000426995.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100189510.hg.1 ENST00000544614.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100189510.hg.1 NM_003356.3 Refseq
Affymetrix Human Clariom D Assay PSR1100189510.hg.1 NM_022803.2 Refseq
Affymetrix Human Clariom D Assay PSR1100189510.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Clariom D Assay PSR1100189510.hg.1 ENST00000426995.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100189510.hg.1 ENST00000544614.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100189510.hg.1 NM_003356.3 Refseq
Affymetrix Human Clariom D Assay PSR1100189510.hg.1 NM_022803.2 Refseq
Affymetrix Human Clariom D Assay PSR1100189510.hg.1 TC1100011605.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000426995.2 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000426995.2 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000426995.2 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000426995.2 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000426995.2 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000426995.2 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000426995.2 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000426995.2 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000426995.2 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000544614.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000544614.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000544614.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000544614.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000545271.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000545271.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 NM_003356.3 Refseq
Affymetrix Human Clariom D Assay TC1100011605.hg.1 NM_003356.3 Refseq
Affymetrix Human Clariom D Assay TC1100011605.hg.1 NM_003356.3 Refseq
Affymetrix Human Clariom D Assay TC1100011605.hg.1 NM_003356.3 Refseq
Affymetrix Human Clariom D Assay TC1100011605.hg.1 NM_003356.3 Refseq
Affymetrix Human Clariom D Assay TC1100011605.hg.1 NM_003356.3 Refseq
Affymetrix Human Clariom D Assay TC1100011605.hg.1 NM_003356.3 Refseq
Affymetrix Human Clariom D Assay TC1100011605.hg.1 NM_003356.3 Refseq
Affymetrix Human Clariom D Assay TC1100011605.hg.1 NM_003356.3 Refseq
Affymetrix Human Clariom D Assay TC1100011605.hg.1 NM_003356.3 Refseq
Affymetrix Human Clariom D Assay TC1100011605.hg.1 NM_003356.3 Refseq
Affymetrix Human Clariom D Assay TC1100011605.hg.1 NM_003356.3 Refseq
Affymetrix Human Clariom D Assay TC1100011605.hg.1 NM_022803.2 Refseq
Affymetrix Human Clariom D Assay TC1100011605.hg.1 NM_022803.2 Refseq
Affymetrix Human Clariom D Assay TC1100011605.hg.1 NM_022803.2 Refseq
Affymetrix Human Clariom D Assay TC1100011605.hg.1 NM_022803.2 Refseq
Affymetrix Human Clariom D Assay TC1100011605.hg.1 NM_022803.2 Refseq
Affymetrix Human Clariom D Assay TC1100011605.hg.1 NM_022803.2 Refseq
Affymetrix Human Clariom D Assay TC1100011605.hg.1 NM_022803.2 Refseq
Affymetrix Human Clariom D Assay TC1100011605.hg.1 NM_022803.2 Refseq
Affymetrix Human Clariom D Assay TC1100011605.hg.1 NM_022803.2 Refseq
Affymetrix Human Clariom D Assay TC1100011605.hg.1 NM_022803.2 Refseq
Affymetrix Human Clariom D Assay TC1100011605.hg.1 NM_022803.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rr9bx:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0