This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01106051_g1
See other UCP3 GE Assays ›
Gene Symbol:
UCP3
Gene Name:
uncoupling protein 3
Gene Aliases:
SLC25A9
Chromosome Location:
Chr.11: 74000281 - 74009237 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01106051_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UCP3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_003356.3 NP_003347.1 3 - 4 686 88
NM_022803.2 NP_073714.1 3 - 4 686 88
GenBank mRNA AF001787.1 - 3 - 4 514 88
AF011449.1 - 2 - 3 429 88
AK092685.1 - 3 - 4 333 88
AK291889.1 - 3 - 4 500 88
AK309873.1 - 3 - 4 534 88
KU178372.1 - 2 - 3 331 88
U82818.1 - 3 - 4 484 88
U84763.1 - 3 - 4 484 88

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100011605.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11002075.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene UCP3

Panther Classification:

Molecular Function -

transporter secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

response to superoxide
response to hypoxia
translation
lipid metabolic process
fatty acid metabolic process
mitochondrial transport
aging
response to nutrient
respiratory gaseous exchange
response to cold
response to activity
proton transport
response to insulin
cellular response to hormone stimulus
response to glucocorticoid
structural constituent of ribosome
transporter activity
protein binding
oxidative phosphorylation uncoupler activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR11024866.hg.1 ENST00000314032.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11024866.hg.1 ENST00000426995.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11024866.hg.1 NM_003356.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11024866.hg.1 NM_022803.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11024866.hg.1 TC11002075.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000426995.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000426995.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000426995.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000426995.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000426995.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000426995.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000426995.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000426995.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000426995.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000544614.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000544614.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000544614.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000544614.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000545271.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 ENST00000545271.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 NM_003356.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 NM_003356.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 NM_003356.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 NM_003356.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 NM_003356.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 NM_003356.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 NM_003356.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 NM_003356.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 NM_003356.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 NM_003356.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 NM_003356.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 NM_003356.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 NM_022803.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 NM_022803.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 NM_022803.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 NM_022803.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 NM_022803.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 NM_022803.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 NM_022803.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 NM_022803.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 NM_022803.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 NM_022803.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011605.hg.1 NM_022803.2 Refseq
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000426995.2 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000426995.2 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000426995.2 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000426995.2 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000426995.2 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000426995.2 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000426995.2 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000426995.2 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000426995.2 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000544614.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000544614.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000544614.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000544614.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000545271.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 ENST00000545271.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011605.hg.1 NM_003356.3 Refseq
Affymetrix Human Clariom D Assay TC1100011605.hg.1 NM_003356.3 Refseq
Affymetrix Human Clariom D Assay TC1100011605.hg.1 NM_003356.3 Refseq
Affymetrix Human Clariom D Assay TC1100011605.hg.1 NM_003356.3 Refseq
Affymetrix Human Clariom D Assay TC1100011605.hg.1 NM_003356.3 Refseq
Affymetrix Human Clariom D Assay TC1100011605.hg.1 NM_003356.3 Refseq
Affymetrix Human Clariom D Assay TC1100011605.hg.1 NM_003356.3 Refseq
Affymetrix Human Clariom D Assay TC1100011605.hg.1 NM_003356.3 Refseq
Affymetrix Human Clariom D Assay TC1100011605.hg.1 NM_003356.3 Refseq
Affymetrix Human Clariom D Assay TC1100011605.hg.1 NM_003356.3 Refseq
Affymetrix Human Clariom D Assay TC1100011605.hg.1 NM_003356.3 Refseq
Affymetrix Human Clariom D Assay TC1100011605.hg.1 NM_003356.3 Refseq
Affymetrix Human Clariom D Assay TC1100011605.hg.1 NM_022803.2 Refseq
Affymetrix Human Clariom D Assay TC1100011605.hg.1 NM_022803.2 Refseq
Affymetrix Human Clariom D Assay TC1100011605.hg.1 NM_022803.2 Refseq
Affymetrix Human Clariom D Assay TC1100011605.hg.1 NM_022803.2 Refseq
Affymetrix Human Clariom D Assay TC1100011605.hg.1 NM_022803.2 Refseq
Affymetrix Human Clariom D Assay TC1100011605.hg.1 NM_022803.2 Refseq
Affymetrix Human Clariom D Assay TC1100011605.hg.1 NM_022803.2 Refseq
Affymetrix Human Clariom D Assay TC1100011605.hg.1 NM_022803.2 Refseq
Affymetrix Human Clariom D Assay TC1100011605.hg.1 NM_022803.2 Refseq
Affymetrix Human Clariom D Assay TC1100011605.hg.1 NM_022803.2 Refseq
Affymetrix Human Clariom D Assay TC1100011605.hg.1 NM_022803.2 Refseq
Affymetrix Human Clariom D Assay PSR1100189508.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Clariom D Assay PSR1100189508.hg.1 ENST00000426995.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100189508.hg.1 NM_003356.3 Refseq
Affymetrix Human Clariom D Assay PSR1100189508.hg.1 NM_022803.2 Refseq
Affymetrix Human Clariom D Assay PSR1100189508.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Clariom D Assay PSR1100189508.hg.1 ENST00000426995.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100189508.hg.1 NM_003356.3 Refseq
Affymetrix Human Clariom D Assay PSR1100189508.hg.1 NM_022803.2 Refseq
Affymetrix Human Clariom D Assay PSR1100189508.hg.1 TC1100011605.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1100189509.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Clariom D Assay PSR1100189509.hg.1 ENST00000426995.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100189509.hg.1 NM_003356.3 Refseq
Affymetrix Human Clariom D Assay PSR1100189509.hg.1 NM_022803.2 Refseq
Affymetrix Human Clariom D Assay PSR1100189509.hg.1 ENST00000314032.8 Ensembl
Affymetrix Human Clariom D Assay PSR1100189509.hg.1 ENST00000426995.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100189509.hg.1 NM_003356.3 Refseq
Affymetrix Human Clariom D Assay PSR1100189509.hg.1 NM_022803.2 Refseq
Affymetrix Human Clariom D Assay PSR1100189509.hg.1 TC1100011605.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0