This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01106208_m1
See other PTPN13 GE Assays ›
Gene Symbol:
PTPN13
Gene Name:
protein tyrosine phosphatase, non-receptor type 13
Gene Aliases:
FAP-1, PNP1, PTP-BAS, PTP-BL, PTP1E, PTPL1, PTPLE, hPTP1E
Chromosome Location:
Chr.4: 86594307 - 86815179 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01106208_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PTPN13
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_006264.2 NP_006255.1 24 - 25 4574 95
NM_080683.2 NP_542414.1 25 - 26 4631 95
NM_080684.2 NP_542415.1 22 - 23 4058 95
XM_017008511.1 XP_016864000.1 25 - 26 4639 95
XM_017008513.1 XP_016864002.1 24 - 25 4582 95
GenBank mRNA AB208904.1 - 23 - 24 3998 95
AF233323.1 - 2 - 3 185 95
BC039610.1 - 2 - 3 185 95
BC140777.1 - 23 - 24 4113 9021645 95
D21209.1 - 25 - 26 4214 95
D21210.1 - 24 - 25 4157 95
D21211.1 - 22 - 23 3641 95
X80289.1 - 24 - 25 4171 95

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Protein Phosphatase Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PTPN13

Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
regulation of phosphatidylinositol 3-kinase signaling
peptidyl-tyrosine dephosphorylation
protein tyrosine phosphatase activity
protein binding
phosphatidylinositol 3-kinase regulatory subunit binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline