This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01106269_g1
See other ATP5C1 GE Assays ›
Gene Symbol:
ATP5C1
Gene Name:
ATP synthase, H+ transporting, mitochondrial F1 complex, gamma polypeptide 1
Gene Aliases:
ATP5C, ATP5CL1
Chromosome Location:
Chr.10: 7788104 - 7807801 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01106269_g1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATP5C1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001001973.2 NP_001001973.1 9 - 10 1029 105
NM_001320886.1 NP_001307815.1 8 - 9 994 105
GenBank mRNA BC000470.2 - 9 - 10 944 2961142 105
BC000931.3 - 9 - 10 977 3445817 105
BC013394.2 - 9 - 10 944 2961142 105
BC013865.1 - 9 - 10 942 105
BC016812.1 - 9 - 10 959 4092259 105
BC020824.1 - 9 - 10 965 4733791 105
BC026049.1 - 9 - 10 904 105
D16562.1 - 9 - 10 956 105

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000006703.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10000067.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Transporter 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ATP5C1

Panther Classification:

Molecular Function -

ATP synthase

Gene Ontology Categories:

Function(s) Process(es)

oxidative phosphorylation
ATP biosynthetic process
mitochondrial ATP synthesis coupled proton transport
ATPase activity
transmembrane transporter activity
poly(A) RNA binding
proton-transporting ATP synthase activity, rotational mechanism
proton-transporting ATPase activity, rotational mechanism

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR10000764.hg.1 ENST00000335698.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10000764.hg.1 ENST00000356708.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10000764.hg.1 ENST00000493053.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10000764.hg.1 NM_001001973.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10000764.hg.1 NM_005174.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10000764.hg.1 TC10000067.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1000145991.hg.1 ENST00000335698.4 Ensembl
Affymetrix Human Clariom D Assay PSR1000145991.hg.1 ENST00000356708.11 Ensembl
Affymetrix Human Clariom D Assay PSR1000145991.hg.1 ENST00000465936.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000145991.hg.1 ENST00000473809.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000145991.hg.1 ENST00000480528.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000145991.hg.1 ENST00000493053.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000145991.hg.1 NM_001001973.2 Refseq
Affymetrix Human Clariom D Assay PSR1000145991.hg.1 NM_005174.3 Refseq
Affymetrix Human Clariom D Assay PSR1000145991.hg.1 ENST00000335698.4 Ensembl
Affymetrix Human Clariom D Assay PSR1000145991.hg.1 ENST00000356708.11 Ensembl
Affymetrix Human Clariom D Assay PSR1000145991.hg.1 ENST00000465936.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000145991.hg.1 ENST00000473809.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000145991.hg.1 ENST00000480528.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000145991.hg.1 ENST00000493053.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000145991.hg.1 NM_001001973.2 Refseq
Affymetrix Human Clariom D Assay PSR1000145991.hg.1 NM_005174.3 Refseq
Affymetrix Human Clariom D Assay PSR1000145991.hg.1 TC1000006703.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0