This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01106651_s1
See other CPTP GE Assays ›
Gene Symbol:
CPTP
Gene Name:
ceramide-1-phosphate transfer protein
Gene Aliases:
GLTPD1
Chromosome Location:
Chr.1: 1324756 - 1328896 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01106651_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CPTP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001029885.1 NP_001025056.1 3 2123 73
XM_005244801.3 XP_005244858.1 3 2236 73
XM_005244802.1 XP_005244859.1 2 1933 73
XM_011542200.2 XP_011540502.1 3 2132 73
GenBank mRNA AL122073.1 - 1 1231 73
BC004366.1 - 1 1134 73
BC098429.1 - 1 1547 73

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CPTP

Panther Classification:

Molecular Function -

transfer/carrier protein

Gene Ontology Categories:

Function(s) Process(es)

glycosphingolipid metabolic process
glycolipid transport
ceramide 1-phosphate transport
phospholipid binding
phospholipid transporter activity
glycolipid binding
ceramide 1-phosphate binding
ceramide 1-phosphate transporter activity

MicroArray Probes:

Agilent Affymetrix ArrayStar ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 260233 - -
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 449577 - -
Agilent SurePrint G3 Human CNV Microarray 2x400K 96770 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_23_P63281 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3266396 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_24_P225518 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3343220 - -
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P234105 - -
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P225518 - -
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P63281 - -
Agilent Whole Human Genome Oligo Microarray 4x44K 27652 - -
Agilent Whole Human Genome Oligo Microarray 4x44K 18946 - -
Agilent Whole Human Genome Oligo Microarray 4x44K 7698 - -
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 53625 - -
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 58788 - -
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 875 - -
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 45820 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_23_P63281 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3343220 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3266396 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_24_P225518 - -
SurePrint G3 Human GE Microarray G4851A A_23_P63281 - -
SurePrint G3 Human GE Microarray G4851A A_33_P3266396 - -
SurePrint G3 Human GE Microarray G4851A A_33_P3343220 - -
SurePrint G3 Human GE Microarray G4851A A_24_P225518 - -
Whole Human Genome Oligo Microarray G4112A A_23_P63281 - -
Whole Human Genome Oligo Microarray G4112A A_24_P225518 - -
Whole Human Genome Oligo Microarray G4112A A_24_P234105 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P00001794 - -
Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P63281 - -
Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P234105 - -
Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P225518 - -
Agilent Human 2 cDNA Microarray (G4101A) 1676226 - -
Agilent Human 2 cDNA Microarray (G4101A) 1676226 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Gene 1.0 ST Array NM_001029885_at - -
Affymetrix Human Gene 1.0 ST Array 8039938 - -
Affymetrix Human Gene 1.0 ST Array 7896918 - -
Affymetrix Human Gene 1.0 ST Array 7896919 - -
Affymetrix Human Gene 1.0 ST Array 7896920 - -
Affymetrix Human Gene 1.1 ST Array 8039937 - -
Affymetrix Human Gene 1.1 ST Array 7896917 - -
Affymetrix HT HG-U133+ PM Array Plate 220734_PM_s_at - -
Affymetrix Human Genome U219 Array 11724058_PM_x_at - -
Affymetrix Human Genome U219 Array 11724058_x_at - -
Human Genome U133 Plus 2.0 Array 220734_PM_s_at - -
Human Genome U133 Plus 2.0 Array 220734_s_at - -
Human Genome U133A 2.0 Array 220734_PM_s_at - -
Human Genome U133A 2.0 Array 220734_s_at - -
Human Genome U95 Set 90503_PM_at - -
Human Genome U95 Set 73257_at - -
Human Genome U95 Set 90503_at - -
Human Genome U95 Set 73257_PM_at - -
Human Genome U133 Set 220734_PM_s_at - -
Human Genome U133 Set 220734_s_at - -
Array Name Probe IDs Targets Target Type
Arraystar Human LncRNA microarray V2.0 ASHG19A3A014297 - -
Arraystar Human LncRNA microarray V2.0 ASHG19A3A014297 - -
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 151262 - -
ABI Human Genome Survey Microarray Version 2.0 178690 - -
ABI Human Genome Survey Microarray Version 2.0 215822 - -
Array Name Probe IDs Targets Target Type
HumanHT-12 v4 Expression BeadChip ILMN_2234310 CR599582.1 human_transcriptome
HumanHT-12 v4 Expression BeadChip ILMN_2234310 BC098429.1 human_transcriptome
HumanHT-12 v4 Expression BeadChip ILMN_2234310 AL122073.1 human_transcriptome
HumanHT-12 v4 Expression BeadChip ILMN_2234310 BC004366.1 human_transcriptome
HumanHT-12 v4 Expression BeadChip ILMN_2234310 NM_001029885.1 human_transcriptome
Illumina HumanWG-6 v2.0 expression beadchip ILMN_1681325 CR599582.1 human_transcriptome
Illumina HumanWG-6 v2.0 expression beadchip ILMN_1681325 BC098429.1 human_transcriptome
Illumina HumanWG-6 v2.0 expression beadchip ILMN_1681325 AL122073.1 human_transcriptome
Illumina HumanWG-6 v2.0 expression beadchip ILMN_1681325 BC004366.1 human_transcriptome
Illumina HumanWG-6 v2.0 expression beadchip ILMN_1681325 NM_001029885.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_2234310 CR599582.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_2234310 BC098429.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_2234310 AL122073.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_2234310 BC004366.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_2234310 NM_001029885.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2234310 CR599582.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2234310 BC098429.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2234310 AL122073.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2234310 BC004366.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2234310 NM_001029885.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2234310 CR599582.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2234310 BC098429.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2234310 AL122073.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2234310 BC004366.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2234310 NM_001029885.1 human_transcriptome
Illumina HumanRef-8 v2 Expression BeadChip GI_4647-S - -
Illumina Sentrix Human-6 v2 Expression BeadChip ILMN_4647 - -
Illumina HumanRef-6 v2.0 expression beadchip ILMN_4647 - -
Illumina HumanRef-8 v3.0 expression beadchip ILMN_2234310 CR599582.1 human_transcriptome
Illumina HumanRef-8 v3.0 expression beadchip ILMN_2234310 BC098429.1 human_transcriptome
Illumina HumanRef-8 v3.0 expression beadchip ILMN_2234310 AL122073.1 human_transcriptome
Illumina HumanRef-8 v3.0 expression beadchip ILMN_2234310 BC004366.1 human_transcriptome
Illumina HumanRef-8 v3.0 expression beadchip ILMN_2234310 NM_001029885.1 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_1681325 CR599582.1 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_1681325 BC098429.1 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_1681325 AL122073.1 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_1681325 BC004366.1 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_1681325 NM_001029885.1 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_2234310 CR599582.1 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_2234310 BC098429.1 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_2234310 AL122073.1 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_2234310 BC004366.1 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_2234310 NM_001029885.1 human_transcriptome
Illumina HumanHT-12 V3.0 expression beadchip ILMN_2234310 CR599582.1 human_transcriptome
Illumina HumanHT-12 V3.0 expression beadchip ILMN_2234310 BC098429.1 human_transcriptome
Illumina HumanHT-12 V3.0 expression beadchip ILMN_2234310 AL122073.1 human_transcriptome
Illumina HumanHT-12 V3.0 expression beadchip ILMN_2234310 BC004366.1 human_transcriptome
Illumina HumanHT-12 V3.0 expression beadchip ILMN_2234310 NM_001029885.1 human_transcriptome
Illumina HumanRef-8 WG-DASL v3.0 ILMN_2234310 CR599582.1 human_transcriptome
Illumina HumanRef-8 WG-DASL v3.0 ILMN_2234310 BC098429.1 human_transcriptome
Illumina HumanRef-8 WG-DASL v3.0 ILMN_2234310 AL122073.1 human_transcriptome
Illumina HumanRef-8 WG-DASL v3.0 ILMN_2234310 BC004366.1 human_transcriptome
Illumina HumanRef-8 WG-DASL v3.0 ILMN_2234310 NM_001029885.1 human_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0