This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01106985_m1
See other NDUFB6 GE Assays ›
Gene Symbol:
NDUFB6
Gene Name:
NADH:ubiquinone oxidoreductase subunit B6
Gene Aliases:
B17, CI
Chromosome Location:
Chr.9: 32553526 - 32573184 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01106985_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NDUFB6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_002493.4 NP_002484.1 2 - 3 120
GenBank mRNA AF035840.1 - 2 - 3 120
AF067167.1 - 2 - 3 120
AK289702.1 - 2 - 3 120
BC009801.1 - 2 - 3 4334414 120
BC016708.1 - 2 - 3 120
BC029247.1 - 2 - 3 5171627 120
BP208846.1 - 2 - 3 120
BU952486.1 - 2 - 3 466 120
CR456920.1 - 2 - 3 120

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0900009829.hg.1
This assay targets the same location as HTA transcript cluster(s) TC09001001.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NDUFB6

Panther Classification:

Molecular Function -

dehydrogenase oxidoreductase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

mitochondrial electron transport, NADH to ubiquinone
mitochondrial respiratory chain complex I assembly
NADH dehydrogenase (ubiquinone) activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200013262.hg.1 ENST00000605302.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09013098.hg.1 TC09001001.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR09013098.hg.1 ENST00000379847.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09013098.hg.1 NM_001199987.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09013098.hg.1 NM_002493.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000350021.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000350021.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000350021.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000350021.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000350021.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000350021.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000350021.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000366466.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000366466.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000366466.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000366466.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000366466.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000366466.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000366466.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000366466.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000379847.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000379847.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000379847.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000379847.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000379847.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000379847.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000379847.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000379847.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000379847.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 ENST00000379847.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_001199987.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_001199987.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_001199987.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_001199987.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_001199987.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_001199987.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_001199987.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_001199987.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_002493.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_002493.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_002493.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_002493.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_002493.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_002493.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_002493.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_002493.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_002493.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_182739.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_182739.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_182739.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_182739.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_182739.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_182739.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_182739.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009829.hg.1 NM_182739.2 Refseq
Affymetrix Human Clariom D Assay PSR0900167206.hg.1 ENST00000366466.5 Ensembl
Affymetrix Human Clariom D Assay PSR0900167206.hg.1 ENST00000379847.7 Ensembl
Affymetrix Human Clariom D Assay PSR0900167206.hg.1 NM_001199987.1 Refseq
Affymetrix Human Clariom D Assay PSR0900167206.hg.1 NM_002493.4 Refseq
Affymetrix Human Clariom D Assay PSR0900167206.hg.1 ENST00000366466.5 Ensembl
Affymetrix Human Clariom D Assay PSR0900167206.hg.1 ENST00000379847.7 Ensembl
Affymetrix Human Clariom D Assay PSR0900167206.hg.1 NM_001199987.1 Refseq
Affymetrix Human Clariom D Assay PSR0900167206.hg.1 NM_002493.4 Refseq
Affymetrix Human Clariom D Assay PSR0900167206.hg.1 TC0900009829.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0200013262.hg.1 ENST00000605302.1 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000350021.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000350021.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000350021.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000350021.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000350021.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000350021.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000350021.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000366466.5 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000366466.5 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000366466.5 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000366466.5 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000366466.5 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000366466.5 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000366466.5 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000366466.5 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000379847.7 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000379847.7 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000379847.7 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000379847.7 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000379847.7 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000379847.7 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000379847.7 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000379847.7 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000379847.7 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 ENST00000379847.7 Ensembl
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_001199987.1 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_001199987.1 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_001199987.1 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_001199987.1 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_001199987.1 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_001199987.1 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_001199987.1 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_001199987.1 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_002493.4 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_002493.4 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_002493.4 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_002493.4 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_002493.4 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_002493.4 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_002493.4 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_002493.4 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_002493.4 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_182739.2 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_182739.2 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_182739.2 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_182739.2 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_182739.2 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_182739.2 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_182739.2 Refseq
Affymetrix Human Clariom D Assay TC0900009829.hg.1 NM_182739.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0