This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01107135_m1
See other RPIA GE Assays ›
Gene Symbol:
RPIA
Gene Name:
ribose 5-phosphate isomerase A
Gene Aliases:
RPI, RPIAD
Chromosome Location:
Chr.2: 88691658 - 88750935 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01107135_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RPIA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_144563.2 NP_653164.2 5 - 6 569 86
GenBank mRNA AY050633.1 - 5 - 6 545 86
BC015529.1 - 5 - 6 541 3904718 86

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200008351.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02000546.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RPIA

Panther Classification:

Molecular Function -

isomerase

Gene Ontology Categories:

Function(s) Process(es)

pentose-phosphate shunt
pentose-phosphate shunt, non-oxidative branch
ribose-5-phosphate isomerase activity
protein binding
monosaccharide binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR02008452.hg.1 ENST00000283646.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02008452.hg.1 ENST00000543560.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02008452.hg.1 NM_144563.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02008452.hg.1 TC02000546.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0200008351.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008351.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008351.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008351.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008351.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008351.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008351.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008351.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008351.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008351.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008351.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008351.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008351.hg.1 NM_144563.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008351.hg.1 NM_144563.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008351.hg.1 NM_144563.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008351.hg.1 NM_144563.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008351.hg.1 NM_144563.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008351.hg.1 NM_144563.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008351.hg.1 NM_144563.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008351.hg.1 NM_144563.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008351.hg.1 NM_144563.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008351.hg.1 NM_144563.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008351.hg.1 NM_144563.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008351.hg.1 NM_144563.2 Refseq
Affymetrix Human Clariom D Assay PSR0200158062.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Clariom D Assay PSR0200158062.hg.1 NM_144563.2 Refseq
Affymetrix Human Clariom D Assay PSR0200158062.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Clariom D Assay PSR0200158062.hg.1 NM_144563.2 Refseq
Affymetrix Human Clariom D Assay PSR0200158062.hg.1 TC0200008351.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0200008351.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Clariom D Assay TC0200008351.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Clariom D Assay TC0200008351.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Clariom D Assay TC0200008351.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Clariom D Assay TC0200008351.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Clariom D Assay TC0200008351.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Clariom D Assay TC0200008351.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Clariom D Assay TC0200008351.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Clariom D Assay TC0200008351.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Clariom D Assay TC0200008351.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Clariom D Assay TC0200008351.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Clariom D Assay TC0200008351.hg.1 ENST00000283646.4 Ensembl
Affymetrix Human Clariom D Assay TC0200008351.hg.1 NM_144563.2 Refseq
Affymetrix Human Clariom D Assay TC0200008351.hg.1 NM_144563.2 Refseq
Affymetrix Human Clariom D Assay TC0200008351.hg.1 NM_144563.2 Refseq
Affymetrix Human Clariom D Assay TC0200008351.hg.1 NM_144563.2 Refseq
Affymetrix Human Clariom D Assay TC0200008351.hg.1 NM_144563.2 Refseq
Affymetrix Human Clariom D Assay TC0200008351.hg.1 NM_144563.2 Refseq
Affymetrix Human Clariom D Assay TC0200008351.hg.1 NM_144563.2 Refseq
Affymetrix Human Clariom D Assay TC0200008351.hg.1 NM_144563.2 Refseq
Affymetrix Human Clariom D Assay TC0200008351.hg.1 NM_144563.2 Refseq
Affymetrix Human Clariom D Assay TC0200008351.hg.1 NM_144563.2 Refseq
Affymetrix Human Clariom D Assay TC0200008351.hg.1 NM_144563.2 Refseq
Affymetrix Human Clariom D Assay TC0200008351.hg.1 NM_144563.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline