This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01107657_m1
See other DDOST GE Assays ›
Gene Symbol:
DDOST
Gene Name:
dolichyl-diphosphooligosaccharide--protein glycosyltransferase non-catalytic subunit
Gene Aliases:
AGER1, CDG1R, OKSWcl45, OST, OST48, WBP1
Chromosome Location:
Chr.1: 20651767 - 20661544 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01107657_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DDOST
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005216.4 NP_005207.2 8 - 9 1137 61
GenBank mRNA AB062391.1 - 8 - 9 970 61
AK296041.1 - 7 - 8 903 61
AK297009.1 - 8 - 9 1010 61
AK315633.1 - 8 - 9 1117 61
BC002594.2 - 8 - 9 1044 3161588 61
D29643.1 - 8 - 9 1100 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100018421.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01002316.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene DDOST

Panther Classification:

Molecular Function -

glycosyltransferase

Gene Ontology Categories:

Function(s) Process(es)

protein glycosylation
protein N-linked glycosylation
protein N-linked glycosylation via asparagine
response to cytokine
T cell activation
oligosaccharyl transferase activity
dolichyl-diphosphooligosaccharide-protein glycotransferase activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR01035433.hg.1 TC01002316.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01035433.hg.1 ENST00000375048.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01035433.hg.1 ENST00000415136.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01035433.hg.1 NM_005216.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000375048.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000375048.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000375048.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000375048.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000375048.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000375048.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000375048.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000375048.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000375048.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000375048.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000375048.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000375048.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000375048.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000375048.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000415136.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000415136.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000415136.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000415136.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000415136.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000415136.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000415136.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000415136.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000415136.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000415136.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000415136.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000415136.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000415136.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000415136.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000415136.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000464364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000464364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000464364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000464364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000475210.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000475210.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000475210.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000477229.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000477229.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000477229.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000602624.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000602624.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000602624.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000602624.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000602624.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000602624.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000602624.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000602624.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000602624.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000602624.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000602624.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 ENST00000602624.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 NM_005216.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 NM_005216.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 NM_005216.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 NM_005216.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 NM_005216.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 NM_005216.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 NM_005216.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 NM_005216.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 NM_005216.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 NM_005216.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 NM_005216.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 NM_005216.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 NM_005216.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 NM_005216.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018421.hg.1 NM_005216.4 Refseq
Affymetrix Human Clariom D Assay PSR0100241644.hg.1 ENST00000375048.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100241644.hg.1 ENST00000415136.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100241644.hg.1 ENST00000475210.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100241644.hg.1 ENST00000602624.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100241644.hg.1 NM_005216.4 Refseq
Affymetrix Human Clariom D Assay PSR0100241644.hg.1 ENST00000375048.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100241644.hg.1 ENST00000415136.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100241644.hg.1 ENST00000475210.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100241644.hg.1 ENST00000602624.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100241644.hg.1 NM_005216.4 Refseq
Affymetrix Human Clariom D Assay PSR0100241644.hg.1 TC0100018421.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000375048.7 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000375048.7 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000375048.7 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000375048.7 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000375048.7 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000375048.7 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000375048.7 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000375048.7 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000375048.7 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000375048.7 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000375048.7 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000375048.7 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000375048.7 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000375048.7 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000415136.6 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000415136.6 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000415136.6 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000415136.6 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000415136.6 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000415136.6 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000415136.6 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000415136.6 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000415136.6 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000415136.6 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000415136.6 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000415136.6 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000415136.6 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000415136.6 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000415136.6 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000464364.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000464364.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000464364.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000464364.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000475210.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000475210.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000475210.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000477229.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000477229.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000477229.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000602624.6 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000602624.6 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000602624.6 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000602624.6 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000602624.6 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000602624.6 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000602624.6 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000602624.6 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000602624.6 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000602624.6 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000602624.6 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 ENST00000602624.6 Ensembl
Affymetrix Human Clariom D Assay TC0100018421.hg.1 NM_005216.4 Refseq
Affymetrix Human Clariom D Assay TC0100018421.hg.1 NM_005216.4 Refseq
Affymetrix Human Clariom D Assay TC0100018421.hg.1 NM_005216.4 Refseq
Affymetrix Human Clariom D Assay TC0100018421.hg.1 NM_005216.4 Refseq
Affymetrix Human Clariom D Assay TC0100018421.hg.1 NM_005216.4 Refseq
Affymetrix Human Clariom D Assay TC0100018421.hg.1 NM_005216.4 Refseq
Affymetrix Human Clariom D Assay TC0100018421.hg.1 NM_005216.4 Refseq
Affymetrix Human Clariom D Assay TC0100018421.hg.1 NM_005216.4 Refseq
Affymetrix Human Clariom D Assay TC0100018421.hg.1 NM_005216.4 Refseq
Affymetrix Human Clariom D Assay TC0100018421.hg.1 NM_005216.4 Refseq
Affymetrix Human Clariom D Assay TC0100018421.hg.1 NM_005216.4 Refseq
Affymetrix Human Clariom D Assay TC0100018421.hg.1 NM_005216.4 Refseq
Affymetrix Human Clariom D Assay TC0100018421.hg.1 NM_005216.4 Refseq
Affymetrix Human Clariom D Assay TC0100018421.hg.1 NM_005216.4 Refseq
Affymetrix Human Clariom D Assay TC0100018421.hg.1 NM_005216.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0