This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01107997_m1
See other TOM1L2 GE Assays ›
Gene Symbol:
TOM1L2
Gene Name:
target of myb1 like 2 membrane trafficking protein
Gene Aliases:
-
Chromosome Location:
Chr.17: 17843508 - 17972470 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01107997_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TOM1L2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001033551.2 NP_001028723.1 11 - 12 1290 79
NM_001082968.1 NP_001076437.1 12 - 13 1440 79
NM_001288787.1 NP_001275716.1 11 - 12 1305 79
GenBank mRNA AF467441.1 - 11 - 12 1243 79
AK057308.1 - 6 - 7 1046 79
AK124331.1 - 12 - 13 1368 79
AK301944.1 - 11 - 12 1232 79
BC051650.1 - 11 - 12 1230 5298847 79

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1700009954.hg.1
This assay targets the same location as HTA transcript cluster(s) TC17001194.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TOM1L2

Gene Ontology Categories:

Function(s) Process(es)

intracellular protein transport
signal transduction
negative regulation of mitotic nuclear division
protein binding
protein kinase binding
clathrin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR17016264.hg.1 ENST00000318094.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17016264.hg.1 ENST00000379504.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17016264.hg.1 ENST00000395739.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17016264.hg.1 ENST00000478943.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17016264.hg.1 ENST00000535933.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17016264.hg.1 ENST00000542206.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17016264.hg.1 NM_001033551.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR17016264.hg.1 NM_001082968.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR17016264.hg.1 TC17001194.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1700177851.hg.1 ENST00000318094.14 Ensembl
Affymetrix Human Clariom D Assay PSR1700177851.hg.1 ENST00000379504.7 Ensembl
Affymetrix Human Clariom D Assay PSR1700177851.hg.1 ENST00000395739.8 Ensembl
Affymetrix Human Clariom D Assay PSR1700177851.hg.1 ENST00000478943.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700177851.hg.1 ENST00000486413.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700177851.hg.1 ENST00000535933.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700177851.hg.1 ENST00000542206.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700177851.hg.1 ENST00000581396.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700177851.hg.1 NM_001033551.2 Refseq
Affymetrix Human Clariom D Assay PSR1700177851.hg.1 NM_001082968.1 Refseq
Affymetrix Human Clariom D Assay PSR1700177851.hg.1 NM_001288786.1 Refseq
Affymetrix Human Clariom D Assay PSR1700177851.hg.1 NM_001288787.1 Refseq
Affymetrix Human Clariom D Assay PSR1700177851.hg.1 NM_001288789.1 Refseq
Affymetrix Human Clariom D Assay PSR1700177851.hg.1 ENST00000318094.14 Ensembl
Affymetrix Human Clariom D Assay PSR1700177851.hg.1 ENST00000379504.7 Ensembl
Affymetrix Human Clariom D Assay PSR1700177851.hg.1 ENST00000395739.8 Ensembl
Affymetrix Human Clariom D Assay PSR1700177851.hg.1 ENST00000478943.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700177851.hg.1 ENST00000486413.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700177851.hg.1 ENST00000535933.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700177851.hg.1 ENST00000542206.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700177851.hg.1 ENST00000581396.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700177851.hg.1 NM_001033551.2 Refseq
Affymetrix Human Clariom D Assay PSR1700177851.hg.1 NM_001082968.1 Refseq
Affymetrix Human Clariom D Assay PSR1700177851.hg.1 NM_001288786.1 Refseq
Affymetrix Human Clariom D Assay PSR1700177851.hg.1 NM_001288787.1 Refseq
Affymetrix Human Clariom D Assay PSR1700177851.hg.1 NM_001288789.1 Refseq
Affymetrix Human Clariom D Assay PSR1700177851.hg.1 TC1700009954.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hwq2n:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0