This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01108550_g1
See other PPP4R3B GE Assays ›
Gene Symbol:
PPP4R3B
Gene Name:
protein phosphatase 4 regulatory subunit 3B
Gene Aliases:
FLFL2, PP4R3B, PSY2, SMEK2, smk1
Chromosome Location:
Chr.2: 55547292 - 55617757 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01108550_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPP4R3B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005264442.2 XP_005264499.1 14 - 15 2519 74
XM_005264445.2 XP_005264502.1 13 - 14 2360 74
XM_005264446.2 XP_005264503.1 12 - 13 2264 74
XM_017004531.1 XP_016860020.1 13 - 14 2423 74
XM_017004532.1 XP_016860021.1 13 - 14 1895 74
XM_017004534.1 XP_016860023.1 12 - 13 1799 74
XM_017004536.1 XP_016860025.1 11 - 12 1640 74
XM_017004540.1 XP_016860029.1 14 - 15 2526 74
GenBank mRNA AK023148.1 - 3 - 4 433 74
AK026437.1 - 9 - 10 1542 74

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

ATP synthase

Gene Ontology Categories:

Function(s)

protein dephosphorylation
regulation of lipid metabolic process
positive regulation of gluconeogenesis

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline