This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01108706_g1
See other CADM4 GE Assays ›
Gene Symbol:
CADM4
Gene Name:
cell adhesion molecule 4
Gene Aliases:
IGSF4C, NECL4, Necl-4, TSLL2, synCAM4
Chromosome Location:
Chr.19: 43622368 - 43641984 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01108706_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CADM4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_145296.1 NP_660339.1 5 - 6 706 70
XM_005258620.2 XP_005258677.1 5 - 6 688 70
XM_017026452.1 XP_016881941.1 5 - 6 615 70
GenBank mRNA AF363368.1 - 5 - 6 706 70
AK313028.1 - 5 - 6 688 70
BC068457.1 - 5 - 6 687 70
BC113963.1 - 2 - 3 279 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1900010855.hg.1
This assay targets the same location as HTA transcript cluster(s) TC19001592.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CADM4

Gene Ontology Categories:

Function(s) Process(es)

homophilic cell adhesion via plasma membrane adhesion molecules
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
cell recognition
receptor activity
receptor binding
protein homodimerization activity
cell adhesion molecule binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR19021805.hg.1 ENST00000222374.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19021805.hg.1 NM_145296.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19021805.hg.1 TC19001592.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1900010855.hg.1 ENST00000222374.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010855.hg.1 ENST00000222374.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010855.hg.1 ENST00000222374.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010855.hg.1 ENST00000222374.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010855.hg.1 ENST00000222374.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010855.hg.1 ENST00000222374.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010855.hg.1 ENST00000222374.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010855.hg.1 ENST00000222374.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010855.hg.1 ENST00000222374.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010855.hg.1 ENST00000222374.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010855.hg.1 ENST00000222374.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010855.hg.1 ENST00000222374.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010855.hg.1 ENST00000593506.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010855.hg.1 ENST00000593506.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010855.hg.1 ENST00000593506.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010855.hg.1 NM_145296.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010855.hg.1 NM_145296.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010855.hg.1 NM_145296.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010855.hg.1 NM_145296.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010855.hg.1 NM_145296.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010855.hg.1 NM_145296.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010855.hg.1 NM_145296.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010855.hg.1 NM_145296.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010855.hg.1 NM_145296.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010855.hg.1 NM_145296.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010855.hg.1 NM_145296.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010855.hg.1 NM_145296.1 Refseq
Affymetrix Human Clariom D Assay PSR1900190072.hg.1 ENST00000222374.2 Ensembl
Affymetrix Human Clariom D Assay PSR1900190072.hg.1 NM_145296.1 Refseq
Affymetrix Human Clariom D Assay PSR1900190072.hg.1 TC1900010855.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900190072.hg.1 ENST00000222374.2 Ensembl
Affymetrix Human Clariom D Assay PSR1900190072.hg.1 NM_145296.1 Refseq
Affymetrix Human Clariom D Assay TC1900010855.hg.1 ENST00000222374.2 Ensembl
Affymetrix Human Clariom D Assay TC1900010855.hg.1 ENST00000222374.2 Ensembl
Affymetrix Human Clariom D Assay TC1900010855.hg.1 ENST00000222374.2 Ensembl
Affymetrix Human Clariom D Assay TC1900010855.hg.1 ENST00000222374.2 Ensembl
Affymetrix Human Clariom D Assay TC1900010855.hg.1 ENST00000222374.2 Ensembl
Affymetrix Human Clariom D Assay TC1900010855.hg.1 ENST00000222374.2 Ensembl
Affymetrix Human Clariom D Assay TC1900010855.hg.1 ENST00000222374.2 Ensembl
Affymetrix Human Clariom D Assay TC1900010855.hg.1 ENST00000222374.2 Ensembl
Affymetrix Human Clariom D Assay TC1900010855.hg.1 ENST00000222374.2 Ensembl
Affymetrix Human Clariom D Assay TC1900010855.hg.1 ENST00000222374.2 Ensembl
Affymetrix Human Clariom D Assay TC1900010855.hg.1 ENST00000222374.2 Ensembl
Affymetrix Human Clariom D Assay TC1900010855.hg.1 ENST00000222374.2 Ensembl
Affymetrix Human Clariom D Assay TC1900010855.hg.1 ENST00000593506.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010855.hg.1 ENST00000593506.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010855.hg.1 ENST00000593506.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010855.hg.1 NM_145296.1 Refseq
Affymetrix Human Clariom D Assay TC1900010855.hg.1 NM_145296.1 Refseq
Affymetrix Human Clariom D Assay TC1900010855.hg.1 NM_145296.1 Refseq
Affymetrix Human Clariom D Assay TC1900010855.hg.1 NM_145296.1 Refseq
Affymetrix Human Clariom D Assay TC1900010855.hg.1 NM_145296.1 Refseq
Affymetrix Human Clariom D Assay TC1900010855.hg.1 NM_145296.1 Refseq
Affymetrix Human Clariom D Assay TC1900010855.hg.1 NM_145296.1 Refseq
Affymetrix Human Clariom D Assay TC1900010855.hg.1 NM_145296.1 Refseq
Affymetrix Human Clariom D Assay TC1900010855.hg.1 NM_145296.1 Refseq
Affymetrix Human Clariom D Assay TC1900010855.hg.1 NM_145296.1 Refseq
Affymetrix Human Clariom D Assay TC1900010855.hg.1 NM_145296.1 Refseq
Affymetrix Human Clariom D Assay TC1900010855.hg.1 NM_145296.1 Refseq
Affymetrix Human Clariom D Assay PSR1900190071.hg.1 ENST00000222374.2 Ensembl
Affymetrix Human Clariom D Assay PSR1900190071.hg.1 ENST00000593506.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900190071.hg.1 NM_145296.1 Refseq
Affymetrix Human Clariom D Assay PSR1900190071.hg.1 TC1900010855.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900190071.hg.1 ENST00000222374.2 Ensembl
Affymetrix Human Clariom D Assay PSR1900190071.hg.1 ENST00000593506.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900190071.hg.1 NM_145296.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0