This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01108978_m1
See other ASXL2 GE Assays ›
Gene Symbol:
ASXL2
Gene Name:
additional sex combs like 2, transcriptional regulator
Gene Aliases:
ASXH2
Chromosome Location:
Chr.2: 25733758 - 25878485 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01108978_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ASXL2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_018263.4 NP_060733.4 4 - 5 480 116
XM_011532950.2 XP_011531252.1 3 - 4 519 116
XM_011532951.2 XP_011531253.1 2 - 3 207 116
XM_017004429.1 XP_016859918.1 4 - 5 310 116
GenBank mRNA AB051472.2 - 4 - 5 480 116
AB084279.1 - 4 - 5 343 116
BX648218.1 - 4 - 5 505 116
CR933613.1 - 4 - 5 524 116

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200016645.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02001663.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ASXL2

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
positive regulation of peroxisome proliferator activated receptor signaling pathway
positive regulation of fat cell differentiation
positive regulation of transcription from RNA polymerase II promoter
DNA binding
peroxisome proliferator activated receptor binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR02026608.hg.1 TC02001663.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR02026608.hg.1 ENST00000272341.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02026608.hg.1 ENST00000336112.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02026608.hg.1 ENST00000435504.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02026608.hg.1 ENST00000497092.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02026608.hg.1 NM_018263.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02026613.hg.1 TC02001663.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR02026613.hg.1 ENST00000272341.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02026613.hg.1 ENST00000336112.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02026613.hg.1 ENST00000435504.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02026613.hg.1 NM_018263.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02026612.hg.1 TC02001663.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR02026612.hg.1 ENST00000272341.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02026612.hg.1 ENST00000336112.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02026612.hg.1 ENST00000435504.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02026612.hg.1 NM_018263.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000497092.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000497092.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000497092.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000497092.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000497092.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02026607.hg.1 TC02001663.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR02026607.hg.1 ENST00000272341.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02026607.hg.1 ENST00000336112.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02026607.hg.1 ENST00000435504.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02026607.hg.1 ENST00000497092.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02026607.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay PSR0200225939.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay PSR0200225939.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay PSR0200225939.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay PSR0200225939.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay PSR0200225939.hg.1 TC0200016645.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000497092.1 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000497092.1 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000497092.1 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000497092.1 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000497092.1 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay PSR0200225935.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay PSR0200225935.hg.1 ENST00000497092.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200225935.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay PSR0200225935.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay PSR0200225935.hg.1 ENST00000497092.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200225935.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay PSR0200225935.hg.1 TC0200016645.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0200225940.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay PSR0200225940.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay PSR0200225940.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay PSR0200225940.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay PSR0200225940.hg.1 TC0200016645.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0200225936.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay PSR0200225936.hg.1 ENST00000497092.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200225936.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay PSR0200225936.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay PSR0200225936.hg.1 ENST00000497092.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200225936.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay PSR0200225936.hg.1 TC0200016645.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rhr4g:80/100.66.78.163:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0