This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01109043_s1
See other ASXL2 GE Assays ›
Gene Symbol:
ASXL2
Gene Name:
additional sex combs like 2, transcriptional regulator
Gene Aliases:
ASXH2
Chromosome Location:
Chr.2: 25733758 - 25878485 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01109043_s1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ASXL2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_006712039.3 XP_006712102.1 9 11097 108
XM_006712040.2 XP_006712103.1 9 8383 108
XM_011532950.2 XP_011531252.1 12 9015 108
XM_011532951.2 XP_011531253.1 11 8703 108
XM_017004429.1 XP_016859918.1 13 8806 108
XM_017004430.1 XP_016859919.1 10 11163 108

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
positive regulation of peroxisome proliferator activated receptor signaling pathway
positive regulation of fat cell differentiation
positive regulation of transcription from RNA polymerase II promoter
DNA binding
peroxisome proliferator activated receptor binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000497092.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000497092.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000497092.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000497092.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 ENST00000497092.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000336112.8 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000404843.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000497092.1 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000497092.1 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000497092.1 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000497092.1 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 ENST00000497092.1 Ensembl
Affymetrix Human Clariom D Assay TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay TC0200016645.hg.1 NM_018263.5 Refseq
Affymetrix Human Clariom D Assay TC0200016645.hg.1 NM_018263.5 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0